2D–5D software for visualization, processing and analysis of microscopy images serving life sciences
Thermo Scientific Amira Software is a powerful, comprehensive, and versatile software solution for visualizing, analyzing and understanding life science and biomedical research images from many image modalities, including Optical and Electron Microscopy, CT, MRI and other imaging modalities. 
With incredible speed and flexibility, Amira Software supports advanced 2D–5D bioimaging workflows in research areas ranging from structural and cellular biology to tissue imaging, neuroscience, anatomical imaging and bioengineering. 
Amira Software harnesses the power of artificial intelligence to provide a broad spectrum of data visualization, processing, and analysis capabilities for 3D image data, including time series and multi-channel datasets. Its advanced features enable life science and biomedical researchers to extract valuable insights from their data, regardless of the modality or scale.
Imaging data
Amira Software workflow
Processing
Amira Software workflow
Visualization
Amira Software workflow
Analysis
Amira Software workflow
Customization
Confidence
Thanks to powerful segmentation and image processing capabilities and workflows, and after 20+ years of collaboration with the scientific community and thousands of researchers, it has been now proven that our digital imaging-based workflows provide reliable answers to biomedical and life science research.
Customization
Customization
Because your needs are unique and keep evolving, our software solutions are fully flexible and open to customization. Thanks to our scripting interface (Python, TCL), bridge with MATLAB, and our programming API, you can expand our software solution and integrate your own IP (Intellectual Property). And, if needed, our professional service team can help you design unique solutions tailored to your needs.
icon-improve_collaboration_64x64
Support
Thanks to our dedicated professional support team, you get access to our top experts to ensure that no question is left unanswered. And with our training, consulting options, and ever-expanding collection of tutorials and how-to’s, you can reduce your learning curve and focus on getting the answers you are pursuing.
Software_Automation_65x64
Automation
Thanks to our automation capabilities, and with the addition of our always expanding online repository of add-ons (Xtra Gallery), you can encapsulate repeatable workflows into easy-to-reproduce recipes. With the addition of artificial intelligence, analysis can be performed by non-image processing experts, allowing them to save time on complex analysis while ensuring results consistency.
Applications
Insight from your peers
Explore industry-leading insights and research that can support your lab workflow. Amira Software can empower your lab with a cutting-edge, comprehensive imaging data analysis toolbox.
Helping scientists answer questions that enable breakthrough discoveries in life sciences, materials science, and industry.
Amira Software Supports Mitochondria Research at Vanderbilt University
Read more
Correlative Microscopy: Using Amira Software to Understand the Spread of Cancer
Read more
Amira Software Accelerates Veterinary Research with 3D Visualization and Analysis
Read more
Molecular Clarity—Discovering What’s Possible with Cryo-Electron Tomography
Read more
Cryo-Electron Tomography and 3D Software Advances Coronavirus Research
Read more
Register now to get exclusive content delivered directly to your inbox.
Register
Techniques
Services
Amira-Avizo-Introductory-training_1160x600
Introductory training
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-advanced-training_1160x600
Advanced training
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-Custom-Dev_1160x600
Custom development
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.
Testimonials
Video Player is loading.
Current Time 0:00
Duration 2:26
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:26
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Dr. Antentor Hinton Jr, has made extensive use of Amira Software as part of his lab’s work in mitochondrial research.
    "Amira Software is crucial for visualizing SBF-SEM (serial block-face SEM) and FIB-SEM images, so we can analyze the 3D morphology of organelles."
    Dr. Antentor Hinton Jr
    Department of Molecular Physiology and Biophysics, Vanderbilt University
    Video Player is loading.
    Current Time 0:00
    Duration 5:10
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 5:10
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      VetCore, University of Veterinary Medicine uses Amira Software
      "Research at Vetmeduni Vienna is quite diverse, covering both clinical and preclinical research areas. We are currently four people working in the imaging facility, providing equipment and supports for researchers in both image acquisition (widefield microscopy, CLSM, spinning disk confocal microscopy, soon also STORM super-resolution microscopy, microscopic X-ray computed tomography) and in data visualization and analysis. Amira Software allows you to build networks, enabling complex interactions between data objects. Biology is also complex, and biological interactions are complex networks. Complex questions require complex tools to address them. In this respect, analysis networks such as used in Amira Software outperform most other software approaches when it comes to complex data analysis. Furthermore, I like that Amira Software is quite open concerning integration of deep learning networks and scripting (Tcl, Matlab, Python,...)."
      Stephan Handschuh, PhD
      VetCore, University of Veterinary Medicine, Vienna
      Video Player is loading.
      Current Time 0:00
      Duration 6:00
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 6:00
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        EMBL and DKFZ uses Amira Software
        The present testimonial highlights the way Amira Software supported Matthia A. Karreman, MSc, PhD at EMBL and DKFZ, Heidelberg, Germany, in her project on multimodal correlative microscopy to efficiently target single tumor cells in vivo. Read reference paper.
        Video: 6 min
        Features
        Style Sheet for Instrument Cards Original
        Products
        Thermo Scientific Amira 2D-5D visualization Software
        Amira Software
        Life Sciences
        • Explore 2D-5D bioimaging data
        • Identify and understand structures
        • Obtain statistical information
        • Share reports and stunning animations
        Thermo Scientific Krios G4 Cryo transmission electron microscope (TEM)
        Krios 5 Cryo-TEM
        • Fast, high-quality data collection
        • High workflow efficiency
        • Increased productivity and automation
        • Designed-in sustainability
        Thermo Scientific Aquilos 2 cryo focused ion beam (FIB)
        Aquilos 2 Cryo-FIB
        • Automation enables production of multiple lamellas
        • Target and extract your structure of interest with lift-out nano-manipulator
        • 3D visualization for high-resolution tomography
        Thermo Scientific Apreo 2 scanning electron microscope (SEM)
        Apreo 2 SEM
        • High-performance SEM for all-round nanometer or sub-nanometer resolution
        • In-column T1 backscatter detector for sensitive, TV-rate materials contrast
        • Excellent performance at long working distance (10 mm)