Compounding and extrusion instruments features
Materials mixing, compounding, and processing
Reliably scale-up and reduce time to market with the controlled, continuous processing of Thermo Scientific compounders and extruders. Our twin-screw extruders offer flexible configurations for small batches, ideally suited for pilot-scale production or low-volume manufacturing. Extruders are also a critical tool for research and development across the polymer, pharmaceutical, battery, and food sectors. 
Extruders designed to adapt to your compounding processes
The barrels and screws of Thermo Scientific compounders and extruders are specifically designed to maximize process flexibility.
A fully segmented screw design matches process requirements with a wide range of conveying, mixing, and extrusion elements.
The horizontal split barrel design has a removable top that can be opened for easy cleaning and process inspection. Multiple optional barrel ports can be utilized for feeding of liquids and solids. Program multiple temperature zones to adjust temperature along the process to fit any application.
Open horizontal split barrel showing segmented parallel screws
Larger extrusion free volume enhances productivity
Increased extruder output in a manufacturing environment can typically be achieved by increasing the specific torque [Nm/cm3] of the extruder (i.e., increasing motor power). Transferring this strategy to R&D and pilot labs would require screws with larger inner diameters, thus automatically reducing the free volume in the extruder. This free volume is much more important than high torque, especially in small extruders, as it would leave very little volume to process the material. To ensure all material can enter the extruder in these environments, our extruder design uses a high-free-volume approach (e.g., do/di = 1.73) with appropriate specific torque to generate the necessary throughput. 
Free volume diagram showing the space in a twin screw extruder that material travels through.
The comparatively small change from 1.50 to 1.73 (Process 16
Twin-Screw Extruder) shows a significant increase in available space between the twin-screws (green area) where the material must travel to be processed.
Easily scale up extrusion processes
Successful scale-up is only possible if materials on production-scale extruders undergo the same processes as on the smaller, lab-scale extruders. (i.e., similar screw geometries, process lengths, and specific energies.) Generally, this also means that the residence time of the material within the extruder would be the same. Thermo Scientific screw geometries are intentionally similar throughout our range of parallel twin-screw extruders. This helps to transfer lab processes into production, following a scientific approach and avoiding intermediate steps.
4 screw profiles of increasing size, showing the scalability of Thermo Scientific twin-screw extruders
Intuitive and convenient extruder operation
The functionality of our stand-alone extruders can easily be controlled via the integrated human-machine interface touchscreen. It allows for fast toggling between separate overview, setup, and parameter screens; attached feeders and downstream equipment are represented using clear graphics. One click opens a context menu for full control over all respective parameters. Consistent user guidance simplifies handling, lowers the learning curve, and minimizes operator errors.
 Screenshot of HMI showing extruder layout, with full control of all extrusion processes
Efficient material compounding
The Thermo Scientific Process 11 Parallel Twin-Screw Extruder helps you reduce the time and money spent formulating new polymer compounds. It uses 67% less material than a comparable 16 mm extruder while still delivering meaningful results in process and end-product optimization. Process conditions are reached quickly, allowing nearly twice as many experiments to be performed when compared to a standard production-scale (i.e., 24 mm) extruder. 
Process 11 Twin-screw Extruder
Modular torque rheometers for extrusion process and materials characterization
The Thermo Scientific HAAKE PolyLab OS System is a modular torque rheometer that combines measuring, mixing, and extrusion to meet your quality control and R&D needs. The drive unit can be connected to an interchangeable measuring sensor as well as a mixer, single-screw extruder, or conical/parallel twin-screw extruder.
This measurement platform lets you analyze material properties, simulate production processes, and perform capillary rheology to determine flow characteristics under real process conditions.
HAAKE PolyLab OS Modular Torque Rheometer System