Search Thermo Fisher Scientific
Fetal Bovine Serum (FBS) for Cell Culture |
Fetal bovine serum (FBS), also known as fetal calf serum, offers essential nutrients and growth factors for the maintenance and growth of cultured cells. Cell-based applications are an integral part of basic research, biochemical studies, vaccine development, and drug discovery to name a few. FBS is the most used serum in cell-based applications as it provides the most robust culture system for the widest range of cell types.
Since 1962, Gibco FBS products have been a global market leader. Through Thermo Fisher Scientific’s unwavering commitment to quality, scientists using Gibco FBS have been provided with consistent reliability, service, value, and innovation. Discover the performance differences that Gibco Fetal Bovine Serum can deliver for your research and production.
Learn more about the various market scenarios that drive FBS availability and price
Specifications, not origin, help drive consistent performance. We commissioned an independent research group to survey more than 500 researchers around the globe. This is what we learned:
Based on these findings, we transformed our Gibco FBS portfolio into categories focused on quality indicators rather than origin; Value FBS, Premium FBS, and Premium Plus FBS. Now you can easily select the best sera for your needs based on how they perform, not where they came from. This also enables us to minimize supply disruption since there is less reliance on origin.
Quality profile | Value FBS | Premium FBS | Premium Plus FBS |
---|---|---|---|
Endotoxin | ≤20 EU/mL | ≤10 EU/mL | ≤5 EU/mL |
Haemoglobin/hemoglobin | ≤25 mg/dL | ≤25 mg/dL | ≤20 mg/dL |
Sterility: bacterial and fungal testing | ✓ | ✓ | ✓ |
Osmolality | ✓ | ✓ | ✓ |
pH | ✓ | ✓ | ✓ |
Mycoplasma: supplemental testing (H-stain) | ✓ | ✓ | ✓ |
Mycoplasma | ✓ | ✓ | ✓ |
Country of origin | ✓ | ✓ | ✓ |
Oritain™ testing (origin confirmation) | – | ✓ | ✓ |
Biochemical and hormonal profile | |||
Biochemical: Alkaline phosphatase, ALT, AST, bicarbonate, bilirubin (total), BUN, BUN/creatinine ratio, calcium, chloride, cholesterol, creatinine, GGPT, glucose, HDL, iron, iron saturation, LDH, LDL, phosphorous (inorganic), potassium, sodium, TIBC, triglycerides (TG), uric acid | – | ✓ | ✓ |
Hormonal: Estradiol, insulin, progesterone, testosterone, thyroxine (T4) | – | ✓ | ✓ |
Virus testing | |||
Serology: BVDV serum neutralization titer (Ab) | – | ✓ | ✓ |
Serology: Anti-BVDV antibody test (EMA lots only) | – | ✓ | ✓ |
Virus (9CFR): Bluetongue Virus, Bovine Adenovirus, Bovine Parvovirus, Bovine Viral Diarrhoea Virus, Bovine Respiratory Syncytial Virus (BRSV), Rabies Virus, Reo Virus Cytopathogenic Agents - Including Bovine Herpesvirus 1 (BHV-1/IBR) Haemadsorbing Agents - Including Bovine Parainfluenza Virus 3 (PI-3) | ✓ | ✓ | ✓ |
Virus (EMA): Bluetongue Virus, Bovine Adenovirus, Bovine Parvovirus, Bovine Viral Diarrhoea Virus, Bovine Respiratory Syncytial Virus (BRSV), Rabies Virus, Reo Virus Cytopathogenic Agents- Including Bovine Herpesvirus 1 (BHV-1/IBR) Haemadsorbing Agents- Including Bovine Parainfluenza Virus 3 (PI-3) | – | ✓ | ✓ |
USDA safety testing: Bluetongue virus (Mexican and Australian origin) | ✓ (Mexican only) | ✓ | ✓ (Australian only) |
USDA safety testing: Akabane virus (Australian only) | – | ✓ | ✓ (Australian only) |
Protein electrophoresis | |||
Identification:Electrophoretic profile | ✓ | ✓ | ✓ |
Identification: Bovine gamma globulin (≤500 mg/L) | ✓ | ✓ | ✓ |
Protein: Albumin, alpha globulin, beta globulin, total protein | ✓ | ✓ | ✓ |
Performance testing | |||
Relative growth promotion (RGP), relative cloning efficiency (RCE), relative plating efficiency (RPE) | ✓ | ✓ | ✓ |
Documentation | |||
Certificate of Suitability (TSE CEP) | – | ✓ | ✓ |
Important to note that our FBS is sourced from BSE-negligible countries
Choose One Shot FBS in convenient 50 mL bottles to end aliquoting, stop the freeze-thaw cycle, and help minimize contamination risk.
Choose heat-inactivated FBS for immunological applications requiring inactivation of complement proteins.
Choose gamma-irradiated FBS when your research requires the highest level of biosecurity, ensuring safer and more consistent outcomes.
We provide a simplified three-tiered offering—Gibco Value FBS, Premium FBS, and Premium Plus FBS—where each category is clearly delineated by relevant performance markers and testing levels to help ensure you can confidently select the right serum for your research.
Designed for consistent growth for robust cell lines used within all standard cell culture research applications. Our Value FBS products offer excellent value with up to 30 quality specification tests.
Our most popular FBS product; high quality sera and exceptional value with up to 60 qualities specification tests
Our highest quality FBS for use with the most sensitive cells; up to 70 quality specification tests, including our lowest levels of endotoxin and hemoglobin release specifications
Designed for specific experimental needs and sensitive cell culture, including stem cell research, cancer research, reporter assays, immunoassays, antibodies, and more.
Quality profile | Value FBS | Premium FBS | Premium Plus FBS |
---|---|---|---|
Endotoxin | ≤20 EU/mL | ≤10 EU/mL | ≤5 EU/mL |
Haemoglobin/hemoglobin | ≤25 mg/dL | ≤25 mg/dL | ≤20 mg/dL |
Sterility: bacterial and fungal testing | ✓ | ✓ | ✓ |
Osmolality | ✓ | ✓ | ✓ |
pH | ✓ | ✓ | ✓ |
Mycoplasma: supplemental testing (H-stain) | ✓ | ✓ | ✓ |
Mycoplasma | ✓ | ✓ | ✓ |
Country of origin | ✓ | ✓ | ✓ |
Oritain™ testing (origin confirmation) | – | ✓ | ✓ |
Biochemical and hormonal profile | |||
Biochemical: Alkaline phosphatase, ALT, AST, bicarbonate, bilirubin (total), BUN, BUN/creatinine ratio, calcium, chloride, cholesterol, creatinine, GGPT, glucose, HDL, iron, iron saturation, LDH, LDL, phosphorous (inorganic), potassium, sodium, TIBC, triglycerides (TG), uric acid | – | ✓ | ✓ |
Hormonal: Estradiol, insulin, progesterone, testosterone, thyroxine (T4) | – | ✓ | ✓ |
Virus testing | |||
Serology: BVDV serum neutralization titer (Ab) | – | ✓ | ✓ |
Serology: Anti-BVDV antibody test (EMA lots only) | – | ✓ | ✓ |
Virus (9CFR): Bluetongue Virus, Bovine Adenovirus, Bovine Parvovirus, Bovine Viral Diarrhoea Virus, Bovine Respiratory Syncytial Virus (BRSV), Rabies Virus, Reo Virus Cytopathogenic Agents - Including Bovine Herpesvirus 1 (BHV-1/IBR) Haemadsorbing Agents - Including Bovine Parainfluenza Virus 3 (PI-3) | ✓ | ✓ | ✓ |
Virus (EMA): Bluetongue Virus, Bovine Adenovirus, Bovine Parvovirus, Bovine Viral Diarrhoea Virus, Bovine Respiratory Syncytial Virus (BRSV), Rabies Virus, Reo Virus Cytopathogenic Agents- Including Bovine Herpesvirus 1 (BHV-1/IBR) Haemadsorbing Agents- Including Bovine Parainfluenza Virus 3 (PI-3) | – | ✓ | ✓ |
USDA safety testing: Bluetongue virus (Mexican and Australian origin) | ✓ (Mexican only) | ✓ | ✓ (Australian only) |
USDA safety testing: Akabane virus (Australian only) | – | ✓ | ✓ (Australian only) |
Protein electrophoresis | |||
Identification:Electrophoretic profile | ✓ | ✓ | ✓ |
Identification: Bovine gamma globulin (≤500 mg/L) | ✓ | ✓ | ✓ |
Protein: Albumin, alpha globulin, beta globulin, total protein | ✓ | ✓ | ✓ |
Performance testing | |||
Relative growth promotion (RGP), relative cloning efficiency (RCE), relative plating efficiency (RPE) | ✓ | ✓ | ✓ |
Documentation | |||
Certificate of Suitability (TSE CEP) | – | ✓ | ✓ |
Important to note that our FBS is sourced from BSE-negligible countries
Watch these videos to learn how FBS performance is based on science, not origin. Explore customer stories, FBS FAQs, how-to explanations, and product innovations.
Choosing the Right FBS: Why Science, Not Geography, Drives Performance and Reproducibility in Cell Culture Research.
This video will show you how to streamline your lab work by removing the need to test your FBS saving you time and money.
Join Bay Vagher, a graduate student at the University of Vermont, shares her enthusiasm for the Gibco iMATCH Sera Lot Matching Tool.
"Learn more and see why researchers worldwide recommend Gibco sera more than any other sera at thermofisher.com/fbs This video explains what specifications researchers consider when selecting their FBS. We surveyed over 500 cell culture researchers
If you’re one of the more than 80% of researchers who aliquots their FBS into 50 mL conical tubes, your life is about to change for the better. Pour and pipette with the greatest of ease.
Why do FBS prices fluctuate constantly
Learn more at thermofisher.com/oneshot Hear what Dr. Christian is saying about the Gibco One Shot FBS 50 mL bottle. Saving time in the lab and reducing the risk of contamination with this aliquot-free solution is a great thing.
Learn more at thermofisher.com/oneshot Our researchers discuss how a unique shaped bottle can help reduce plastic waste in lab. Not only that, it will remove the need to aliquot your serum.
Learn more at thermofisher.com/oneshot Two researchers from neighboring cell culture labs discuss the frustation of aliquoting many bottles of serum into 50 mL conical tubes.
Learn more at thermofisher.com/oneshot. Let our scientist show you how to properly aliquot your FBS into 50 mL conical tubes as well as heat inactivate the FBS to ensure an optimal outcome.
The benefits of using Gibco® FBS for your cell culture research. Click here to learn more: lifetechnologies.com/fbs
Learn more and see other SerumSmart videos at thermofisher.com/gibco. This video gives you an understanding of what to expect from our SerumSmart video series. We will cover all sorts of topics like myths and misconceptions of FBS/serum
In this episode of Serum Smart, learn about Grand Island, NY and it's important history, and impact to cell culture research.
Explanation of dynamics in the FBS industry and how it's impacted by the meat-packing industry. Sera products are a byproduct of this industry, the number of cows being processed will determine the serum supply to researchers.
In a world full of compromise, we refuse to cut corners. Our customers demand the best, so we take extraordinary measures in the collection of FBS. Find out what makes Gibco FBS the most trusted in the world.
When others cut corners, we refuse to be ordinary. We take steps in the processing of our FBS to help ensure our customers the highest quality product. Witness these extraordinary steps that enable extraordinary breakthroughs.
A new era of quality is upon us. Unmatched innovation, reliability and traceability have arrived. See how we provide researchers the transparency and confidence they deserve from their FBS supplier.
Fetal bovine serum (FBS) is a byproduct of harvesting cattle for the meatpacking industry–it offers essential growth factors for the maintenance and growth of cultured cells. FBS is used as a supplement to basal growth medium in cell culture applications.
Since its launch over 60 years ago, Gibco FBS has been a part of historical scientific breakthroughs, including cell line development, cloning, and gene editing. Gibco sera are the most cited in global scientific journals and are used and trusted by many top pharma companies. Furthermore, Gibco FBS is origin guaranteed through fingerprinting technology and is ISIA traceability certified.
Learn more about FBS fingerprinting and ISIA traceability certification
Extraordinary measures are taken during the manufacturing and production of Gibco FBS, with careful consideration taken in each step of the journey from farm to bench.
Did you know there are lower-cost alternatives to fetal bovine serum? These products include bovine serum, horse serum, newborn calf serum, goat serum, rabbit serum, porcine serum, and chicken serum.
Gibco sera for bioprocessing can support your cell culture needs as you scale up production. As one of the largest serum suppliers from BSE-negligible risk regions, Gibco serum can help meet biopharmaceutical manufacturing requirements.
Sera sourced from BSE-free regions for preclinical, industrial, and academic research applications requiring low risk.
Sera specifically qualified for specialty research & specific assays, including stem cell research, immunoassays, & antibody production.
Learn more about how we’ve become a world leader for sera, media, and reagents.
Discover the important factors to consider in FBS selection and learn how scientists are utilizing the benefits of FBS in their research.
We have dedicated regional sales representatives to assist you in all things FBS. Connect directly with a FBS representative today!
Have questions about using FBS in your cell culture experiments? Find the answers you need in this extensive FAQ database.
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Fetal Bovine Serum Basics
Learn the basics of FBS for cell culture, including information on the FBS uses, components, and the market dynamics driving this industry.
Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Find your regional sales representative
Connect with a regional FBS sales representative.
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support?
Contact our expert team for technical and application support of laboratory products.
For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.