The Wohl–Ziegler reaction, or the Wohl–Ziegler bromination, is a type of the free radical substitution
Wohl-Ziegler reaction overview
The Wohl–Ziegler reaction is a chemical reaction that involves the allylic or benzylic bromination of hydrocarbons using an N-bromosuccinimide and a radical initiator.
Named Reactions in Organic Synthesis
Wohl-Ziegler Bromination Mechanism
Watch our video to learn more about the Wohl-Ziegler bromination and its mechanisms.
Video Player is loading.
Current Time 0:00
Duration 3:02
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:02
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Wohl-Ziegler Bromination?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Wohl-Ziegler Bromination. 
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Wohl-Ziegler Bromination
    In 1919, German chemist Alfred Wohl investigated the reaction between 2,3-dimethyl-2-butene and N-bromoacetamide in diethyl ether, and discovered that the double bond of the substrate remained unchanged and one of the methyl groups was replaced by bromine. This discovery was interesting, as the reaction was previously thought to require the use of bromine at high temperatures to react with the alkenes. In 1942, Karl Ziegler carried out a comprehensive study of the utility of N-bromosuccinimide (NBS) in the allylic bromination of olefins and demonstrated the synthetic value of the process. Subsequently, the addition of bromine at the allylic position of olefins or at the benzylic position of alkylated aromatic or heteroaromatic compounds became known as the Wohl-Ziegler bromination.
    The Wohl-Ziegler bromination has been utilized in several total synthesis campaigns, including James M. Cook’s preparation of (-)-tryprostatin A, a natural product isolated from a marine fungus that inhibits cell cycle progression.
    Other Free Radical Reactions
    Other Free Radical Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 4:00