Oxidations using Dess-Martin periodinane (DMP) are now known as Dess-Martin oxidations
Dess-Martin Oxidation
DMP, a hypervalent iodine compound, is the reagent of choice for the selective and very mild oxidation of primary alcohols to aldehydes and secondary alcohols to ketones.
Named Reactions in Organic Synthesis
Dess-Martin Oxidation Mechanism
Watch our video to learn more about the Dess-Martin Oxidation and its mechanisms.
Video Player is loading.
Current Time 0:00
Duration 2:54
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:54
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Dess-Martin Oxidation?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Dess-Martin Oxidation. 
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Dess-Martin Oxidation
    During the 1980s, hypervalent iodine reagents were developed as selective, mild, and environmentally friendly oxidizing agents for organic synthesis. Perhaps the most important group of these reagents are the periodinanes (derivatives of pentacoordinate iodine (V)), and the most well-known of these include the reagents 2-iodoxybenzoic acid (IBX) and Dess-Martin periodinane (DMP). While IBX had been known since 1893, its insolubility in most organic solvents inhibited its use in organic synthesis. However, in 1983 D.B. Dess and J.C. Martin described the preparation of DMP, a far more soluble alternative. Since this discovery, DMP has become the reagent of choice for the oxidation of alcohols to their corresponding carbonyl compounds, and oxidations using DMP are now known as Dess-Martin oxidations.
    In the total synthesis of ustiloxin D, a highly potent inhibitor of microtubule assembly, M.M. Joullié and co-workers utilized DMP to convert a macrocyclic primary alcohol into its corresponding aldehyde.
    Other Oxidation Reactions
    Other Oxidation Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 4.09%
    Stream Type LIVE
    Remaining Time 4:00