Rapidly detect Mycoplasma with confidence
Applied Biosystems MycoSEQ Mycoplasma Detection Assays help deliver highly accurate and sensitive results that meet or exceed regulatory guidance for Mycoplasma detection while facilitating same-day actionable results to enable early detection of Mycoplasma contamination.
  • Proven—Regulatory-accepted method for Mycoplasma detection, with Drug Master File (DMF), filed with the United States Food and Drug Administration (FDA) and Health Canada
  • Sensitive—Meets or exceeds the recommended regulatory guidance of 10 CFU/mL or 10 genome copies (GC)/mL for a wide variety of Mycoplasma species, including those commonly known to contaminate bioprocessing cultures
  • Specific—Validated in numerous independent studies* to specifically detect a wide variety of Mycoplasma, acholeplasma, and spiroplasma species with no known cross-reactivity with closely related bacterial species
  • Fast—qPCR-based workflow helps deliver actionable results in less than 5 hours, enabling quick detection of in-process, raw materials, or harvest contamination—helping expedite the release of your product
  • Accurate—Discriminatory positive control allows discrimination between true Mycoplasma and accidental contamination of test sample with positive control
        *MycoSEQ Detection Assay
MycoSEQ kits include:
PrepSEQ Sample prep
Optional
TaqMan or SYBR Green Assay
TaqMan or SYBR Green Master Mix
Discriminatory Positive Control
Negative
control
DNA dilution buffer
Featured solution: MycoSEQ Plus Mycoplasma Detection Kit
MycoSEQ Plus Mycoplasma Detection Kit
The Applied Biosystems MycoSEQ Plus Mycoplasma Detection Kit is a probe-based quantitative PCR (qPCR) assay kit featuring trusted TaqMan chemistry. Developed specifically for the detection of Mycoplasma in cell therapy and complex bioproduction samples, TaqMan-based assays help deliver performance that meets or exceeds the regulatory requirements of 10 CFU/mL, the high level of performance needed to facilitate product lot release testing, including the ability to help meet regulatory guidance for assay sensitivity and specificity.
  • Multiplexed assay enables simultaneous screening of over 200 species of Mycoplasma, with no demonstrated cross-reactivity to closely related bacterial species
  • Demonstrated to meet or exceed LOD less than/equal to 10 GC/mL for all Mycoplasma species listed in Pharmacopoeia (USP, EP, JP), as well as representative hemoplasma, mesoplasma, and ureaplasma
  • Unique discriminatory positive control that can differentiate a control-based contamination event from a true positive sample call
  • Flexible and scalable throughput supports fluctuating testing needs and enables reduced cost per reaction by batch testing multiple samples
  • Limits risk to production facilities as it does not require live Mycoplasma for testing or process validation
  • Internal positive control supports PCR reaction consistency across all samples
MycoSEQ Mycoplasma Detection Kit
MycoSEQ Mycoplasma Detection Assays Kit
The tried-and-true Applied Biosystems MycoSEQ Mycoplasma Detection Kit set the standard for rapid Mycoplasma testing and has been validated in more than 44 therapeutics across a variety of modalities.
This dye-based assay design utilizes a proprietary bioinformatics pipeline to detect >90 Mycoplasma species with high specificity and limited cross-reactivity with closely related bacterial species. Its sensitivity has been demonstrated by internal and external validation methods to detect 10 CFU/mL, or the genomic equivalent of 10 GC/mL, as recommended by regulatory guidance.
Mycoplasma testing in monoclonal antibody production bioreactor growth and bulk harvest lot release
Learn about implementing the MycoSEQ Plus Mycoplasma detection system throughout various stages of a CHO cell–based bioproduction process in this application note.
Compare MycoSEQ Assays
MycoSEQ Plus Kit
MycoSEQ Kit
Sample type
Bioproduction media, cells
Bioproduction media, cells
Therapeutic type
Cell therapies, gene therapies, viral vector-based therapies, biosimilars
Cell therapies, gene therapies, viral vector-based therapies, biosimilars
Detection chemistry
TaqMan (probe-based)
SYBR (dye-based)
Number of reactions per kit
100
100
Hours per workflow
≤5 hours
≤5 hours
Discriminatory positive control
Yes
Yes
Internal positive control
Yes
No
Performance capable of meeting lot release requirements
Yes
Yes
Number of species
200+
90+
On-demand webinars
On-demand webinar: Detecting contaminants, ensuring safety – high-performance Mycoplasma testing solution
On-demand webinar: A journey to Cell Therapy manufacturing—from R&D to lot-release
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
Shop MycoSEQ solutions
Additional bioprocessing resources
As a trusted partner, we are committed to supporting your journey toward innovative biotech solutions.
Accelerate your therapeutic monoclonal antibody development and manufacturing program.                             
Learn, evaluate, connect.   
Connect with a Bioprocessing representative or request more info.                                                               