NGS Library Preparation Kits for Illumina Systems
The Invitrogen Collibri portfolio of library prep kits and accessories generate libraries more quickly, with less bias, in order to match the speed and capacity of Illumina instruments. Advantages available only from Invitrogen Collibri Library Prep Kits:
Research SARS-CoV-2 mutations and gene expression profiles
Collibri Library Prep kits for Illumina Systems enhance NGS-based viral surveillance with higher quality sequences and rapid protocols.
  • Identify emerging strains
  • Rapidly generate gene expression profiles
  • Research target sequences for vaccine or therapeutic development
Identify the best technology for your research
Review optimized protocol to sequence SARS-CoV-2
Visual feedback at each step in the protocol indicates proper addition and mixing of reagents
Video: Improve success rates of library generation using visual feedback
Video Player is loading.
Current Time 0:00
Duration 1:10
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:10
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Collibri PS DNA Library Prep Kit for Illumina systems
    Figure 1. An example of the Collibri PS DNA Library Prep Kit for Illumina systems workflow in which each reagent contains a unique tracking dye that provides visual feedback of proper reagent addition and mixing.
    Shorter total workflow times, greater support
    The Collibri library prep workflows match the pace needed to efficiently utilize Illumina systems. Reduce time spent on placing orders, managing inventory and contacting technical support from multiple companies. Thermo Fisher Scientific offers technical and ordering support for most steps in the total pre-sequencing workflow for improved efficiency of your lab operations.
    Whole-genome sequencing PCR-free library prep workflow
    RNA-seq library prep workflow
    Visual feedback is provided by the entire Collibri portfolio of library prep kits and accessories
    Whole-genome sequencing
    Reduce bias in genomic interpretation
    RNA sequencing
    Clean data reveals transcriptome complexity
    Library quantification
    Determine the concentration of your Illumina NGS libraries
    Library amplification
    Amplify libraries for sequencing on Illumina systems
    Technical Note: Understanding library quantification assays for next-generation sequencing applications
    NGS libraries must be carefully quantified and normalized so that each sample achieves the required number of reads.
    Download
    For Research Use Only. Not for use in diagnostic procedures.