Integrative structural biology
Integrative structural biology is when two analytical techniques are combined to achieve a complete and accurate determination of the 3D protein or protein complex structure. The precise and accurate characterization of protein structure and protein complexes is essential for understanding protein function and the mechanisms of action in a biological system. Solving the structure of large dynamic complexes often requires integrating several complementary techniques, such as mass spectrometry (MS) and cryo-electron microscopy (cryo-EM)—an approach known as integrative structural biology.
Cryo-EM makes it possible to reproducibly produce near-atomic resolution of proteins in all their complex conformations, structures, and modified forms. This has made cryo-EM the go-to technique for scientists around the world, generating breakthroughs in research for infectious disease, neurodegenerative disease, and cancer, among others.
MS can provide complementary information to cryo-EM by guiding downstream structure determination. For example, information on sample purity, stability and sample homogeneity can be ascertained to make intelligent decision about cryo-EM structural determination. Additionally, MS techniques can provide rich constraints to support integrative structural models that deliver greater resolution and accuracy. Adding MS data to cryo-EM techniques can provide the boost needed to generate a complete and accurate 3D structural model.
OptiMSe: An automated sample screening workflow
Learn about the Thermo ScientificOptiMSe workflow. A fast, automated, and intuitive way to to eliminate unpromising samples for vitrification and cryo-EM.
Learn more ›
Workflows
Integrative structural biology workflows
There are a growing number of innovative ways researchers are combining techniques to better understand the proteome. Explore pioneering integrative structural biology workflows below, and the available tools to achieve each step of the workflow. 
Fast sample quality screening of cryo-EM samples with automated native MS
Sample integrity, purity, and homogeneity are critical for achieving high-resolution structures using cryo-EM. Rapid and reliable screening methods for assessing sample quality are essential. Native mass spectrometry (nMS) enables direct mass measurement of macromolecular assemblies by maintaining their near-native structures and assembly states upon gas phase transfer from solution. nMS is a powerful diagnostic and a screening platform that can be used for rapid identification of whether the correct protein and/or nucleic acid components as well as bound cofactors and ligands are correctly assembled. With high resolving power, nMS can reveal protein heterogeneity arising from post-translational modifications such as glycosylation and phosphorylation as well as spot samples with contaminations or degradation.
Workflow stepSummaryAvailable tools
Sample preparation
A variety of traditional sample preparation techniques can be used to achieve sample integrity required for cryo-EM. Complete end-to-end solutions are available for even the most challenging protein. 
Protein expression
Protein sample preparation
VitroEase Buffer Screening Kit
Native mass spectrometry 
Protein structures are kept intact and introduced into the mass spectrometer in biologically relevant conditions. Specifically designed software is used to obtain information on the intact protein or protein complex, including subunit stoichiometry, subunit identification, biomolecule binding, protein complex topology and protein dynamics can also be ascertained. 
Q Exactive UHMR Hybrid Quadrupole-Orbitrap MS
NativePac OBE-1 SEC column
Orbitrap Ascend Tribrid Mass Spectrometry
Identification of high-quality samples
Data analysis can be performed with Thermo Scientific BioPharma Finder software. This software produces highly accurate results, even for low-abundance proteins, and enables detection of extremely small protein modifications.  
Biopharma Finder Software
Single particle cryo-EM
Data collection consists of high-resolution imaging with a cryo-TEM. With advances in data collection software, individual particles can be automatically identified in the TEM image and grouped according to particle orientation. For every sample, robust, reliable automation simplifies and accelerates imaging and identification.   
Tundra Cryo TEM- 100 kV
Glacios Cryo TEM- 200 kV
Krios Cryo TEM- 300 kV
Structure visualization 
Once sufficient particle data is collected the data can be recombined into a 3D representation of the protein or protein complex. This uses 2D data from tens of thousands of particles and typically involves multiple data processing steps. A number of professionally developed and open-source data processing solutions exist to simplify and expedite this process.
Smart EPU Software with embedded CryoSparc Live
High fidelity structural models from in cell cryo-electron tomography and crosslinking mass spectrometry
Combining chemical crosslinking mass spectrometry (XL-MS) with cryo-ET can help improve structural resolution of proteins in their native state. XL-MS in combination with cryo-ET can provide confirmation of cryo-EM structural data by mapping crosslinks on proposed structures; fitting/positioning of subunits with the help of XL-derived restraints; modelling of missing regions invisible to EM due to their flexibility. More generally, combination of cryo-EM and XL-MS data with other experimentally or computationally derived information can provide a more complete integrative/hybrid model.
Workflow stepOverviewAvailable tools
Cell culture
Cells prepared by routine culture methods are grown on carbon-coated gold electron microscopy (EM) grids for cryo-TEM analysis.
Cell culture media
Cryo-FIB with correlative light microscopy
Using cryo-correlative microscopy the structures of interest are identified. A dedicated cryo-FLM stage keeps the sample in its vitrified state during cryofluorescence imaging. All Thermo Scientific cryo-FIBs and cryo-PFIBs can be equipped with an iFLM Correlative System, allowing samples to be imaged directly within the high vacuum without additional transfer steps from an external cryo-light microscope. The dedicated cryo-FIB prepares a thin, uniform lamella at the vitreous temperature.
Aquilos 2 Cryo-FIB
Arctis Cryo-PFIB
Helios Hydra 5 DualBeam
Imaging by cryo TEM
During cryo-ET, the sample is tilted in known increments about an axis. The individual projection images from the tomographic tilt series are then combined computationally in a procedure known as back projection, which creates the 3D tomographic volume.  
Glacios Cryo TEM- 200 kV
Krios Cryo TEM- 300 kV
Reconstruction 
The 3D tomogram featuring cellular structures can be segmented and colored in a variety of ways to enhance its display and presentation. From the tomogram small subsets of data containing the structures of interest can be computationally extracted and subjected to image processing methods.   
Amira Software
Protein crosslinking
Crosslinking reagents are used to covalently link interacting proteins that are in close proximity. If the crosslinking is done at the protein level, the samples are digested to peptides with an appropriate enzyme. An enrichment step is incorporated upon digestion to isolate crosslinked peptides.
DSSO (disuccinimidyl sulfoxide)
DSBU (Disuccinimidyl Dibutyric Urea)
DSPP  (Disuccinimidyl Phenyl Phosphonic Acid, PhoX)
Crosslinking mass spectrometry
The samples are separated and introduced into the mass spectrometer for analysis. Proteome Discoverer software with XlinkX node is used for data interpretation. This workflow enables the identification of interacting regions thereby enabling creation of protein-protein interaction maps.
Orbitrap Ascend Tribrid Mass Spectrometer
Proteome Discoverer Software with XlinkX node
Insights into structural dynamics and function with hydrogen-deuterium exchange mass spectrometry and cryo-EM 
Cryo-EM captures a snapshot of a biological process frozen in time, when in fact biological processes are highly dynamic. By combining structural cryo-em data with MS data, additional information on protein dynamics can be identified. For example, for protein complexes that can be studied ex vivo or ex situ, hydrogen-deuterium exchange-MS (HDX-MS) is a powerful technique that can provide information on protein-protein or protein-ligand interaction sites and conformational changes induced by PTMs.
Workflow stepOverviewAvailable tools
Sample preparation
A variety of traditional sample preparation techniques can be used to achieve sample integrity required for cryo-EM. Complete end-to-end solutions are available for even the most challenging protein. 
Protein expression
Protein sample preparation
Single particle cryo-EM
With advances in data collection software, individual particles can be automatically identified in the TEM image and grouped according to particle orientation. Near-atomic structural determination of single particles can be achieved with Thermo Scientific Cryo-TEMs. Once sufficient particle data is collected the data can be recombined into a 3D representation of the protein or protein complex. A number of professionally developed and open-source data processing solutions exist to simplify and expedite this process.   
Tundra Cryo TEM- 100 kV
Glacios Cryo TEM- 200 kV
Krios Cryo TEM- 300 Kv
Hydrogen deuterium exchange mass spectrometry (HDX-MS)
HDX-MS can be used to obtain protein structure and conformation information. Additionally, for protein complexes, HDX-MS can provide information on protein-protein or protein-ligand interaction sites and conformational changes induced by PTMs.
Orbitrap Exploris 240 or 480 MS
Orbitrap Ascend Tribrid Mass Spectrometer
Glycoproteomics
Mass spectrometry based glycoproteomics can be used to ascertain information such as glycosylation sites and types and number of glycoforms that are present.   
Orbitrap Ascend Tribrid Mass Spectrometer
Native MS
Provides information on proteins or protein complexes in their biological state such as mass, subunit stoichiometry, protein complex topology, and dynamics.
eBook: Integrative Structural Biology: High Precision 3D Analysis from Structure to Function
The dynamic and flexible nature of biomolecules poses a major challenge for structural biologists. Thus, the full characterization of a macromolecular system requires the combination of many structure-determination techniques, an approach known as integrative structural biology. In this context, advances in MS techniques in combination with cryo-EM and other structural tools is revolutionizing the understanding of protein structure, function and dynamics. Download this eBook to learn more about: Tools for integrative structural biology. The benefits of combining cryo-EM, cryo-ET and mass spectrometry. The advantages integrated modeling provides.
 Download eBook ›
Using integrative biophysical approaches to understand GPCR regulation by β-arrestins
Over the past five years, the work of Dr. John Janetzko and colleagues in Dr. Brian Kobilka’s lab have provided considerable advances in understanding how β-arrestins recognize GPCRs and how these complexes are regulated inside cells. Using cryo-EM, they obtained one of the first structures of a GPCR-β-arrestin complex. This structure showed a bound membrane phosphoinositide, suggesting a possible mode of regulation for these complexes. Using cell-based and in vitro biochemical and biophysical approaches, including hydrogen-deuterium exchange (HDX) MS, they showed that membrane phosphoinositides act as allosteric modulators of GPCR-β-arrestin complexes, gating their assembly and disassembly. Together these studies provide high-resolution molecular details into the mechanism by which these proteins of critical importance for cellular physiology function.
Video Player is loading.
Current Time 0:00
Duration 40:03
Loaded: 0.41%
Stream Type LIVE
Remaining Time 40:03
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Study small viral glycoproteins using an integrative structural biology approach
    leva Drulyte, Thermo Fisher Scientific
    Joost Snijder, Utrecht University
    Video Player is loading.
    Current Time 0:00
    Duration 54:58
    Loaded: 0.30%
    Stream Type LIVE
    Remaining Time 54:58
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Native mass spectrometry-based screening for optimal sample preparation in single particle cryo-EM
      Dom Olinares, Rockefeller University
      Video Player is loading.
      Current Time 0:00
      Duration 10:59
      Loaded: 1.49%
      Stream Type LIVE
      Remaining Time 10:59
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Integrative Structural Biology Going Viral
        Albert Konijnenberg, Thermo Fisher Scientific
        Video Player is loading.
        Current Time 0:00
        Duration 1:01:24
        Loaded: 0.27%
        Stream Type LIVE
        Remaining Time 1:01:24
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Mass spectrometry for 3D structural determination with cryo-electron microscopy
          Christian Dienemann, Max Planck Institute
          Henning Urlaub, Max Planck Institute
          Register to watch
          Orbitrap Exploris 480 Mass Spectrometer
          Orbitrap Ascend Tribrid Mass Spectrometer
          Q Exactive UHMR Hybrid Quadrupole-Orbitrap MS System
           Orbitrap Exploris 480 Mass SpectrometerOrbitrap Ascend Tribrid Mass SpectrometerQ Exactive UHMR Hybrid Quadrupole-Orbitrap MS System
          Advantages
          • A compact footprint as space is always a premium regardless of lab. 
          • Superior ion transmission, HRAM and FAIMS Pro option. Ideal for examining  large proteins, protein complexes, PTM under biological conditions, 
          • An analytical framework that meets the needs of most structural research. 
          • Most versatile mass spectrometer for any structural biology experiment. 
          • High resolution accurate mass (HRAM), multiple fragmentation, MSn capability and FAIMS Pro option. 
          • Provides insights on the most complex molecules and biological systems, from whole proteome profiling, protein complexes to individual protein structural characterization.
          • Ideal mass spectrometer for native intact mass, top-down analyses and screening sample quality for cryo-EM. 
          • Enables the investigation of heterogeneity of protein complexes. HRAM, multiple fragmentations option, and direct mass technology capability.
          Resources
           Download Orbitrap Exploris 480 MS spec sheet ›
           Orbitrap Ascend Tribrid MS spec sheet ›
           Download Q Exactive UHMR MS spec sheet ›
          Resolving power480,000 at m/z 195 7,500-480,000 FWHM at m/z 200Up to 240,000 at m/z 200 
          Scan speedUp to 40HzOrbitrap mass analyzer MSⁿ up to 45 Hz
          Ion trap MSⁿ up to 50 Hz
          18Hz
          Mass range40 to 6,000 m/z (up to 8,000 m/z with the BioPharma option) Standard mass range m/z 40–2000, mid-mass range m/z 200–6000,
          and optional HMRn+ mass range m/z 500–16,000
          50 to 8,000 m/z
          Dynamic range>5000:1>5,000 within a single MS acquisition>5000:1
          Mass accuracyInternal <1 ppm RMS; External: <3 ppm RMSInternal <1 ppm RMS; External: <3 ppm RMSInternal <1 ppm RMS; External: <3 ppm RMS
          Cryo transmission electron microscopes for structural biology
          The Thermo Scientific Cryo TEMs are revolutionizing life science research through innovation and accessibility. By combining automation, artificial intelligence, and an improved user experience, Thermo Scientific Cryo-TEMs allows you to harness the power of single particle analysis, MicroED and Cryo-ET at resolutions that are accelerating our breakthroughs in structural biology.
          Tundra Cryo-TEM
          Accessible & Smart
          • Easy, iterative loading and imaging for rapid sample-viability determination 
          • AI-guided automation with results displayed progressively 
          • Cost effective and space efficient
           Download Tundra Cryo TEM datasheet ›
          Intermediate-resolution SPA100 kV, <3.5 Å*
          Medium throughputDataset in 24 hours
          Sample typeProteins
          ApplicationsSPA
          Glacios 2 Cryo-TEM
          Capable & Versatile
          • Maximized ease-of-use and excellent performance to offer a complete package for introducing cryo-TEM into your research
          • Compact hardware footprint (minimizes installation requirements) at an affordable price
           Download Glacios Cryo TEM datasheet ›
          High-resolution SPA200 kV, <2.5 Å*
          High throughputDataset in 30 minutes
          Sample typeProteins, crystals, cells
          ApplicationsSPA, Micro-ED, tomography
          Krios 5 Cryo-TEM
          Powerful & Productive
          • Ultimate productivity with fast, high-quality data collection
          • High workflow efficiency for cryo-tomography and single particle analysis
          • Built-in sustainability with energy-saving Eco modes and fast "wake up" scheduling
           Download Krios 5 Cryo-TEM datasheet ›
          Ultra-high resolution SPA300 kV, <2 Å*
          Highest throughputDataset in minutes
          Sample typeProteins, crystals, cells
          ApplicationsSPA, Micro-ED, tomography
          * Indicates a required field
          *
          *
          *
          *
          *
          *
          *
          *
          *
           Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
          This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
          CMD Form Code v3.0
          Style Sheet for Global Design System
          Style Sheet for Komodo Tabs