Cockroach Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
There's a strong link between cockroach allergy, asthma, and allergic rhinitis (aka hay fever).1 Plus, cockroach allergy is an important risk factor for hospital admissions and emergency room visits, as well as a significant cause of asthma related symptoms.2,3 Cockroach allergy also seems to have a larger impact on inner-city youth as an estimated 70 to 80 percent of asthmatic children in inner-city areas are sensitized to cockroach, compared to only 21 percent in suburban environments.4
There are roughly 4,600 species of cockroach, but only a few are considered pests, and just five varieties are frequently found in homes: German, American, Oriental, dusky/smokey brown, and brown-banded.5,6 Of these five, the German and American varieties are the most common species to cause allergies and to infest human dwellings.3,6 Reddish brown in color, the American cockroach measures roughly 30 to 50 millimeters (approximately 2 inches), and the smaller German roach, which is often carried into homes in grocery bags, is typically 12 millimeters (less than 0.5 inches) and is light brown with two dark stripes. Most species prefer warm, humid, and dark environments, and they eat both plant and animal products, as well as items such as paper, clothing, books, dead insects, and human food.5
Allergenic cockroach proteins from cast skins, secretions, egg casings, whole bodies, and fecal materials can become airborne and cause allergy symptoms.2 Cockroaches also spread filth and ruin food and materials, as they disgorge portions of partially digested food, drop feces, and emit secretions that have an offensive smell.7 Unfortunately, cockroaches are commonly found in urban dwellings across the globe.6
Cockroaches prefer warm, moist environments where food is available.7 Factors that increase cockroach infestations include building degradation and high population density.3 Active at night, cockroaches tend to live in groups. They often hide in cracks and crevices in walls, door frames, furniture, cupboards, steam tunnels, basements, televisions, electronic devices, drains, and sewer systems.7 Causing year-round symptoms, allergens can be found throughout dwellings in locales such as beds, furniture, and carpets, with the highest concentrations typically located in the kitchen.11 Even after extermination, however, cockroach allergens may remain in the environment for several months.6
There's both interspecies and cross species reactivity with cockroaches. For example, if you're allergic to American cockroaches, you could also be allergic to German varieties. Plus, you may be allergic to other arthropods (e.g., shrimp and crab), insects (e.g., butterflies and silverfish), arachnids (e.g., dust mites), and mollusks (e.g., oysters and scallops). This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.2
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
If you are allergic to cockroaches, your healthcare provider may recommend a plan that includes the following.2,3,4,7,8,9,12,13
Your healthcare provider may direct you to employ one of the following therapies to improve your allergy symptoms:
Symptoms of cockroach allergy can include:8,9
For those with asthma, cockroach allergy may also cause any of the following symptoms:8
There have been some cases of Oral Allergy Syndrome (OAS) in cockroach allergic patients who consumed crustaceans such as shrimp.6 This syndrome is caused by cross-reacting allergens in both crustaceans and cockroaches. Symptoms of OAS include the following:10
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are allergic to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Cockroach allergens are strong inducers of sensitization and asthma, and cockroach allergy is an important risk factor for emergency room visits and hospital admissions.2