Peanut Allergen Facts, Symptoms, and Treatment
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Access videos and webinars delivered by key experts in the field of allergy.
Part of the legume family, peanuts (aka ground nuts, earthnuts, and goobers) are valued as a nutritionally dense food that's rich in protein and fat.1 While consumed mostly in roasted form throughout the world, peanuts are often eaten whole, made into peanut butter, and incorporated into many food products, such as bakery goods, candies, cereals, sauces, and marinades.2,3
Despite the nutritional value of peanuts, peanut allergy is the most common cause of fatal food allergic reactions.4 Plus, prevalence of peanut allergy has increased steadily in recent years, making it the most common food allergy in children and the second most common food allergy in adults.4,5 Furthermore, in the United Kingdom, peanut allergy affects up to 2 percent of children, and in the United States roughly 2.5 percent of children may be allergic according to at least one study.3,4
Unfortunately, only about 20 percent of affected children outgrow the allergy, so untreated peanut allergies tend to be lifelong.5 Since peanuts are used in a host of foods, avoiding them can be difficult, and accidental reactions are common. There's also a risk for cross-contamination in food-manufacturing facilities that handle peanuts. And according to at least one study, nearly one-third of nut allergic children can't identify the nut to which they're allergic, which further increases the chance of accidental ingestion.4
Peanuts may be found in the following:8 artificial flavoring, baked goods, candy, chili, chocolate, crumb toppings, egg rolls, enchilada sauces, fried foods, flavorings, graham cracker crusts, hydrolyzed plant protein, hydrolyzed vegetable protein, marzipan, mole sauces, natural flavorings, nougats, and various cuisines (e.g., African, Asian, Chinese, Indian, Indonesian, Thai, Vietnamese, and Mexican).
If found on a food label, the following indicates the presence of peanut protein:8 arachic oil, arachis, arachis hypogaea, artificial nuts, beer nuts, boiled peanuts, cold pressed peanut oil (and extruded or expelled peanut oil), crushed nuts, earth nuts, goober peas, ground nuts, hydrolyzed peanut protein, mandelonas, mixed nuts, monkey nuts, flavored nuts, nut pieces, nut meat, peanut butter, peanut butter chips, peanut butter morsels, peanut flour, peanut paste, peanut sauce, peanut syrup, Spanish peanuts, and Virginia peanuts.
Most peanut allergy reactions are due to eating peanuts or foods containing peanuts, but merely touching peanuts can trigger a reaction in some allergic individuals.9 In addition, the food manufacturing and preparation processes also pose a risk for cross-contamination.3 Peanut protein that has become airborne via peanut flour and peanut cooking spray or during grinding or pulverization processes can also cause reactions.3,9 Finally, since peanuts and tree nuts (e.g., pecans, cashews, and walnuts) often touch one another during manufacturing and serving processes, talk with your healthcare provider to determine if it's best to also avoid all tree nuts.5
While relatively new to the United States market, lupine is a common food ingredient in Europe that has a high rate of cross-reactivity with peanuts. That is, if you're allergic to peanut, you might also react to lupine. Also known as lupinus albus, lupine is used in seed, bean, and flour forms, and is a popular ingredient in gluten free foods.8
Some people with a peanut allergy may also experience symptoms when eating other seemingly unrelated foods. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response. The most common cross-reactivities with peanuts are plant foods, e.g., tree nuts, fruits, soybeans, vegetables, and legumes.2
If you experience an itchy mouth or ears, scratchy throat, hives on the mouth, or swelling of the lips, mouth, tongue, or throat after eating peanuts or other related fresh fruits, raw vegetables, or tree nuts, you may suffer from Pollen Food Allergy Syndrome (PFAS) also called Oral Allergy Syndrome (OAS). This condition is caused by your immune system's reaction to similar proteins, or components, found in foods and pollens.7 It is quite common, as one study suggests that up to 25 percent of children with allergic rhinitis (aka hay fever) also suffer from PFAS.11 Common pollen allergies that could cause OAS when eating peanuts include tree (e.g., birch), grass, and weed.2
Peanut consists of different types of proteins that all have different characteristics and different levels of risk for causing symptoms.2 Some people with peanut allergy may tolerate peanuts if they are extensively cooked, as high temperatures break down the causative proteins. However, roasting seems to have the opposite effect, making the peanut more allergenic. For another patient, peanut should be avoided completely, as the proteins are stable even when heated, and could potentially cause a severe event called anaphylaxis. Your specific risk profile depends on which proteins in the peanut you are allergic to.2
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Your healthcare provider may recommend a plan that includes the following.12-15
Your health care provider may direct you to take one of the following medications:
If you're with someone who's having an allergic reaction and shows signs of shock, act fast. Look for pale, cool, and clammy skin; a weak, rapid pulse; trouble breathing; confusion; and loss of consciousness. Do the following immediately:
Peanut allergy can range from mild to severe and may vary over time, resulting in mild symptoms during one episode and severe symptoms in another. Although food allergy symptoms can start a few minutes to several hours after ingestion, most begin within two hours. Symptoms may involve the skin, gastrointestinal tract, cardiovascular system, and respiratory tract, and may include one or more of the following:6
Peanut allergy symptoms can also include anaphylaxis, a whole-body reaction that can impair breathing, cause a dramatic drop in blood pressure, and affect heart rate. The most severe allergic reaction, anaphylaxis can come on within minutes of exposure and can be fatal.6
Symptoms may also include the following, which are associated with Oral Allergy Syndrome (OAS), aka Pollen Food Allergy Syndrome (PFAS):7,8
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are allergic to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
Please note that allergies can change over time, and 10 to 20 percent of children with peanut and tree nut allergies may outgrow them.10
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Knowing the proteins, or components, within each allergen that are triggering your symptoms can help guide your management plan. With that in mind, and based on your symptom history, your healthcare provider may suggest something called a specific IgE component test, which can help reveal other pollens and foods you may react to.2
Already have your specific IgE component test results?
Your component test results will include the name of the components (a series of letters and numbers). Your healthcare provider will likely review the results with you, but here you'll find an at-a-glance breakdown you can use as a reference. Simply match the component names to the list below to see what they mean in terms of symptom management.2
rAra h 1, rAra h 2, rAra h 3, Ara h 6
rAra h 9
rAra h 8
rBet v 2 (profilin)
MUXF3 (CCD)
Test results should be interpreted by your healthcare provider in the context of your clinical history. Final diagnosis and decision on further management is made by your healthcare provider.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.