Meet the next generation of CTS Detachable Dynabeads

Learn more

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
  • Special Offers
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

ELISA Kits
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            LOADING ...
            ...
            • ELISA Kits ›
            • C1S ELISA Kits

            Invitrogen

            Human C1S/Complement Component C1s ELISA Kit

            Technical guide & protocol Tech Support
            Promotion Available
            Learn more
            Catalog # EH57RB
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            666.00
             
            Estimated availability date: 28-May-2025
            Check your price
            Request bulk or custom format
            Join Aspire™ for free ELISA kits
            Technical guide & protocol Tech Support
            Technical guide & protocol Tech Support
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected

              This is a modal window.

              Beginning of dialog window. Escape will cancel and close the window.

              Text
              Text Background
              Caption Area Background
              Font Size
              Text Edge Style
              Font Family

              End of dialog window.

              This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

              Human C1S/Complement Component C1s ELISA Kit (EH57RB)
              Group 53 Created with Sketch.

              FIGURE: 1 / 2

              Human C1S/Complement Component C1s ELISA Kit

              Sandwich ELISA Kit
              View Product

              Human C1S/Complement Component C1s ELISA Kit (EH57RB)
              videoThumbnail
              ►

              Product Specifications

              Analytical sensitivity

              4.1 ng/mL

              Assay range

              4.1-260 ng/mL

              Sample type/volume

              Serum
              50 µL
              Plasma
              50 µL
              Supernatant
              50 µL

              Hands-on time

              1 hr 20 min

              Time-to-result

              4 hr 45 min

              Homogenous (no wash)

              No

              Interassay CV

              <12%

              Intraassay CV

              <10%

              Instrument

              Absorbance Microplate Reader

              Product size

              96 Tests

              Contents

              Pre-coated 96 well plate
              Standard
              Assay Diluent concentrate
              Biotinylated Detection Antibody
              SAV-HRP
              Wash Buffer
              Chromogen
              Stop Solution
              Adhesive Plate Covers

              Shipping conditions

              Wet ice

              Storage

              2-8°C

              Protein name

              C1S

              Species (tested)

              Human

              Assay kit format

              Sandwich ELISA Kit

              Detector antibody conjugate

              Biotin

              Label or dye

              HRP

              About This Kit

              Human C1S/Complement Component C1s quantitates human C1S/Complement Component C1s in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human C1S/Complement Component C1s.

              Target information

              This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.

              For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

              References (0)

              Have you cited this product in a publication?
              Let us know so we can reference it here.
              Cite this product

              Bioinformatics

              Gene aliases : C1S

              Gene ID : (Human) 716

              Gene symbol : C1S

              Protein Aliases : basic proline-rich peptide IB-1, C1 esterase, Complement C1s subcomponent, Complement component 1 subcomponent s

              UniProt ID (Human) P09871

              Recommended Products

              IL-6 Human Matched Antibody Pair

              Because you are interested in IL-6 Human ELISA Kit.

              SIZE

              10 x 96 tests

              PRICE

              USD 400.00

              Cat # CHC1263

              guarantee_icon

              Performance Guarantee

              If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

              Learn more
              help_icon

              We're here to help

              Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

              Contact tech support
              Ordering Plus Icon Minus Icon
              • Order Status
              • Order Help
              • Quick Order
              • Supply Center
              • eProcurement
              Support Plus Icon Minus Icon
              • Help and Support
              • Contact Us
              • Technical Support Centers
              • Documents and Certificates
              • Report a Site Issue
              Resources Plus Icon Minus Icon
              • Learning Centers
              • Promotions
              • Events and Webinars
              • Social Media
              About Thermo Fisher Plus Icon Minus Icon
              • About Us
              • Careers
              • Investors
              • News
              • Social Responsibility
              • Trademarks
              • Consumer Health Data Privacy Policy
              Our Portfolio Plus Icon Minus Icon
              • Thermo Scientific
              • Applied Biosystems
              • Invitrogen
              • Gibco
              • Ion Torrent
              • Fisher Scientific
              • Unity Lab Services
              • Patheon
              • PPD
              • Terms & Conditions
              • Privacy Information Center
              • Price & Freight Policy
              • Cookie Preferences

                Your choices regarding cookies on this site

                We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
              © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
              united states flag icon
              United States

              Your items have has been added!


              Host server : magellan-search-green-6ff95d844f-9jzxh:80/100.66.79.106:80.
              git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
              git-url: https://github.com/thermofisher/magellan-search
              git-branch: release/2.28.0-Offline