Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human CHST4/GST-3 quantitates human CHST4/GST-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CHST4/GST-3.
This gene encodes an N-acetylglucosamine 6-O sulfotransferase. The encoded enzyme transfers sulfate from 3'phosphoadenosine 5'phospho-sulfate to the 6-hydroxyl group of N-acetylglucosamine on glycoproteins. This protein is localized to the Golgi and is involved in the modification of glycan structures on ligands of the lymphocyte homing receptor L-selectin. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : CHST4, GlcNAc6ST2, GST3, HECGLCNAC6ST, LSST
Gene ID : (Human) 10164
Gene symbol : CHST4
Protein Aliases : carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4, Carbohydrate sulfotransferase 4, galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3, GlcNAc6ST-2, gn6st-2, GST-3, HEC-GlcNAc6ST, High endothelial cells N-acetylglucosamine 6-O-sulfotransferase, L-selectin ligand sulfotransferase, LSST, N-acetylglucosamine 6-O-sulfotransferase 2
UniProt ID (Human) Q8NCG5
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support