Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
900-K30K consists of: 900-K30 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
Human GM-CSF ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human GM-CSF in a sandwich ELISA format. Human GM-CSF ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : CSF2, GMCSF
Gene ID : (Human) 1437
Gene symbol : CSF2
Protein Aliases : colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, granulocyte macrophage-colony stimulating factor, Granulocyte-macrophage colony-stimulating factor, MGC138897, MGC151255, MGC151257, M-GM-CSF, granulocyte macrophage colony stimulating factor, MGC131935, Molgramostin, Sargramostim
UniProt ID (Human) P04141
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support