Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

ELISA Kits
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            LOADING ...
            ...
            • ELISA Kits ›
            • IL-2 ELISA Kits

            Invitrogen

            Human IL-2 Uncoated ELISA Kit with Plates

            8 References
            View all (25) IL-2 ELISA Kits
            Technical guide & protocol Tech Support
            Promotion Available
            Learn more
            Catalog # 88-7025-22
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            343.00
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free ELISA kits
            Technical guide & protocol Tech Support
            Technical guide & protocol Tech Support
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected

              This is a modal window.

              Beginning of dialog window. Escape will cancel and close the window.

              Text
              Text Background
              Caption Area Background
              Font Size
              Text Edge Style
              Font Family

              End of dialog window.

              This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

              Human IL-2 Uncoated ELISA Kit with Plates (88-7025-22)
              Group 53 Created with Sketch.

              FIGURE: 1 / 2

              Human IL-2 Uncoated ELISA Kit with Plates

              Sandwich ELISA Kit
              View Product

              Human IL-2 Uncoated ELISA Kit with Plates (88-7025-22)
              videoThumbnail
              ►

              Product Specifications

              Analytical sensitivity

              2 pg/mL

              Assay range

              2-250 pg/mL

              Sample type/volume

              Plasma
              100 µL
              Serum
              100 µL
              Supernatant
              100 µL

              Hands-on time

              1 hr 45 min

              Time-to-result

              24 hr 30 min

              Homogenous (no wash)

              No

              Instrument

              Absorbance Microplate Reader

              Product size

              2 x 96 Tests

              Contents

              Capture Antibody: Pre-titrated, purified antibody
              Detection Antibody: Pre-titrated, HRP-conjugated antibody
              Standard: Recombinant protein for generating standard curve and calibrating samples
              10X Coating Buffer: Buffer for plating the Capture Antibody
              5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme
              Substrate: 1X TMB Solution
              Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
              96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)

              Shipping conditions

              Wet ice

              Storage

              2-8°C

              Protein name

              IL-2

              Species (tested)

              Human

              Assay kit format

              Uncoated ELISA Kit with Plates

              Detector antibody conjugate

              Biotin

              Label or dye

              HRP

              About This Kit

              The Human Interleukin-2 (IL-2) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of Humanmouse IL-2. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately.

              Principle of the method

              ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

              Target information

              Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.

              For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

              Bioinformatics

              Gene aliases : IL-2, IL2, lymphokine, TCGF

              Gene ID : (Human) 3558

              Gene symbol : IL2

              Protein Aliases : aldesleukin, IL-2, T-cell growth factor, TCGF, H-IL-2, il 2, ILN, Interleukin, Interleukin2, Involved in regulation of T cell clonal expansion, M-IL-2, T-cell growth factor (TCGF), Interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor

              UniProt ID (Human) P60568

              Recommended Products

              IL-6 Human Matched Antibody Pair

              Because you are interested in IL-6 Human ELISA Kit.

              SIZE

              10 x 96 tests

              PRICE

              USD 400.00

              Cat # CHC1263

              guarantee_icon

              Performance Guarantee

              If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

              Learn more
              help_icon

              We're here to help

              Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

              Contact tech support
              Ordering Plus Icon Minus Icon
              • Order Status
              • Order Help
              • Quick Order
              • Supply Center
              • eProcurement
              Support Plus Icon Minus Icon
              • Help and Support
              • Contact Us
              • Technical Support Centers
              • Documents and Certificates
              • Report a Site Issue
              Resources Plus Icon Minus Icon
              • Learning Centers
              • Promotions
              • Events and Webinars
              • Social Media
              About Thermo Fisher Plus Icon Minus Icon
              • About Us
              • Careers
              • Investors
              • News
              • Social Responsibility
              • Trademarks
              • Consumer Health Data Privacy Policy
              Our Portfolio Plus Icon Minus Icon
              • Thermo Scientific
              • Applied Biosystems
              • Invitrogen
              • Gibco
              • Ion Torrent
              • Fisher Scientific
              • Unity Lab Services
              • Patheon
              • PPD
              • Terms & Conditions
              • Privacy Information Center
              • Price & Freight Policy
              • Cookie Preferences

                Your choices regarding cookies on this site

                We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
              © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
              united states flag icon
              United States

              Your items have has been added!


              Host server : magellan-search-green-7c4bf66445-frnln:80/100.66.78.233:80.
              git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
              git-url: https://github.com/thermofisher/magellan-search
              git-branch: release/2.30.1-2025.05.22-1.0