Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
900-K39K consists of: 900-K39 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
Human CXCL10 (IP-10) ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human CXCL10 (IP-10) in a sandwich ELISA format. Human CXCL10 (IP-10) ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : C7, crg-2, CXCL10, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10
Gene ID : (Human) 3627
Gene symbol : CXCL10
Protein Aliases : 10 kDa interferon gamma-induced protein, C Cmotif chemokine, C X C motif chemokine, CC motif chemokine, CCmotif chemokine, CXC, CXC motif chemokine, CXCL, H-IP-10, M-IP-10, C-X-C motif chemokine 10, chemokine (C-X-C motif) ligand 10, gamma IP10, Gamma-IP10, interferon-inducible cytokine IP-10, protein 10 from interferon (gamma)-induced cell line, small inducible cytokine subfamily B (Cys-X-Cys), member 10, Small-inducible cytokine B10
UniProt ID (Human) P02778
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support