  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
ELISA Kits
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            LOADING ...
            ...
            • ELISA Kits ›
            • Luteinizing Hormone ELISA Kits
            Invitrogen
            Human Luteinizing Hormone ELISA Kit
            2 References
            View all (3) Luteinizing Hormone ELISA Kits
            Technical guide & protocol Tech Support
            Promotion Available
            Learn more
            Catalog # EHLH
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
            Price (USD)
            672.00
            Estimated availability date: 09-Jun-2025
            Check your price
            Request bulk or custom format
            Join Aspire™ for free ELISA kits
            Technical guide & protocol Tech Support
            Technical guide & protocol Tech Support
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            Stream Type LIVE
            Remaining Time -0:00
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
              This is a modal window.
              Beginning of dialog window. Escape will cancel and close the window.
              Text
              Text Background
              Caption Area Background
              Font Size
              Text Edge Style
              Font Family
              End of dialog window.
              This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
              Human Luteinizing Hormone ELISA Kit (EHLH)
              Group 53 Created with Sketch.
              FIGURE: 1 / 2
              Human Luteinizing Hormone ELISA Kit
              Sandwich ELISA Kit
              View Product
              Human Luteinizing Hormone ELISA Kit (EHLH)
              videoThumbnail
              ►
              Product Specifications
              Analytical sensitivity
              140 pg/mL
              Assay range
              156.3-10,000 pg/mL
              Sample type/volume
              Plasma
              50 µL
              Serum
              50 µL
              Supernatant
              100 µL
              Hands-on time
              1 hr 20 min
              Time-to-result
              4 hr 45 min
              Homogenous (no wash)
              No
              Interassay CV
              <12%
              Intraassay CV
              <10%
              Instrument
              Absorbance Microplate Reader
              Product size
              96 Tests
              Contents
              Pre-coated 96 well plate
              Standard
              Assay Diluent concentrate
              Biotinylated Detection Antibody
              SAV-HRP
              Wash Buffer
              Chromogen
              Stop Solution
              Adhesive Plate Covers
              Shipping conditions
              Wet ice
              Storage
              2-8°C
              Protein name
              Luteinizing Hormone
              Protein family
              Other Proteins
              Species (tested)
              Human
              Assay kit format
              Sandwich ELISA Kit
              Detector antibody conjugate
              Biotin
              Label or dye
              HRP
              About This Kit
              The Human Luteinizing Hormone (Hu LH) ELISA quantitates Hu LH in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu LH.
              Principle of the method
              The Human LH solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
              Rigorous validation
              Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
              Target information
              LH is a member of the glycoprotein hormone family, Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. LH is expressed in the pituitary gland and promotes spermatogenesis and ovulation by stimulating the testes and ovaries to synthesize steroids. The genes for the beta chains of chorionic gonadotropin and for luteinizing hormone are contiguous on chromosome 19q13.3. Mutations in this gene are associated with hypogonadism which is characterized by infertility and pseudohermaphroditism.
              For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
              Bioinformatics
              Gene aliases : CG-ALPHA, CGA, CGB4, FSHA, GPHa, GPHA1, HCG, HH23, LHA, LHB, LSH-B, LSH-beta, TSHA
              Gene ID : (Human) 1081, (Human) 3972
              Gene symbol : CGA, LHB
              Protein Aliases : Anterior pituitary glycoprotein hormones common subunit alpha, CG-alpha, Choriogonadotropin alpha chain, Chorionic gonadotrophin subunit alpha, chorionic gonadotropin, alpha polypeptide, Follicle-stimulating hormone alpha chain, follicle-stimulating hormone alpha subunit, Follitropin alpha chain, FSH-alpha, Glycoprotein hormones alpha chain, interstitial cell stimulating hormone, beta chain, LH-B, LSH-alpha, Luteinizing hormone alpha chain, luteinizing hormone beta subunit, Luteinizing hormone subunit beta, Lutropin alpha chain, Lutropin beta chain, LSH-B, LSH-beta, follitropin, FSH-B, FSH-beta, hLHB, Lutropin subunit beta, Thyroid-stimulating hormone alpha chain, Thyrotropin alpha chain, TSH-alpha
              UniProt ID (Human) P01215, (Human) P01229
              Recommended Products
              IL-6 Human Matched Antibody Pair
              Because you are interested in IL-6 Human ELISA Kit.
              SIZE
              10 x 96 tests
              PRICE
              USD 400.00
              Cat # CHC1263
              guarantee_icon
              Performance Guarantee
              If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
              Learn more
              help_icon
              We're here to help
              Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
              Contact tech support
              Ordering Plus Icon Minus Icon
              • Order Status
              • Order Help
              • Quick Order
              • Supply Center
              • eProcurement
              Support Plus Icon Minus Icon
              • Help and Support
              • Contact Us
              • Technical Support Centers
              • Documents and Certificates
              • Report a Site Issue
              Resources Plus Icon Minus Icon
              • Learning Centers
              • Promotions
              • Events and Webinars
              • Social Media
              About Thermo Fisher Plus Icon Minus Icon
              • About Us
              • Careers
              • Investors
              • News
              • Social Responsibility
              • Trademarks
              • Consumer Health Data Privacy Policy
              Our Portfolio Plus Icon Minus Icon
              • Thermo Scientific
              • Applied Biosystems
              • Invitrogen
              • Gibco
              • Ion Torrent
              • Fisher Scientific
              • Unity Lab Services
              • Patheon
              • PPD
              • Terms & Conditions
              • Privacy Information Center
              • Price & Freight Policy
              • Cookie Preferences
                Your choices regarding cookies on this site
                We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
              © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
              united states flag icon
              United States
              Your items have has been added!
              Host server : magellan-search-green-7c4bf66445-kltx8:80/100.66.73.216:80.
              git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
              git-url: https://github.com/thermofisher/magellan-search
              git-branch: release/2.30.1-2025.05.22-1.0