Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
900-T36K consists of: 900-T36 (capture antibody, detection antibody, and protein standard), and HRP-17UL (streptavidin HRP-conjugate).
Human CCL4 TMB ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinanthuman CCL4 in a sandwich ELISA format. Human CCL4 TMB ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
The TMB ELISA Buffer Kit (Product # 900-T00), sold separately, is recommended to be used in conjunction with this TMB ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
CCL4 (macrophage inflammatory protein 1-beta, MIP1B) belongs to the intercrine beta (chemokine CC) family. Functionally, CCL4 is involved in chemotactic and proinflammatory effects, and homeostasis. CCL4 is produced by macrophages upon stimulation by bacterial endotoxins. CCL4 recruits and stimulates various inflammatory cells at sites of inflammation. CCL4 is produced by lymphocytes, macrophages and dendritic cells. Both CCL4 and the related protein CCL3 participate in the host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells. While both CCL4 and CCL3 exert similar effects on monocytes, their effect on lymphocytes differ, with CCL4 selectively attracting CD4+ lymphocytes and CCL3 selectively attracting CD8+ lymphocytes. Additionally, both have been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. The processed form of CCL4 can induce down-modulation of surface expression of the chemokine receptor CCR5, thus inhibiting the CCR5-mediated entry of HIV-1 in T cells. CCL4 binds with high affinity to CCR5 receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : ACT2, AT744.1, CCL4, G-26, HC21, LAG-1, LAG1, MIP-1-beta, MIP1B, MIP1B1, SCYA2, SCYA4
Gene ID : (Human) 6351
Gene symbol : CCL4
Protein Aliases : ACT-2, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, G-26 T-lymphocyte-secreted protein, HC21, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MGC104418, MGC126025, MGC126026, MIP1beta, MIP1-beta, RP23-320E6.8, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2
UniProt ID (Human) P13236
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support