Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
900-K10K consists of: 900-K10 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
Human VEGF-165 ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human VEGF-165 in a sandwich ELISA format. Human VEGF-165 ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : MVCD1, VEGF, VEGFA, VPF
Gene ID : (Human) 7422
Gene symbol : VEGFA
Protein Aliases : H-VEGF-165, M-VEGF-165, VEGF165, L-VEGF, Vascular endothelial growth factor A, long form, vascular endothelial growth factor A121, vascular endothelial growth factor A165, Vascular permeability factor, VPF
UniProt ID (Human) P15692
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support