Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The mouse Interleukin-17A (IL-17) ELISA kit facilitates the accurate quantification of mouse IL-17 in various biological samples, including murine serum, plasma, buffered solutions, or cell culture medium. This highly specific assay is designed to exclusively detect both endogenous and recombinant forms of mouse IL-17.
Principle of the method
The mouse IL-17 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) utilizes a meticulously paired set of antibodies to capture and measure the target analyte. The microplate wells are pre-coated with a target-specific antibody, enabling the binding of the analyte during the assay. Following the addition of samples or standards to the wells, they bind to the immobilized (capture) antibody. The formation of a sandwich complex is achieved by introducing a second (detector) antibody. Subsequently, a substrate solution is added, initiating a reaction with the enzyme-antibody-target complex and generating a measurable signal. The intensity of this signal is directly proportional to the concentration of the target present in the original specimen.
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : Ctla-8, Ctla8, IL-17, IL-17A, Il17, Il17a
Gene ID : (Mouse) 16171
Gene symbol : Il17a
Protein Aliases : CTLA-8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17, IL-17A, il 17, ILN, Interleukin, interleukin 17, Interleukin17A, R-IL-17-A, Interleukin-17A
UniProt ID (Mouse) Q62386
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support