Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Mouse Interleukin 28B (IFN lambda 3) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-28B. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately.
Principle of the method
ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-28B encodes a cytokine distantly related to type I interferons and the IL-10 family. IL-28B, interleukin 28A (IL28A), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : IFL-1, Ifnl3, IL-28B, Il28, Il28b, INF-alpha, INF-lambda
Gene ID : (Mouse) 338374
Gene symbol : Ifnl3
Protein Aliases : IFN-lambda-3, Interleukin-28B, IL-28B, IFN-lambda 3, ILN, Interferon lambda-4, Interleukin, interleukin 28C, interferon alpha, Interferon lambda-3, interleukin 28, interleukin 28B
UniProt ID (Mouse) Q8CGK6
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support