Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Mouse Lipocalin-2 (LCN2) ELISA quantitates Ms Lipocalin-2 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Lipocalin-2.
Principle of the method
The Mouse Lipocalin-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : 24p3, AW212229, Lcn2, Sip24
Gene ID : (Mouse) 16819
Gene symbol : Lcn2
Protein Aliases : 24p3, Lipocalin-2, Neutrophil gelatinase-associated lipocalin, NGAL, SV-40-induced 24P3 protein, Siderocalin LCN2, p25, HGNC:6526, Lipocalin2, OTTHUMP00000022240, RP23-161B9.11, uterocalin, Oncogene 24p3, secreted inducible protein 24
UniProt ID (Mouse) P11672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support