Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
900-K126K consists of: 900-K126 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
Mouse CCL2 (MCP-1) ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant mouse CCL2 (MCP-1) in a sandwich ELISA format. Mouse CCL2 (MCP-1) ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : AI323594, Ccl2, HC11, JE, MCAF, MCP-1, MCP1, Scya2, Sigje, SMC-CF
Gene ID : (Mouse) 20296
Gene symbol : Ccl2
Protein Aliases : C-C motif chemokine 2, MCP-1, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, Monocyte chemotactic protein 1, Platelet-derived growth factor-inducible protein JE, small inducible cytokine A2, Small-inducible cytokine A2
UniProt ID (Mouse) P10148
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support