Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated human, mouse, and hamster ERK 1/2 and AKT 1/2/3 in cell lysates. Detection of rat ERK 1/2 and AKT 1/2/3 is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Extracellular signal-regulated kinase (ERK) is the founding member and a key component of the classical Mitogen-Activated Protein Kinase (MAPK) pathway. ERK1 (p44, MAPK3) and ERK2 (p42, MAPK2) are both activated via the MAPK/ERK pathway, which is downstream of various Receptor Tyrosine Kinases (RTKs) or GPCRs. MEK, a Ser/Thr and Tyr kinase, activates ERK by phosphorylation of its TxY motifs, namely Thr202/Tyr204 and Thr185/Tyr187 of ERK1 and ERK2, respectively. Once activated, ERK phosphorylates a variety of proteins that regulate cellular processes such as cell division, proliferation, survival, differentiation, apoptosis, motility, and metabolism. AKT, which exists as multiple isoforms, is one of the principle kinases activated by phosphoinositide 3-kinase (PI3K). Activation of PI3K results in the generation of phosphatidylinositol (3,4,5)-triphosphate (PIP3). These lipid second messengers bind to the pleckstrin homology domain of AKT to promote its translocation to the plasma membrane for activation via phosphorylation at Thr308 and Ser473 by PDK1 and the mTOR TORC2 complex, respectively. Phosphorylation at both these sites is required for full activation of AKT Ser/Thr kinase activity. AKT phosphorylates over 50 known substrates, including GSK3, AS160, PRAS40, TSC 1, TCS 2, Raf-1, Bad, the FOXO family of transcription factors, and PFK2. Due to its established critical role in numerous biological processes, AKT is a frequent target for oncology-related drug development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : AKT, AKT1, AKT2, AKT3, CWS6, ERK, ERK-1, ERK-2, ERK1, ERK2, ERT1, ERT2, HIHGHH, HS44KDAP, HUMKER1A, MAPK1, MAPK2, MAPK3, MPPH, MPPH2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, p44-ERK1, p44-MAPK, P44ERK1, P44MAPK, PKB, PKB-ALPHA, PKB-GAMMA, PKBB, PKBBETA, PKBG, PRKBA, PRKBB, PRKBG, PRKM1, PRKM2, PRKM3, RAC, RAC-ALPHA, RAC-BETA, RAC-gamma, RAC-PK-gamma, STK-2
Gene ID : (Human) 207, (Human) 208, (Human) 10000, (Human) 5594, (Human) 5595
Gene symbol : AKT1, AKT2, AKT3, MAPK1, MAPK3
Protein Aliases : AKT1m, ERK, pan AKT, ERK-1, ERK-2, ERT1, ERT2, Extracellular signal-regulated kinase 1, Extracellular signal-regulated kinase 2, extracellular signal-related kinase 1, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 2, MAP kinase 3, MAP kinase isoform p42, MAP kinase isoform p44, MAPK 1, MAPK 2, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, Mitogen-activated protein kinase 3, murine thymoma viral (v-akt) homolog-2, p42-MAPK, p44-ERK1, p44-MAPK, PKB, PKB alpha, PKB beta, PKB gamma, Protein kinase Akt-2, Protein kinase Akt-3, Protein kinase B, Protein kinase B alpha, Protein kinase B beta, Protein kinase B gamma, protein tyrosine kinase ERK2, Proto-oncogene c-Akt, putative v-akt murine thymoma viral oncoprotein 2, rac protein kinase alpha, RAC protein kinase beta, RAC-alpha serine/threonine-protein kinase, RAC-beta serine/threonine-protein kinase, RAC-gamma serine/threonine protein kinase, RAC-gamma serine/threonine-protein kinase, RAC-PK-alpha, RAC-PK-beta, RAC-PK-gamma, serine-threonine protein kinase, STK-2, v-akt murine thymoma viral oncogene homolog 1, v-akt murine thymoma viral oncogene homolog 2, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma), v-akt murine thymoma viral oncogene-like protein 1
UniProt ID (Human) P31749, (Human) P31751, (Human) Q9Y243, (Human) P28482, (Human) P27361
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support