Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

ELISA Kits
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            LOADING ...
            ...
            • ELISA Kits ›
            • VEGF-165 ELISA Kits
            The PeproTech website has now moved to Thermofisher.com. All orders for PeproTech products must be placed on thermofisher.com or offline via phone, fax, email, or distributor partner. For any questions, please contact PeproTech.CustomerService@thermofisher.com

            Invitrogen

            Rat VEGF-165 ELISA Development Kit (ABTS), PeproTech®

            View all (4) VEGF-165 ELISA Kits
            Technical guide & protocol Tech Support
            Promotion Available
            Learn more
            Catalog # 900-K436K
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            440.00
            10 x 96 Tests
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free ELISA kits
            Technical guide & protocol Tech Support
            Technical guide & protocol Tech Support
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected

              This is a modal window.

              Beginning of dialog window. Escape will cancel and close the window.

              Text
              Text Background
              Caption Area Background
              Font Size
              Text Edge Style
              Font Family

              End of dialog window.

              This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

              Rat VEGF-165 ELISA Development Kit (ABTS)
              Group 53 Created with Sketch.

              FIGURE: 1 / 3

              Rat VEGF-165 ELISA Development Kit (ABTS)

              Rat VEGF-165 ABTS ELISA standard curve (Product # 900-K436K).
              View Product

              Rat VEGF-165 ELISA Development Kit (ABTS)
              Rat VEGF-165 ELISA Development Kit (ABTS) (900-K436K)
              videoThumbnail
              ►

              Product Specifications

              Analytical sensitivity

              Lot-specific

              Assay range

              Lot-specific

              Hands-on time

              1 hr 45 min

              Time-to-result

              24 hr 30 min

              Homogenous (no wash)

              No

              Instrument

              Absorbance Microplate Reader

              Product size

              10 x 96 Tests

              Contents

              This Rat VEGF-165 ELISA Kit includes Capture Antibody, Detection Antibody, Protein Standard, and HRP-conjugate. The HRP-conjugate is packaged separately.

              Storage

              Refer to product documentation for component specific storage temperature.

              Protein name

              VEGF-165

              Protein family

              Growth Factors & Receptors

              Species (tested)

              Rat

              Assay kit format

              ELISA Development Kit

              Detector antibody conjugate

              Biotin

              Label or dye

              HRP

              About This Kit

              900-K436K consists of: 900-K436 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).

              Rat VEGF-165 ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant rat VEGF-165 in a sandwich ELISA format. Rat VEGF-165 ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.

              The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.

              Target information

              The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.

              For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

              References (0)

              Have you cited this product in a publication?
              Let us know so we can reference it here.
              Cite this product

              Bioinformatics

              Gene aliases : Vegf, VEGF-A, VEGF164, Vegfa, VPF

              Gene ID : (Rat) 83785

              Gene symbol : Vegfa

              Protein Aliases : H-VEGF-165, M-VEGF-165, VEGF165, Vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VPF

              UniProt ID (Rat) P16612

              Recommended Products

              IL-6 Human Matched Antibody Pair

              Because you are interested in IL-6 Human ELISA Kit.

              SIZE

              10 x 96 tests

              PRICE

              USD 400.00

              Cat # CHC1263

              guarantee_icon

              Performance Guarantee

              If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

              Learn more
              help_icon

              We're here to help

              Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

              Contact tech support
              Ordering Plus Icon Minus Icon
              • Order Status
              • Order Help
              • Quick Order
              • Supply Center
              • eProcurement
              Support Plus Icon Minus Icon
              • Help and Support
              • Contact Us
              • Technical Support Centers
              • Documents and Certificates
              • Report a Site Issue
              Resources Plus Icon Minus Icon
              • Learning Centers
              • Promotions
              • Events and Webinars
              • Social Media
              About Thermo Fisher Plus Icon Minus Icon
              • About Us
              • Careers
              • Investors
              • News
              • Social Responsibility
              • Trademarks
              • Consumer Health Data Privacy Policy
              Our Portfolio Plus Icon Minus Icon
              • Thermo Scientific
              • Applied Biosystems
              • Invitrogen
              • Gibco
              • Ion Torrent
              • Fisher Scientific
              • Unity Lab Services
              • Patheon
              • PPD
              • Terms & Conditions
              • Privacy Information Center
              • Price & Freight Policy
              • Cookie Preferences

                Your choices regarding cookies on this site

                We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
              © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
              united states flag icon
              United States

              Your items have has been added!


              Host server : magellan-search-green-7c4bf66445-kjlsz:80/100.66.72.149:80.
              git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
              git-url: https://github.com/thermofisher/magellan-search
              git-branch: release/2.30.1-2025.05.22-1.0