Overview
The Friedel-Crafts reactions are prominent processes for the substitution of both aromatic and aliphatic compounds with a variety of alkylating agents in the presence of a Lewis acid
Friedel-Crafts Acylation and Alkylation overview
Named Reactions in Organic Synthesis
Friedel-Crafts Acylation and Alkylation Mechanism
Watch our video to learn more about the Friedel-Crafts Acylation and Alkylation and its mechanisms. 
Video Player is loading.
Current Time 0:00
Duration 2:31
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:31
1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
    • en (Main), selected
    Need reagents for the Friedel-Crafts Acylation and Alkylation?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Friedel-Crafts Acylation and Alkylation.
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Friedel-Crafts Acylation and Alkylation
    In 1877, C.Friedel and J.M. Crafts reacted amyl chloride with aluminum pieces in benzene, forming amyl benzene. The reaction of alkyl halides with benzene was found to be general, and aluminum chloride (AlCl3) was identified as the catalyst. Since this discovery, the substitution of both aromatic and aliphatic compounds with a variety of alkylating agents in the presence of a Lewis acid is known as Friedel-Crafts alkylation.
    Before the 1940s, the alkylation of aromatic compounds was the foremost application, but later the alkylation of aliphatic systems also gained in importance. In addition to aluminum chloride, other Lewis acids can also be used: BeCl2, CdCl2, BF3, BBr3, GaCl3, AlBr3, FeCl3, TiCl4, SnCl4, SbCl5, lanthanide trihalides, and alkyl aluminum halides.
    Closely related to the Friedel-Crafts alkylation is the introduction of a keto group into an aromatic or aliphatic compound using an acyl halide or anhydride in the presence of a Lewis acid catalyst, known as the Friedel-Crafts acylation. Compounds that undergo the Friedel-Crafts alkylation are, in most cases, also easily acylated. One drawback of the Friedel-Crafts acylation reaction is that the Lewis acid catalyst often cannot be recovered once the reaction is complete. However, heterogeneous catalysts such as zeolites make this reaction more feasible on an industrial scale.
    Other Electrophilic Aromatic Substitution Reactions
    Other Electrophilic Aromatic Substitution Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:00
    1x
      • Chapters
      • descriptions off, selected
      • subtitles off, selected