Overview
The Clemmensen Reduction converts carbonyl groups to the corresponding methylene group.
Clemmensen Reduction overview
Named Reactions in Organic Synthesis
Clemmensen Reduction Mechanism
Watch our video to learn more about the Clemmensen Reduction and its mechanisms. 
Video Player is loading.
Current Time 0:00
Duration 2:53
Loaded: 0%
Stream Type LIVE
Remaining Time 2:53
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Clemmensen Reduction?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Clemmensen Reduction. 
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Clemmensen Reduction
    n 1913, the Danish chemist Erik Christian Clemmensen reported that simple ketones and aldehydes reacted with amalgamated zinc (Zn/Hg) in the presence of 40% aqueous hydrochloric acid, and in a hydrophobic solvent, such as toluene, to give the corresponding alkanes after several hours under reflux conditions. Ever since, this method of converting carbonyl groups to the corresponding methylene group has been known as the Clemmensen reduction.
    The Clemmensen reduction's original harsh conditions are not conducive to acid-sensitive substrates, so several modifications have been made to increase its synthetic utility by expanding the functional group tolerance. Yamamura and his colleagues developed a milder procedure using organic solvents such as tetrahydrofuran, saturated with hydrogen halides such as hydrogen chloride or bromide, in the presence of activated zinc dust at ice-bath temperatures.
    Many heterocyclic 1,3-dicarbonyl compounds possessing alkyl substituents at the electronegative “2” position exhibit interesting biological properties. Synthesis of many of these molecules was expedited by Thomas Kappe and co-workers using a version of the Clemmensen reduction.
    Other Reduction Reactions
    Other Reduction Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 4:00