Overview of ELISA kits and products
High-quality optimized Invitrogen ELISA kits can help you measure target-specific proteins with confidence, reliability, and consistency. A variety of ELISA kit formats are available which include complete, ready-to-use kits as well as preoptimized reagents to design your own assay. ELISA kits and antibody pairs are available for a range of different species including human, mouse, rat, non-human primate, canine, porcine, bovine, equine, and feline. In addition to the off-the-shelf ELISA formats, special services such as: lot reservation, bulk packaging, and custom target development are available for flexibility and convenience as you start your next research project.
Search ELISA targets  ELISA for endocrinology
90-minute Rapid ELISA kits now available. Discover now
Choosing the right ELISA kit format
 Coated ELISA kit componentsRapid ELISA kit componentsInstant ELISA kit componentsCoated ELISA kit componentsDevelopment ELISA kit components
Coated ELISA kits*Rapid ELISA kitsInstant ELISA kitsUncoated ELISA kitsMatched antibody pair & PeproTech development kits
Explore coated ELISA targetsExplore Rapid ELISA targetsExplore Instant ELISA targetsExplore uncoated ELISA targetsExplore matched antibody pair targets
Features 
Overnight coatingNot requiredNot requiredNot requiredRequiredRequired
Analytical sensitivity**++++++++
Sample volume10–100 µL50 µL25–50 µL10–100 µL10–100 µL
Sample-to-result time125–380 min
(kit specific)
90 min130–250 min
(kit specific)
24 hrs>24 hrs
Data analysisPlate reader with curve-fitting software
 Learn more below about coated ELISA kitsLearn more below about Rapid ELISA kitsLearn more below about Instant ELISA kitsLearn more below about uncoated ELISA kitsLearn more about matched antibody pair kits
*Values in this table refer to our Standard Colorimetric kits. A select number of ultrasensitive kits are also available.
**Every assay has its own specifications. Please consult the protocol for your specific immunoassays/kits.
ProQuantum High Sensitivity Immunoassays
Utilizing proximity-based amplification technology, ProQuantum assays offer the analyte specificity of antibody-antigen binding with the signal detection and amplification capabilities of qPCR to achieve a highly sensitive assay.
ELISA Builder
ELISA Builder can make it easy to find a great solution for developing your own ELISA assay. Based on a short list of questions about your experimental needs, the ELISA Builder tool recommends everything needed to perform an ELISA, from plates to stop solution.
Toxicology Research Products
Explore the newest products for toxicology including ELISA kits for key targets as well as new ProcartaPlex multiplex immunoassays.
Coated ELISA kits
Invitrogen coated ELISA kits are ready-to-use, and have been tested to provide very low inter- and intra-assay variability, with very high lot-to-lot consistency. ELISA kits with coated plates help ensure that you obtain consistent ELISA performance and are designed for labs that require validated and comprehensively tested kits.
Benefits of coated ELISA kits:
  • Validated performance⁠—tested for plate variability (CV) and final kit validation testing before shipping out
  • Easy-to-use⁠—includes all the required assay buffers and reagents, and a straightforward protocol
  • Fast⁠—pre-coated plates allow for a workflow typically completed within four hours
Coated ELISA kit contents
  • Coated 96-well plate
  • Pre-titrated capture antibody
  • Pre-titrated detection antibody Streptavidin-HRP conjugate
  • Recombinant protein standard
  • Stop solution
  • Substrate solution
  • Assay buffer (sample diluent)
  • Wash buffer
Video Player is loading.
Current Time 0:00
Duration 4:46
Loaded: 0%
Stream Type LIVE
Remaining Time 4:46
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Using Invitrogen pre-coated ELISA kits
    View the instructional video showing how to use Invitrogen pre-coated ready-to-use ELISA kits. Please make sure to carefully read and review your ELISA kit’s specific instruction sheet, located inside the package, for detailed information before beginning the procedure.
    Rapid ELISA kits
    Invitrogen Rapid ELISA kits have a condensed workflow with only one wash step, reducing the overall assay time to 90 minutes. Compared with conventional ELISA that requires three incubations for samples, biotinylated antibodies and enzyme conjugates, Rapid ELISA kits can be completed in one step, reducing operating steps by 1–2 hours.
    Benefits of Rapid ELISA kits
    • Short assay time—only 90 min from sample to results
    • Ease of use—simplified ELISA procedure with a single wash step
    • Excellent performance—Comparable sensitivity and performance with conventional ELISA
    Rapid ELISA kit contents
    • ELISA Plate​
    • Reference Standard ​
    • Concentrated HRP Linked Detection Ab (100X)​
    • Reference Standard & Sample Diluent
    • HRP Linked Ab Diluent​
    • Concentrated Wash Buffer (25X)​
    • Substrate Reagent​
    • Stop Solution​
    Figure 1. Rapid ELISA kit run time compared to conventional ELISA.
    Instant ELISA kits
    Invitrogen Instant ELISA Kits have a condensed workflow format designed to alleviate the inconveniences of multiple preparation and incubation steps. In conventional ELISA kits, the precoated plates provide only the capture antibody, to which the sample and detection reagents must be added sequentially. In contrast, Instant ELISA Kits come with ready-to-use plates that contain all necessary assay components—including capture antibody and lyophilized detection antibody, streptavidin-HRP, and sample diluent. These prepared plates greatly reduce pipetting time and enable a simple one-wash protocol. Furthermore, serially diluted, lyophilized protein standards are provided in additional strip wells, eliminating the need for plate setup and serial dilutions to create a standard curve, leaving less room for error.
    Benefits of Instant ELISA kits
    • Ready-to-use—microplates contain all necessary assay components
    • Time saving—standards are already serially diluted in the plate
    • Convenient—sequential addition of reagents during assay minimized
    Instant ELISA kit contents
    • 96-well plate (12 strips with 8 wells each) coated with capture antibody, biotin-conjugated detection antibody, streptavidin-HRP conjugate and assay buffer (lyophilized)
    • Recombinant lyophilized protein standard dilutions (in additional strips)
    • Stop solution
    • Substrate solution
    • Wash buffer
    Figure 2. Comparison of Instant ELISA technology vs. conventional ELISA procedures. With Instant ELISA kits, all the preparation work has been done for you, helping decrease hands-on time per experiment. In addition, Instant ELISA kits come with a full 96-well plate for your samples and two separately packed standards. This enables the use of all 96 wells for your samples expanding your ELISA capacity by 20%.
    Table 1. Comparing conventional ELISA and Instant ELISA workflow steps. Fewer steps help save time and reduce variation. There are only 7 working steps when using an Instant ELISA kit, as opposed to 17 steps when performing a conventional ELISA. Therefore, you can decrease total hands-on time per experiment to about 1 hour by adopting the Instant ELISA technique.
    7 steps for Instant ELISA kits 17 steps for conventional ELISA kits
      1. Washing of coated plates
    2. Reconstitution of standard protein
    3. Addition of diluent to standard wells
    1. Rehydration of standard and sample wells on plate 4.Titration of standard curve
    5. Addition of sample diluent
    2. Sample addition 6. Sample addition
      7. Dilution of biotin conjugate
    8. Addition of biotin conjugate
    3. Incubation 9. Incubation
      10. Preparation of streptavidin-HRP conjugate
    11. Washing
    12. Addition of streptavidin-HRP conjugate
    13. Incubation
    4. Washing 14. Washing
    5. Addition of TMB substrate 15. Addition of TMB substrate
    6. Addition of stop solution 16. Addition of stop solution
    7. Calculation of results 16. Calculation of results
    ELISA components—create your own assay
    ELISAs can be used to identify and quantitate just about any type of analyte. When designing an ELISA for your unique target of interest, deciding which components are right for you is often the most difficult part. The ELISA Builder is an online selection tool that guides you through a short series of questions about your specific ELISA and then recommends all the components you’ll need—from plates to stop solution.
    Explore additional ELISA products
    ELISA Enzyme Substrates
    ELISA Enzyme Substrates
    Find HRP and other enzyme substrates for ELISA applications in a range of sensitivities for different methods including colorimetric, chemiluminescent, and fluorescent detection.
    Learn more
    ELISA Blocking Buffers and Reagents
    ELISA Blocking Buffers and Reagents
    Select from a wide variety of easy-to-use and reliable blocking buffers and reagents for ELISA applications.
    Learn more
    ELISA Instrumentation and Equipment
    ELISA Instrumentation and Equipment
    We offer centrifuges, cold storage equipment, dispensers, incubator/shakers, microplates, pipettes and tips, and microplate readers and washers to help meet all your ELISA needs.
    Learn more
    ELISA Microplates an Plasticware
    ELISA Microplates and Plasticware
    Choose from an array of microplates with a variety of surfaces optimized to capture the biomolecule of your choice.
    Learn more
    Uncoated ELISA kits
    Invitrogen uncoated kits are ideal for experienced ELISA users on a budget with less stringent requirements for inter- and intra- assay variance. These kits have the reagents needed to coat-your-own plates using an overnight process and run the assay. Kit configurations can provide easy-to-use, flexible options.
    Benefits of uncoated ELISA kits
    • Pre-titrated and optimized—matched antibody pairs and all provided reagents optimized for ELISA development
    • Many options for kit configuration—purchase based on the volume of your assay through a choice of package sizes, including 2-, 10-, and 20-plate kits
    • Complete—includes all the essential reagents and buffers to limit variability. Generic reagents like wash and stop solution not included
    • Affordable—priced to accommodate demanding budgets and help maximize your research
    Uncoated ELISA kit contents
    • Pre-titrated capture antibody
    • Pre-titrated detection antibody, biotin-conjugated
    • Recombinant protein standard
    • Substrate solution
    • Assay buffer (sample diluent)
    • Coating buffer
    • Avidin- or streptavidin-HRP conjugate (kit specific)
    Matched antibody pair and PeproTech Development ELISA kits
    Invitrogen antibody pair kits contain pre-matched antibody pairs, standards, and streptavidin-HRP to develop your own ELISA assays. PeproTech Development kits are also used to design new ELISA protocols, with a variety of targets available in both ABTS and TMB formats for flexibility.
    Benefits of matched antibody pair kits
    • Flexible—matched antibody pairs can be used for ELISA or with other immunoassay platforms
    • Convenient packaging option—All-in-one buffer kit sold separately, if needed for ELISA development
    Matched antibody pair kit contents
    • Capture (coating) antibody
    • Detection antibody
    • Streptavidin-HRP conjugate
    • Standard
    • Sample diluent (not included in all kits)
    Explore matched antibody pair targets
    Benefits of ELISA development kits
    • Flexibility—available in both ABTS and TMB formats for design flexibility
    • Convenience—specifically formulated Buffer Kits available
    ELISA development kit contents
    • Capture (coating) antibody
    • Detection antibody
    • Protein standard
    • HRP-conjugate
    Video Player is loading.
    Current Time 0:00
    Duration 5:04
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 5:04
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      How to coat a plate and use Invitrogen uncoated ELISA kits
      View the instructional video showing how to coat a plate and use Invitrogen uncoated ELISA kits. Please make sure to carefully read and review your ELISA kit’s specific instruction sheet, located inside the package, for detailed information before beginning the procedure.
      Ordering information for related reagents and buffers
      Catalog # Name Size Price (USD) Qty
      CNB0011 ELISA Buffer Kit Each
      589.65
      Online Exclusive
      624.00
      Save 34.35 (6%)
      900-T00 TMB ELISA Buffer Kit, PeproTech® Each
      269.00
      900-K00 ABTS ELISA Buffer Kit, PeproTech® Each
      269.00
      Resources and related products
      Inside cell11 luminex eyecandy
      Immunoassay Research Areas
      Explore our immunoassays for different research areas, including immune-oncology, neurobiology, isotyping and immunoglobulin characterization, hallmarks of cancer, car t cells, and more.
      Learn more
      A range of plate readers to enable maximum flexibility and performance
      Family
      Visualize results quickly and easily with fast plate reads and an intuitive suite of software. Experience a software suite and application options that evolve with your lab. Measure a variety of signals including fluorescence, absorbance, luminescence, or time-resolved fluorescence on a Thermo Scientific microplate reader. From upgradable multimode systems to dedicated readers, we offer solutions that fit your current budget, with options to meet your lab’s future needs.
      Compare readers
      For Research Use Only. Not for use in diagnostic procedures.
      Stylesheet for Classic Wide Template adjustments
      Hero banner white gradient overlay