Terrific broth (TB) is a nutritionally rich medium for the growth of bacteria (Tartoff, et al., 1987). The formulation of TB medium contains increased concentrations of peptone, yeast extract, and glycerol as a carbon source. We offer TB media in both powder and ready-to-use liquid formats for convenience and ease of use.
Ordering Information
Catalog # Name Size Price (USD) Qty
A1374301 Terrific Broth Each
50.65
Online Exclusive
55.50
Save 4.85 (9%)
22711022 Terrific Broth Each
124.65
Online Exclusive
132.00
Save 7.35 (6%)
For Research Use Only. Not for use in diagnostic procedures.