History of innovation and passion for technology
Thermo Scientific Centrifuges are products of decades of experience in manufacturing and a deep understanding of our customers’ needs. With many proprietary technologies and innovative ideas, we support our customers to manage separation, harvesting, isolation and purification steps as efficiently as possible.
Auto-Lock rotor technology
Our Auto-Lock rotor exchange capability lets you safely and securely swap a rotor with one push of a button. Labs have the flexibility to change rotors and application protocols instantly with no tools or complicated procedures. In as short as three seconds, with just one hand, remove one rotor and replace it with another. Operators can quickly and efficiently access the entire centrifuge chamber for cleaning.
Auto-Lock system is available with our general purpose, compact benchtop, and superspeed models.
Intuitive interface
X Pro series interface Centri-Touch interface Simplified control panel
Available on the General Purpose X Pro Centrifuge Family
The touchscreen interface makes programming easy from a menu bar and simple buttons. An enlarged time tracker and visual and audible end-of-run alerts let you conveniently monitor runs, even from across the lab. Pre-programmed compressor startup has the centrifuge ready to run once you arrive, and shutdown extends compressor life and helps reduce energy costs.
Read Smart Note
Available on superspeed and large capacity floor models
The Centri-Touch interface offers a user-friendly experience with up to 120 programs accessible through alpha-numeric naming. It provides in-use training through tutorial videos and a quick-start manual. The integrated rotor calculator simplifies protocol modifications, and enhanced data management allows user access control and easy USB export of event logging. The backlit display helps ensure visibility from across the lab, with the option to enlarge during runs. Multilingual instructions enable easy navigation for users of different language backgrounds.
Watch video
Available on compact benchtop centrifuges and other select models
Our user-friendly, easy-to-use control panel for centrifuges allows one-touch operation and offers pre-saved protocols. The backlit display allows easy reading from a distance, helping reduce errors. Optional indicators, like automatic lid opening and adjustable signals, offer customizable notifications. The panel is glove- and detergent-friendly. Multilingual instructions in various languages enhance usability for users of different backgrounds.
Biocontainment
The ClickSeal lids provide a biocontainment sealing option that offers glove-friendly, one-handed open/close capability. These lids are ergonomically designed to accommodate both right and left-handed operations, helping ensure easy operation for all laboratory users. By eliminating the need for multi-turn screw caps and complicated high-pressure clips, the ClickSeal lids simplify the sealing process.
To enhance safety and security, the ClickSeal lids feature audible and visual closure cues. These cues provide clear indications when the lids are properly closed, minimizing the risk of accidental exposure or contamination.
The ClickSeal lids are certified and provide a secure and quick solution to help ensure the safe containment of pathogens. In the event of tube breakage, tube leakage, external tube contamination, or over-fill contamination within tube and cap threads, the ClickSeal lids help prevent the release of pathogens when used correctly.
These lids are available for use with multiple swing-out and fixed angle rotors, providing versatility and compatibility with various centrifuge setups.
It is important to note that the ClickSeal lids have obtained biocontainment certification from reputable organizations such as the National Institute for Health Protection in Porton Down, UK, or TÜV NORD CERT GmbH in Germany. This certification validates their effectiveness in containing pathogens and meeting stringent safety standards.
Data management and connectivity
 Centri-Log Plus Software   Centri-Vue App    SECURE-SpinMode  
For large-capacity, superspeed, bioprocessing, and general purpose centrifuges
  • Data management software for centralized monitoring of unlimited number of centrifuges on the same network
  • Supports GMP and 21 CFR Part 11 compliant data tracking
  • Provides complete traceability by generating barcodes for users, samples, programs and process results centrifuges
  • Can connect to LIMS via a third-party software data access point
  • Real-time run data for all networked centrifuges
Learn more
For large-capacity, superspeed, and bioprocessing centrifuges
  • Mobile app to check centrifuge status at a glance
  • Monitors run data and check for diagnostic errors from your mobile device
  • Enables remotely starting/ stopping the run
Download Centri-Vue flyer
For General Purpose X Pro Centrifuges
  • Advanced user management feature on the centrifuge user interface
  • Can be connected to Centri-Log Plus software
  • Supports 21 CFR Part 11 and GMP compliant user and data management standards
  • Allows use of sample tracking kit
Learn more
Auto-ID instant rotor identification
Available on superspeed and large capacity centrifuges.
Auto-ID Instant Rotor Identification is an advanced feature that allows for the immediate identification of a rotor as soon as it is placed in the centrifuge chamber. This innovative technology automatically loads the specific rotor's specifications into the centrifuge parameters, eliminating the need to manually search for and set rotor codes.
The primary benefit of Auto-ID is the significant reduction in run setup time. By eliminating the manual input of rotor codes, this feature streamlines the process and helps save valuable time in the laboratory, allowing for increased productivity and efficiency. Moreover, Auto-ID enhances safety measures by minimizing the risk of over-speed and reducing the occurrence of error messages. With the rotor parameters automatically loaded, the centrifuge operates with optimal settings, helping ensure the safety of both the centrifuge itself and the samples being processed. This feature contributes to improved overall safety for operators as well.
Utilizing Auto-ID is straightforward. Once the rotor is placed in the centrifuge chamber, the system immediately detects it and loads the corresponding parameters. Users simply need to confirm the settings and initiate the run, resulting in a seamless and hassle-free operation.
Auto-ID Instant Rotor Identification helps simplify the centrifuge workflow by providing quick and accurate rotor identification, optimizing efficiency, and enhancing safety in the laboratory setting.
Sustainability
centrifuge sustainability
Reduced energy consumption—Advanced Thermo Scientific features like Green Mode Technology, Smart Vacuum Technology, Eco-Spin Windshielded Rotors, and Thermo Scientific Fiberlite Carbon Fiber Rotors provide energy savings of up to 74% compared to older centrifuge models.
Efficient cooling systems—To manage energy and environmental impact in high-use environments, certain centrifuge models are designed to shut off the automatic cooling function when the instrument is off or when the door opens, thus helping reduce power consumption and save energy on refrigeration.
Less waste—All excess scrap material created during manufacturing is reused, and the lightweight Fiberlite carbon fiber rotors are corrosion resistant with a lifespan up to 300% longer than comparable metal rotors.
RoHS and WEEE compliant—Thermo Scientific centrifuges are fully compliant with the Restriction of Hazardous Substances (RoHS) directive, which promotes the reduced use of hazardous substances such as lead, cadmium, mercury, chromium6+, PBB, and PBDE. WEEE compliance helps ensure that our centrifuges utilize environmentally sound manufacturing practices.
Review the Green Fact Sheets:
Ergonomics and safety
centrifgue ergonomics
With each updated model, we prioritize ergonomic improvements to help ensure that our centrifuges seamlessly integrate into the actual work environment. Our focus on enhancing loading and unloading processes leads to safer use and improved accessibility. We achieve this through the implementation of motorized gas springs, Auto-Lock rotor technology, and automatic door opening and closing technologies.
We understand the importance of user interface visibility and reach, as well as the need for one-handed and glove-friendly operation. Our centrifuges are designed with these considerations in mind, providing a user-friendly experience that promotes efficiency and ease of use.
To further prioritize safety, we have made efforts to reduce rotor weight and incorporate enhanced safety features. Our carbon fiber rotors are lightweight and easy to handle, helping ensure user comfort and minimizing the risk of accidents. Additionally, our Thermo Scientific SmartSpin imbalance detection system automatically detects imbalances to enable stable runs, while the Thermo Scientific Accumulated Centrifugal Effect (ACE) function adjusts run time to accommodate variations in acceleration.
For our large capacity centrifuges, we offer Auto-Door opening technology, which allows the centrifuge door to be released from the front to the back without the operator having to touch it. This feature promotes convenience and prevents potential contamination. Furthermore, our Auto-Lid opening mechanism instantly releases rotor lids, allowing them to be stored inside the centrifuge door. This not only saves space but also enhances efficiency in the laboratory.
We continuously strive to improve the ergonomic design and safety features of our centrifuges, ensuring that they meet the specific needs of the work environment
Review the Smart Notes:
Standards and certifications
centrifuge certifications
Choose between general use or IVD (clinical) models for many Thermo Scientific centrifuges. Our clinical models are in compliance with the EU IVDR (EU) 2017/746 and are listed by the FDA.
Our centrifuges are available with the following certifications: UL, CE, RoHS, CSA.
Thermo Scientific centrifuges are built to the highest global standards for quality, safety and performance. Contact us to learn more about IVD models, cleanroom compatibility, and global standards.
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.
1x1 image pixel for data collection 1x1 image pixel for data collection