What is Cells-to-CT?
Invitrogen Cells-to-CT kits measure relative gene expression by real-time quantitative PCR (RT-qPCR) without the need for RNA purification prior to amplification. Available in both TaqMan and SYBR Green detection chemistries, Cells-to-CT kits provide a complete workflow for qPCR analysis directly from cultured cells, providing you with excellent performance, reliability, and world-class support.
Benefits of Cells-to-CT kits
Video Player is loading.
Current Time 0:00
Duration 0:58
Loaded: 0%
Stream Type LIVE
Remaining Time 0:58
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    • Extraordinary ease and speed—96 samples for RT-qPCR in less than 10 min
    • Skip RNA purification—no columns, heating, or centrifugation
    • Exceptional performance—designed for consistent accuracy, reproducibility, and sensitivity with 1–100,000 cells
    • Analytically validated workflows—complete sets of reagents pre-optimized to work efficiently right out of the box
    Invitrogen TaqMan Cells-to-CT Express Kit
    Perform RT-PCR directly from cell lysates without extraction using the newest product in the Cells-to-CT family. This kit allows you to prepare cells for RT-qPCR analysis in just 5 minutes with a simplified workflow that is suitable for a few samples, or scalable for automated, high‑throughput applications.
    Order Now
    Cells-to-CT kit selection guide
     TaqMan Cells-to-CT Express KitCells-to-CT 1-Step TaqMan KitSingle Cell-to-CT qRT-PCR KitTaqMan Fast Advanced Cells-to-CT Kit
     product photoproduct photoproduct photoproduct photo
    Cell working range10–100,000 cells10–100,000 cells1–10 cells10–100,000 cells
    1-step or 2-step2-step1-step1-step2-step
     SYBR Green Fast Advanced Cells-to-CT KitCells-to-CT 1-Step Power SYBR Green Kit
     product photoproduct photo
    Cell working range10–100,000 cells10–100,000 cells
    1-step or 2-step2-step1-step
     TaqMan Cells-to-CT Express KitCells-to-CT 1-Step TaqMan KitSingle Cell-to-CT qRT-PCR KitTaqMan Fast Advanced Cells-to-CT Kit
     product photoproduct photoproduct photoproduct photo
    Cell working range10–100,000 cells10–100,000 cells1–10 cells10–100,000 cells
    1-step or 2-step2-step1-step1-step2-step
     SYBR Green Fast Advanced Cells-to-CT KitCells-to-CT 1-Step Power SYBR Green Kit
     product photoproduct photo
    Cell working range10–100,000 cells10–100,000 cells
    1-step or 2-step2-step1-step
    Read more about the difference between 1-step vs 2-step kits
    Read more about the difference between TaqMan vs SYBR Green detection chemistries
    Cell lineGrowth typeSource speciesSource tissue
    A549adherentH. sapiensLung carcinoma
    BJadherentH. sapiensForeskin
    CHO-K1adherentC. griseus (hamster)Ovary
    COS-7adherentC. aethiops (monkey)Kidney
    DU-145adherentH. sapiensProstate
    HEK-293adherentH. sapiensKidney
    HeLaadherentH. sapiensCervical adenocarcinoma
    HepG2adherentH. sapiensLiver carcinoma
    Huh-7adherentH. sapiensLiver
    JurkatsuspensionH. sapiensAcute T cell leukemia
    K562suspensionH. sapiensBone marrow
    ME-180adherentH. sapiensCervical epidermoid carcinoma
    NCI-H460adherentH. sapiensLung
    Neuro 2AadherentM. musculusBrain
    NIH/3T3adherentM. musculus (mouse)Embryonic fibroblast
    PC-12adherentR. norvegicus (rat)Adrenal pheochromocytoma
    PT-K75adherentS. scrofa (pig)Nasal turbinate mucosa
    RajisuspensionH. sapiensB lymphocyte
    SK-N-ASadherentH. sapiensBrain neuroblast
    SK-N-SHadherentH. sapiensBrain fibroblast
    U-2 OSadherentH. sapiensBone
    U-87 MGadherentH. sapiensBrain glioblastoma 
    Note: Due to differences in cell size and composition, the maximum number of cells per lysis reaction may be slightly different for different cell lines. Test for inhibition and minimal sample input by using the TaqMan Cells-to-CT Control Kit.
    Research solutions for downstream RT-PCR (qPCR) analysis
    Leverage our diverse product offerings for real-time, quantitative PCR applications to build a complete experimental workflow solution. Explore our powerful assay design algorithms, optimized master mixes, intuitive data analysis software, and flexible instrumentation to help harness the power of RT-qPCR across a rich and diverse set of applications.
    Featured resources
    Video Player is loading.
    Current Time 0:00
    Duration 32:45
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 32:45
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Cells-to-CT Kits: A fast way to kill your cells for gene expression analysis
      This ASCB presentation shows you how Cells-to-CT kits allow you to measure relative gene expression by qPCR without having to purify RNA prior to amplification.
      Video Player is loading.
      Current Time 0:00
      Duration 3:14
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 3:14
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Cells-to-CT: full video
        Learn how to prepare RNA for gene expression analysis from cultured cells, without the need for RNA purification. Fast track to RT-qPCR in only seven minutes. 
        Technical notes
        Resources for Cells-to-CT kits
        Resources
        • Protocol Videos—Videos to help you isolate nucleic acids using a variety of techniques.
        • DNA & RNA Selection Guide—Find the right purification products.
        • Videos—View our library of DNA & RNA purification and analysis videos.
        • Application Notes—Explore our application notes from scientists sharing data for isolation products
        For Research Use Only. Not for use in diagnostic procedures.
        Stylesheet for Classic Wide Template adjustments