c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
SEM imaging and EDS elemental analysis
Even though scanning electron microscopy (SEM) has made tremendous advances, it can be considered by new users to be a complicated technique. Sample preparation, alignments, cost of ownership, and difficulties with analytical techniques prevent widespread adoption. We set out to resolve these challenges in order to make a truly accessible SEM without compromising on flexibility. Our goal was to make microscopy an easy and enjoyable experience in which you should not have to fight the microscope to obtain quality data. The result is the Thermo Scientific Axia ChemiSEM Scanning Electron Microscope, a system in which obtaining SEM-EDS data is no longer a chore. EDS is no longer just an afterthought—chemical data and imaging have now become one.
SEM EDS analysis
The Axia ChemiSEM is unlike traditional SEMs in that it always collects EDS data in the background. It uses unique algorithms to process the SEM and EDS signals simultaneously, allowing it to display the morphology and quantitative elemental make up of a sample together, in real time. It constantly processes EDS data in the background, giving you live updates on elemental data, as it is acquired. Elements found in your sample can be toggled on and off, allowing you to isolate areas of interest.
The Axia ChemiSEM features a superior user experience enhanced by automation, such as SmartAlign technology for alignment-free operation, newly developed automatic functions, and live quantitative EDS mapping. This always-on processing make analysis twice as fast, compared to traditional methods. The Axia ChemiSEM Scanning Electron Microscope puts SEM/EDS analysis within reach of a wider scientific audience, speeds up your workflow, and saves time on training.
Particle Analysis Learning Center
Learn more
Key Features
Live quantitative elemental mapping
Update your Analytical workflow with fully integrated, cutting-edge Thermo Scientific ChemiSEM technology that provides characterization in half the time.
Easy to get started
Integrated User Guidance, live real-time compositional imaging, and alignment-free operation combine on this instrument to allow you to stay focused on data collection.
Flexible sample loading
The flexible stage accommodates samples up to 10 kg with x-y movement, and the door opens fully for easy loading of large, awkward samples - while retaining a very fast pump down time.
Excellent imaging performance
Easily handle a wide range of applications, including insulating samples with Low Vac operation and advanced beam scanning to ensure high-fidelity imaging
Diverse application software
Handle advanced applications with advanced system automation option, such as Thermo Scientific Maps™ Software, and customize workflow development with Thermo Scientific AutoScript™ Software.
Easy maintenance
Source exchange is easy and can be completed by any user, while the straightforward design translates to high uptime and rapid service when attention is needed.
Specifications
Style Sheet for Products Table Specifications
Standard
  • Motorized retractable backscatter detector
  • TrueSight EDS detector
  • Nav-Cam Camera—color optical camera for sample navigation
  • Manual User Interface
  • IR chamber camera
  • Maps Software for tiling and stitching
Electron beam resolution
  • 3.0 nm @ 30 kV (SE)
  • 3.0 nm @ 30 kV (SE) (low vacuum)
  • 8.0 nm @ 3 kV (SE)
  • 7.0 nm @ 3 kV (BD mode* + BSE)
Chamber
  • Inner width: 280 mm
  • Ports: an Axia ChemiSEM Scanning Electron Microscope configured with BSED and EDS offers 5 available ports
Stage
  • XY: 120x120 mm
  • Tilt: -15 to +90 degrees
  • Rotation: n x 360°
  • With ZTR axes removed:
    • Max. sample height: 128 mm
    • Max. sample weight: 10 kg
Common options
  • Large TrueSight EDS detector
  • Quasor-II EBSD
  • ChemiSEM Advanced Technology for drift correction and quant maps per element
  • ChemiView license for offline EDS data processing
  • Beam Deceleration mode
  • CleanHeater stage (up to 1,100°C)
  • Photon cathodoluminescence detector
  • AutoScript 4 Software—Python-based application programming interface
  • Thermo Scientific TopoMaps Software for image colorization, image analysis, and 3D surface reconstruction
Style Sheet for Komodo Tabs
Resources
On-demand Webinar: Meet our newest SEM
Attend this webinar to:
  • Discover zero-click elemental analysis that enhances accessibility and productivity with live quantitative elemental imaging.
  • Experience the future-proof flexibility of a large chamber, support of samples up to 10 kg, and enough ports to accommodate many accessories.
  • Find out how you can save time with fast pumping, automatic alignments, and an easy interface.
Watch webinar
Meet the Axia ChemiSEM System
Meet the Axia ChemiSEM: a SEM EDS instrument that is flexible, easy to use, and offers instantaneous, quantitative elemental analysis.
Video Player is loading.
Current Time 0:00
Duration 2:32
Loaded: 6.44%
Stream Type LIVE
Remaining Time 2:32
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextMeet the Axia ChemiSEM System
        Meet the Axia ChemiSEM: a SEM EDS instrument that is flexible, easy to use, and offers instantaneous, quantitative elemental analysis.
      2. Now Playing
        Up NextAlignment-free operation
        Axia is always ready to image with an autofocus routine that is easier and quicker than ever. It comes with the ability to focus on the feature of choice with a simple click.
      3. Now Playing
        Up NextZero-click elemental analysis-Linescan Brass
        Reliable quantification can be obtained directly from the live image using a choice of standard EDS functions (region/point ID/line scan).
      4. Now Playing
        Up NextAxia ChemiSEM’s geological section tooltip
        With Axia ChemiSEM, quantitative elemental information is provided through colors in the image. Moreover, a tooltip shows the relative concentrations of the sample constituents, making it the quickest way to obtain composition information during live
      5. Now Playing
        Up NextAxia ChemiSEM: two times faster than traditional SEMs.
        Axia ChemiSEM provides live quantitative compositional information during routine SEM navigation and imaging, two times faster compared to traditional SEMs. With the possibility to easily select and unselect the elements of interest, you can focus on
      6. Now Playing
        Up NextZero-click elemental analysis - conventional mapping
        Reliable quantification can be obtained directly from the live image using a choice of standard EDS functions (region/point ID/line scan).
      7. Now Playing
        Up NextConventional EDS functions in a state-of-the-art system
        Conventional EDS functions are available in a state-of-the-art implementation, such as gross counts mapping, which allows to post-process your data by selecting, for each element, the energy line used to generate the elemental map.
      8. Now Playing
        Up NextAxia ChemiSEM’s tooltip, the quickest way to obtain composition information during live imaging
        Axia ChemiSEM provides live compositional information during routine SEM navigation and imaging. The tooltip shows the relative concentrations of the sample constituents, making it the quickest way to obtain composition information during live imagin
      9. Now Playing
        Up NextZero-click elemental analysis - Geological Section
        Reliable quantitative information of a specific point or region of interest can be obtained using traditional EDS functions, directly on the SEM image. Multi-point analysis allows for easy comparison of different features on the sample.
      10. Now Playing
        Up NextBatteries Zero Click Analysis
        With Axia ChemiSEM, elemental analysis is always on, so that compositional information is never more than a mouse click away. Conventional EDS functions operate directly on the image without intermediate steps.
    ChemiSEM Testimonial - Carlos Angeles Chavez
    Carlos Angeles Chavez from the Mexican Petroleum Institute comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
    Video Player is loading.
    Current Time 0:00
    Duration 1:48
    Loaded: 9.11%
    Stream Type LIVE
    Remaining Time 1:48
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
        1. Now Playing
          Up NextChemiSEM Testimonial - Carlos Angeles Chavez
          Carlos Angeles Chavez from the Mexican Petroleum Institute comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
        2. Now Playing
          Up NextChemiSEM Testimonial - Lifeng Dong
          Lifeng Dong from Hamline University comments on his first impressions of his live demo with the Thermo Scientific™ Prisma E SEM with ChemiSEM technology.
        3. Now Playing
          Up NextChemiSEM Testimonial - Cedric Barroo
          Cedric Barroo from the Universite Libre de Bruxelles comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
        4. Now Playing
          Up NextChemiSEM Testimonial - Houston Electron Microscopy
          Dennis and Marina Manuel comment on the time saving and productivity benefits that they experienced in their live demonstration of the Thermo Scientific Prisma with ChemiSEM Technology.
      Video Player is loading.
      Current Time 0:00
      Duration 2:12
      Loaded: 7.44%
      Stream Type LIVE
      Remaining Time 2:12
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        BioLab Axia Customer Testimonial
        Video Player is loading.
        Current Time 0:00
        Duration 2:43
        Loaded: 6.01%
        Stream Type LIVE
        Remaining Time 2:43
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Video Player is loading.
          Current Time 0:00
          Duration 1:44
          Loaded: 9.38%
          Stream Type LIVE
          Remaining Time 1:44
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Geological section
            Section of a geological sample acquired combining the live quantitative elemental information and the navigation montage feature. The navigation montage allows to collect multiple images over a wide area automatically (in this case 13x13 frames have been acquired). Thanks to the instantaneous elemental information provided by Axia ChemiSEM, the montage shows the chemical variability within the grains over a large thin section and permits the user to identify areas of interest for further analysis.
            Download our ebook, SEM and EDS Analysis in Industry, to learn more about:
            • How the joint application of SEM and EDS analysis supports in-depth material characterization for industrial applications
            • How our unique Thermo Scientific ChemiSEM Technology integrates these two techniques into a single, fast, easy-to-use package, which is accessible and valuable to novices and experts alike
            Download
            On-demand Webinar: Meet our newest SEM
            Attend this webinar to:
            • Discover zero-click elemental analysis that enhances accessibility and productivity with live quantitative elemental imaging.
            • Experience the future-proof flexibility of a large chamber, support of samples up to 10 kg, and enough ports to accommodate many accessories.
            • Find out how you can save time with fast pumping, automatic alignments, and an easy interface.
            Watch webinar
            Meet the Axia ChemiSEM System
            Meet the Axia ChemiSEM: a SEM EDS instrument that is flexible, easy to use, and offers instantaneous, quantitative elemental analysis.
            Video Player is loading.
            Current Time 0:00
            Duration 2:32
            Loaded: 6.44%
            Stream Type LIVE
            Remaining Time 2:32
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • en (Main), selected
                1. Now Playing
                  Up NextMeet the Axia ChemiSEM System
                  Meet the Axia ChemiSEM: a SEM EDS instrument that is flexible, easy to use, and offers instantaneous, quantitative elemental analysis.
                2. Now Playing
                  Up NextAlignment-free operation
                  Axia is always ready to image with an autofocus routine that is easier and quicker than ever. It comes with the ability to focus on the feature of choice with a simple click.
                3. Now Playing
                  Up NextZero-click elemental analysis-Linescan Brass
                  Reliable quantification can be obtained directly from the live image using a choice of standard EDS functions (region/point ID/line scan).
                4. Now Playing
                  Up NextAxia ChemiSEM’s geological section tooltip
                  With Axia ChemiSEM, quantitative elemental information is provided through colors in the image. Moreover, a tooltip shows the relative concentrations of the sample constituents, making it the quickest way to obtain composition information during live
                5. Now Playing
                  Up NextAxia ChemiSEM: two times faster than traditional SEMs.
                  Axia ChemiSEM provides live quantitative compositional information during routine SEM navigation and imaging, two times faster compared to traditional SEMs. With the possibility to easily select and unselect the elements of interest, you can focus on
                6. Now Playing
                  Up NextZero-click elemental analysis - conventional mapping
                  Reliable quantification can be obtained directly from the live image using a choice of standard EDS functions (region/point ID/line scan).
                7. Now Playing
                  Up NextConventional EDS functions in a state-of-the-art system
                  Conventional EDS functions are available in a state-of-the-art implementation, such as gross counts mapping, which allows to post-process your data by selecting, for each element, the energy line used to generate the elemental map.
                8. Now Playing
                  Up NextAxia ChemiSEM’s tooltip, the quickest way to obtain composition information during live imaging
                  Axia ChemiSEM provides live compositional information during routine SEM navigation and imaging. The tooltip shows the relative concentrations of the sample constituents, making it the quickest way to obtain composition information during live imagin
                9. Now Playing
                  Up NextZero-click elemental analysis - Geological Section
                  Reliable quantitative information of a specific point or region of interest can be obtained using traditional EDS functions, directly on the SEM image. Multi-point analysis allows for easy comparison of different features on the sample.
                10. Now Playing
                  Up NextBatteries Zero Click Analysis
                  With Axia ChemiSEM, elemental analysis is always on, so that compositional information is never more than a mouse click away. Conventional EDS functions operate directly on the image without intermediate steps.
              ChemiSEM Testimonial - Carlos Angeles Chavez
              Carlos Angeles Chavez from the Mexican Petroleum Institute comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
              Video Player is loading.
              Current Time 0:00
              Duration 1:48
              Loaded: 9.11%
              Stream Type LIVE
              Remaining Time 1:48
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                  1. Now Playing
                    Up NextChemiSEM Testimonial - Carlos Angeles Chavez
                    Carlos Angeles Chavez from the Mexican Petroleum Institute comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
                  2. Now Playing
                    Up NextChemiSEM Testimonial - Lifeng Dong
                    Lifeng Dong from Hamline University comments on his first impressions of his live demo with the Thermo Scientific™ Prisma E SEM with ChemiSEM technology.
                  3. Now Playing
                    Up NextChemiSEM Testimonial - Cedric Barroo
                    Cedric Barroo from the Universite Libre de Bruxelles comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
                  4. Now Playing
                    Up NextChemiSEM Testimonial - Houston Electron Microscopy
                    Dennis and Marina Manuel comment on the time saving and productivity benefits that they experienced in their live demonstration of the Thermo Scientific Prisma with ChemiSEM Technology.
                Video Player is loading.
                Current Time 0:00
                Duration 2:12
                Loaded: 7.44%
                Stream Type LIVE
                Remaining Time 2:12
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  BioLab Axia Customer Testimonial
                  Video Player is loading.
                  Current Time 0:00
                  Duration 2:43
                  Loaded: 6.01%
                  Stream Type LIVE
                  Remaining Time 2:43
                  1x
                    • Chapters
                    • descriptions off, selected
                    • captions off, selected
                    • en (Main), selected
                    Video Player is loading.
                    Current Time 0:00
                    Duration 1:44
                    Loaded: 9.38%
                    Stream Type LIVE
                    Remaining Time 1:44
                    1x
                      • Chapters
                      • descriptions off, selected
                      • captions off, selected
                      • en (Main), selected
                      Geological section
                      Section of a geological sample acquired combining the live quantitative elemental information and the navigation montage feature. The navigation montage allows to collect multiple images over a wide area automatically (in this case 13x13 frames have been acquired). Thanks to the instantaneous elemental information provided by Axia ChemiSEM, the montage shows the chemical variability within the grains over a large thin section and permits the user to identify areas of interest for further analysis.
                      Download our ebook, SEM and EDS Analysis in Industry, to learn more about:
                      • How the joint application of SEM and EDS analysis supports in-depth material characterization for industrial applications
                      • How our unique Thermo Scientific ChemiSEM Technology integrates these two techniques into a single, fast, easy-to-use package, which is accessible and valuable to novices and experts alike
                      Download
                      Applications
                      Process control using electron microscopy
                      Process control using electron microscopy
                      Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
                      Quality control and failure analysis using electron microscopy
                      Quality control and failure analysis
                      Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
                      Fundamental Materials Research_R&D_Thumb_274x180_144DPI
                      Fundamental Materials Research
                      Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
                      Techniques
                      EDS Analysis with ChemiSEM Technology
                      Energy dispersive X-ray spectroscopy for materials characterization.
                      Learn more ›
                      Imaging Hot Samples
                      Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
                      Learn more ›
                      Particle analysis
                      Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
                      Learn more ›
                      Cathodoluminescence
                      Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.
                      Learn more ›
                      EDS Analysis with ChemiSEM Technology
                      Energy dispersive X-ray spectroscopy for materials characterization.
                      Learn more ›
                      Imaging Hot Samples
                      Studying materials in real-world conditions often involves working at high temperatures. The behavior of materials as they recrystallize, melt, deform, or react in the presence of heat can be studied in situ with scanning electron microscopy or DualBeam tools.
                      Learn more ›
                      Particle analysis
                      Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
                      Learn more ›
                      Cathodoluminescence
                      Cathodoluminescence (CL) describes the emission of light from a material when it is excited by an electron beam. This signal, captured by a specialized CL detector, carries information on the sample’s composition, crystal defects, or photonic properties.