Search
Fetal Bovine Serum (FBS) for Cell Culture |
Fetal bovine serum (FBS), also known as fetal calf serum, offers essential nutrients and growth factors for the maintenance and growth of cultured cells. Cell-based applications are an integral part of basic research, biochemical studies, vaccine development, and drug discovery to name a few. FBS is the most used serum in cell-based applications as it provides the most robust culture system for the widest range of cell types.
Since 1962, Gibco FBS products have been a global market leader. Through Thermo Fisher Scientific’s unwavering commitment to quality, scientists using Gibco FBS have been provided with consistent reliability, service, value, and innovation. Discover the performance differences that Gibco Fetal Bovine Serum can deliver for your research and production.
Learn more about the various market scenarios that drive FBS availability and price
We provide a simplified three-tiered offering—Gibco Value FBS, Premium FBS, and Premium Plus FBS—where each category is clearly delineated by relevant performance markers and testing levels to help ensure you can confidently select the right serum for your research.
Designed for consistent growth for robust cell lines used within all standard cell culture research applications. Our Value FBS products offer excellent value with up to 30 quality specification tests.
Designed for consistent growth for a broad range of cells. Our most popular FBS product, Premium FBS offers high quality and exceptional value with up to 60 quality specification tests.
Offers consistent growth for sensitive cells. Our highest quality FBS with up to 70 quality specification tests, including lowest levels of endotoxin and hemoglobin release specifications.
Designed for specific experimental needs and sensitive cell culture, including stem cell research, cancer research, reporter assays, immunoassays, antibodies, and more.
Specifications, not origin, help drive consistent performance. We commissioned an independent research group to survey more than 500 researchers around the globe. This is what we learned:
Based on these findings, we transformed our Gibco FBS portfolio into categories focused on quality indicators rather than origin; Value FBS, Premium FBS, and Premium Plus FBS. Now you can easily select the best sera for your needs based on how they perform, not where they came from. This also enables us to minimize supply disruption since there is less reliance on origin.
| Quality profile | Value FBS | Premium FBS | Premium Plus FBS |
|---|---|---|---|
| Endotoxin | ≤20 EU/mL | ≤10 EU/mL | ≤5 EU/mL |
| Haemoglobin/hemoglobin | ≤25 mg/dL | ≤25 mg/dL | ≤20 mg/dL |
| Sterility: bacterial and fungal testing | ✓ | ✓ | ✓ |
| Osmolality | ✓ | ✓ | ✓ |
| pH | ✓ | ✓ | ✓ |
| Mycoplasma: supplemental testing (H-stain) | ✓ | ✓ | ✓ |
| Mycoplasma | ✓ | ✓ | ✓ |
| Country of origin | ✓ | ✓ | ✓ |
| Oritain™ testing (origin confirmation) | – | ✓ | ✓ |
| Biochemical and hormonal profile | |||
| Biochemical: Alkaline phosphatase, ALT, AST, bicarbonate, bilirubin (total), BUN, BUN/creatinine ratio, calcium, chloride, cholesterol, creatinine, GGPT, glucose, HDL, iron, iron saturation, LDH, LDL, phosphorous (inorganic), potassium, sodium, TIBC, triglycerides (TG), uric acid | – | ✓ | ✓ |
| Hormonal: Estradiol, insulin, progesterone, testosterone, thyroxine (T4) | – | ✓ | ✓ |
| Virus testing | |||
| Serology: BVDV serum neutralization titer (Ab) | – | ✓ | ✓ |
| Serology: Anti-BVDV antibody test (EMA lots only) | – | ✓ | ✓ |
| Virus (9CFR): Bluetongue Virus, Bovine Adenovirus, Bovine Parvovirus, Bovine Viral Diarrhoea Virus, Bovine Respiratory Syncytial Virus (BRSV), Rabies Virus, Reo Virus Cytopathogenic Agents - Including Bovine Herpesvirus 1 (BHV-1/IBR) Haemadsorbing Agents - Including Bovine Parainfluenza Virus 3 (PI-3) | ✓ | ✓ | ✓ |
| Virus (EMA): Bluetongue Virus, Bovine Adenovirus, Bovine Parvovirus, Bovine Viral Diarrhoea Virus, Bovine Respiratory Syncytial Virus (BRSV), Rabies Virus, Reo Virus Cytopathogenic Agents- Including Bovine Herpesvirus 1 (BHV-1/IBR) Haemadsorbing Agents- Including Bovine Parainfluenza Virus 3 (PI-3) | – | ✓ | ✓ |
| USDA safety testing: Bluetongue virus (Mexican and Australian origin*) | ✓ | ✓ | ✓ |
| USDA safety testing: Akabane virus (Australian only*) | ✓ | ✓ | ✓ |
| Protein electrophoresis | |||
| Identification:Electrophoretic profile | ✓ | ✓ | ✓ |
| Identification: Bovine gamma globulin (≤500 mg/L) | ✓ | ✓ | ✓ |
| Protein: Albumin, alpha globulin, beta globulin, total protein | ✓ | ✓ | ✓ |
| Performance testing | |||
| Relative growth promotion (RGP), relative cloning efficiency (RCE), relative plating efficiency (RPE) | ✓ | ✓ | ✓ |
| Documentation | |||
| Certificate of Suitability (TSE CEP) | – | ✓ | ✓ |
| Regulatory | |||
FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin; please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support. | ✓ | ✓ | ✓ |
| *US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US. | ✓ | ✓ | ✓ |
| Important to note also that our FBS is sourced from BSE-negligible countries. | ✓ | ✓ | ✓ |
Choose One Shot FBS in convenient 50 mL bottles to end aliquoting, stop the freeze-thaw cycle, and help minimize contamination risk.
Choose heat-inactivated FBS for immunological applications requiring inactivation of complement proteins.
Choose gamma-irradiated FBS when your research requires the highest level of biosecurity, ensuring safer and more consistent outcomes.
Watch these videos to learn how FBS performance is based on science, not origin. Explore customer stories, FBS FAQs, how-to explanations, and product innovations.
Fetal bovine serum (FBS) is a byproduct of harvesting cattle for the meatpacking industry–it offers essential growth factors for the maintenance and growth of cultured cells. FBS is used as a supplement to basal growth medium in cell culture applications.
Extraordinary measures are taken during the manufacturing and production of Gibco FBS, with careful consideration taken in each step of the journey from farm to bench.
Did you know there are lower-cost alternatives to fetal bovine serum? These products include bovine serum, horse serum, newborn calf serum, goat serum, rabbit serum, porcine serum, and chicken serum.
Gibco sera for bioprocessing can support your cell culture needs as you scale up production. As one of the largest serum suppliers from BSE-negligible risk regions, Gibco serum can help meet biopharmaceutical manufacturing requirements.
Learn more about how we’ve become a world leader for sera, media, and reagents.
Discover the important factors to consider in FBS selection and learn how scientists are utilizing the benefits of FBS in their research.
We have dedicated regional sales representatives to assist you in all things FBS. Connect directly with a FBS representative today!
Have questions about using FBS in your cell culture experiments? Find the answers you need in this extensive FAQ database.
We are dedicated to operational excellence—from how we manage our core quality processes to our risk management systems. Collaborate with Thermo Fisher Scientific for harmonized quality management, multi-site ISO certification, and robust controls designed to help safeguard your cell culture and cell therapy programs.
This detailed guide helps you find recycling programs and manage the end-of-use phase of your research products, including Gibco bottles. Where local recycling infrastructure exists, clean, uncontaminated Gibco bottles and caps may be recyclable through conventional plastic recycling streams aligned to their resin classification.
Keep your cells healthy while helping to reduce your lab’s environmental impact with greener cell culture products. Gibco, Thermo Scientific Nunc, and Invitrogen products can help advance sustainability in your lab by limiting the use of hazardous chemicals, reducing waste and material consumption, and increasing energy efficiency.
Fetal Bovine Serum Basics
Learn the basics of FBS for cell culture, including information on the FBS uses, components, and the market dynamics driving this industry.
Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Gibco iMatch Sera Lot Matching Tool
Provide your previous Gibco FBS lot number or answer a few questions to find your ideal serum match.
Find your regional sales representative
Connect with a regional FBS sales representative.
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support?
Contact our expert team for technical and application support of laboratory products.
For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.