Conceptual art of a bottle of Gibco dialyzed FBS over a cell
Dialyzed FBS is manufactured using a process that significantly reduces the concentrations of small (low molecular weight) molecules such as amino acids, hormones, and cytokines while avoiding the precipitation of serum proteins.
Dialyzed fetal bovine serum is recommended for special applications where the normal concentrations of small molecules in FBS may be of concern, for example, in studies requiring higher levels of control over certain small molecules in the cell culture environment.
Dialyzed FBS vs FBS
Whereas standard FBS will contain higher concentrations of small molecules (e.g., amino acids, hormones, cytokines), the careful filtration process used to produce dialyzed FBS greatly reduces the concentrations of these molecules, facilitating more precise control of nutrients and cell signaling molecules in the cell culture medium.
Why researchers choose dialyzed FBS
Watch to discover how the depletion of small molecules in dialyzed FBS allows researchers to control for molecules of interest in their cell culture experiments.
Video Player is loading.
Current Time 0:00
Duration 3:05
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:05
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Features of Gibco dialyzed fetal bovine serum
        • Dialyzed by tangential flow filtration (MW 10,000 cut-off)
        • Dialyzed against 0.15 M NaCl until glucose <5 mg/dL
        • Performance tested for cloning and plating efficiency
        • Available in the innovatively designed, aliquot-free Gibco One Shot FBS 50 mL bottle
        Artistic rendering of a spheroid of GFP-expressing cells
        Research areas using dialyzed FBS*
        • Proteomics (mass spectrometry)
        • Mitochondrial function studies
        • Glucose/amino acid deprivation studies
        • Isotope labeling
        • Cellular signaling and reporter assays
        * These results are based on a review of approximately 10,000 publications using query terms based on the six specialty FBS products offered by Thermo Fisher Scientific. These terms were generated by the MeSH (medical subject headings) taxonomy based on the full text of the paper.
        Order Gibco dialyzed FBS
        Publications using Gibco dialyzed FBS
        Resources
        Fetal Bovine Serum Basics
        Learn the basics of FBS for cell culture, including information on the FBS uses, components, and the market dynamics driving this industry.
        Cell Culture Basics
        Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
        Gibco iMatch Sera Lot Matching Tool 
        Provide your previous Gibco FBS lot number or answer a few questions to find your ideal serum match.
        Related products
        Support
        Find your regional sales representative
        Connect with a regional FBS sales representative.
        Cell Culture & Transfection Support Center
        Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
        Need technical support?
        Contact our expert team for technical and application support of laboratory products.
        For Research Use or Further Manufacturing Use only. Serum and blood proteins are not for direct administration into humans or animals.
        Stylesheet for Classic Wide Template adjustments