Muntjac fibroblast labeled with probes for actin and the nucleus.
Muntjac fibroblast labeled with probes for actin and the nucleus. The actin cytoskeleton was visualized using the green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379). The nucleus was stained with TO-PRO®-3 reagent (Cat. no. T3605).
Muntjac fibroblast labeled with probes for actin and the nucleus. The actin cytoskeleton was visualized using the green-fluorescent Alexa Fluor® 488 phalloidin (Cat. no. A12379). The nucleus was stained with TO-PRO®-3 reagent (Cat. no. T3605).
Related Products
Catalog # Name Size Price (USD) Qty
A12379 Alexa Fluor™ 488 Phalloidin, Green, 495/518 nm Each
644.65
Online Exclusive
664.00
Save 19.35 (3%)
T3605 TO-PRO™-3 Iodide (642/661), 1 mM Solution in DMSO, 1 mL 1 mL
472.65
Online Exclusive
487.00
Save 14.35 (3%)
Related Images