693f - Key/Specs/Resources/Samples/Tech/Doc/Contact
K-Alpha X-ray Photoelectron Spectrometer
The Thermo Scientific K-Alpha X-ray Photoelectron Spectrometer (XPS) System brings a new approach to surface analysis. Focused on delivering high-quality results using a streamlined workflow, the K-Alpha XPS System makes XPS operation simple and intuitive, with no sacrifice in terms of performance or capabilities.  
State-of-the-art performance, reduced cost of ownership, increased ease of use, and high sample throughput make the K-Alpha XPS System ideal for a multi-user environment. The K-Alpha XPS System gives more researchers around the world access to surface analysis. 
Video Player is loading.
Current Time 0:00
Duration 9:26
Loaded: 1.75%
Stream Type LIVE
Remaining Time 9:26
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Webinar: Thermo Scientific K-Alpha XPS System
    Style Sheet to change H2 style to p with em-h2-header class
    K-Alpha X-ray Photoelectron Spectrometer features
    High-performance X-ray source
    High-performance X-ray source
    The X-ray monochromator allows selection of analysis area from 50 µm to 400 µm in 5 µm steps, fitting it to the feature of interest to maximize the signal.
    Optimized electron optics
    Optimized electron optics
    The high-efficiency electron lens, hemispherical analyzer, and detector allow for superb detectability and rapid data acquisition. 
    Sample viewing
    Sample viewing
    Bring sample features into focus with the K-Alpha XPS System's patented optical viewing system and XPS SnapMap, which helps you pinpoint areas of interest quickly.
    Insulator analysis
    Insulator analysis
    The patented dual-beam flood source couples low-energy ion beams with very low energy electrons (less than 1 eV) to prevent sample charging during analysis, which eliminates the need, in most cases, for charge referencing.
    Depth profiling
    Depth profiling
    Go beyond the surface with the EX06 ion source. Automated source optimization and gas handling ensure excellent performance and experimental reproducibility.
    Digital Control
    Digital Control
    Intuitive operation—guided by the Avantage data system—makes the K-Alpha XPS System ideal for both multi-user, shared facilities and XPS experts who place a premium on efficient operation and high-throughput analysis.
    Optional sample holders
    Optional sample holders
    Specialist sample holders for angle-resolved XPS, sample bias measurements, or for inert transfer from a glove box are available.
    Specifications
    Style Sheet for Products Table Specifications
    Analyzer type 
    • 180° double-focusing hemispherical analyzer, 128-channel detector
    X-ray source type
    • Monochromated, micro-focused, low-power Al K-Alpha X-ray source 
    X-ray spot size
    • 50–400 µm (adjustable in 5 µm steps)
    Depth profiling
    • EX06 ion source
    Maximum Sample area
    • 60x60 mm
    Maximum sample thickness
    • 20 mm
    Vacuum system
    • Two turbo molecular pumps, with automated titanium sublimation pump and backing pump
    Optional accessories
    • ADXPS sample holder, work function sample holder, glove box
    Style Sheet to change H2 style to p with em-h2-header class
    Style Sheet for Komodo Tabs
    Resources
    Thermo Fisher Scientific Avantage Datasystem
    The XPS knowledge view is a powerful reference tool included within Avantage Datasystem
    Video Player is loading.
    Current Time 0:00
    Duration 2:19
    Loaded: 7.11%
    Stream Type LIVE
    Remaining Time 2:19
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
        1. Now Playing
          Up NextThermo Fisher Scientific Avantage Datasystem
          The XPS knowledge view is a powerful reference tool included within Avantage Datasystem
        2. Now Playing
          Up Next50 years of XPS
          Discover how XPS has impacted research over the last 50 years.
        3. Now Playing
          Up NextWebinar: Thermo Scientific K-Alpha XPS System
          The K-Alpha XPS System is a high-performance X-ray Photoelectron Spectrometer, designed to be fast and easy to use for a high-throughput workflow. The K-Alpha XPS System is supplied with everything necessary for XPS analysis, including an ion source
        4. Now Playing
          Up NextXPS Overview with XPS Facility Manager Robert Palgrave
          Mr. Palgrave of University College London goes over analyzing solid-state and thin film materials using XPS.
      Thermo Fisher Scientific Avantage Datasystem
      The XPS knowledge view is a powerful reference tool included within Avantage Datasystem
      Video Player is loading.
      Current Time 0:00
      Duration 2:19
      Loaded: 7.11%
      Stream Type LIVE
      Remaining Time 2:19
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
          1. Now Playing
            Up NextThermo Fisher Scientific Avantage Datasystem
            The XPS knowledge view is a powerful reference tool included within Avantage Datasystem
          2. Now Playing
            Up Next50 years of XPS
            Discover how XPS has impacted research over the last 50 years.
          3. Now Playing
            Up NextWebinar: Thermo Scientific K-Alpha XPS System
            The K-Alpha XPS System is a high-performance X-ray Photoelectron Spectrometer, designed to be fast and easy to use for a high-throughput workflow. The K-Alpha XPS System is supplied with everything necessary for XPS analysis, including an ion source
          4. Now Playing
            Up NextXPS Overview with XPS Facility Manager Robert Palgrave
            Mr. Palgrave of University College London goes over analyzing solid-state and thin film materials using XPS.
        Samples
        Battery Research
        Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
        Learn more ›
        Metals Research and Characterization
        Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
        Learn more ›
        Polymers Research
        Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
        Learn more ›
        Geological Research
        Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
        Learn more ›
        Oil and Gas
        As the demand for oil and gas continues, there is an ongoing need for efficient and effective extraction of hydrocarbons. Thermo Fisher Scientific offers a range of microscopy and spectroscopy solutions for a variety of petroleum science applications.
        Learn more ›
        Nanoparticles
        Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
        Learn more ›
        Forensics
        Micro-traces of crime scene evidence can be analyzed and compared using electron microscopy as part of a forensic investigation. Compatible samples include glass and paint fragments, tool marks, drugs, explosives, and GSR (gunshot residue).
        Learn more ›
        Catalysis Research
        Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
        Learn more ›
        Fibers and Filters
        The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
        Learn more ›
        2D Materials
        Novel materials research is increasingly interested in the structure of low-dimensional materials. Scanning transmission electron microscopy with probe correction and monochromation allows for high-resolution two-dimensional materials imaging.
        Learn more ›
        Automotive Materials Testing
        Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
        Learn more ›
        Style Sheet to change Sample H3 to p with em-h3-header class
        Style Sheet to change H3 to p with em-h3-header class
        Techniques
        X-Ray Photoelectron Spectroscopy
        X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
        Learn more ›
        X-Ray Photoelectron Spectroscopy
        X-ray photoelectron spectroscopy (XPS) enables surface analysis, providing elemental composition as well as the chemical and electronic state of the top 10 nm of a material. With depth profiling, XPS analysis extends to compositional insight of layers.
        Learn more ›
        Contact us
        Style Sheet for Support and Service footer
        Style Sheet for Fonts
        Style Sheet for Cards