Exceptional features of the EyeCGas Multi OGI Camera
The EyeCGas® Multi Optical Gas Imaging (OGI) Camera is designed to deliver remarkable performance and safety for the oil and gas industry. With excellent connectivity, intrinsic safety certifications, and exceptional sensitivity for gas leak detection, this camera sets the standard for optical gas imaging. Its LDAR-ready capabilities, versatile thermographic imaging, and built-in gas quantification help to ensure comprehensive usability and reliability. Explore the advanced features that make the EyeCGas Multi OGI Camera a leading choice for environmental monitoring and industrial safety.
Connectivity
Connectivity icon
Better connectivity – With exceptional connectivity, the EyeCGas Multi OGI Camera offers video streaming and downloading via WiFi, and Bluetooth integration with 3rd party software and devices.
Intrinsically safe
Ex icon
Better safety – An intrinsically safe camera for OGI, the EyeCGas Multi is designed explicitly for the oil and gas industry’s harsh environments. It is certified to meet IEC standards for temperature, humidity, vibration, dust, and IP65 water & dust protection. It is also certified for use in ANSI/ISA-Class I and II, Div. 2, & Class III, CSA C22.2 Class I, Div. 2 and ATEX II, IECEx EMT 19.0006X hazardous environments.
Gas leak detection
Gas leak icon
Higher sensitivity – With NETD of <10mK, the EyeCGas Multi is a highly sensitive OGI camera. It can detect leaks as small as 0.35 g/hr (methane) and is fully certified to meet EPA (NSPS) 40 CFR part 60, subpart OOOOa/b/c requirements.
LDAR-ready capabilities
LDAR Icon
Better usability – The EyeCGas Multi OGI Camera has smart LDAR operations offering wireless integration with leading LDAR software and quantification devices, allowing real-time component tag synchronization and a simple association between components, images, and videos.
Thermographic imaging
Thermographic icon
More versatility – Thermography mode enables accurate temperature measurement, six color palettes, and an option for easy export of radiometric data.
Gas quantification
Gas level icon
Built-in quantification –Concentration as well as mass flow of various VOC emissions can be easily measured in real time using a user-friendly interface.
More features
  • Multi-spectral filters – The EyeCGas Multi OGI Camera offers three operator replaceable filters: for VOC gases 3.2μm to 3.4μm; for heavier alkanes (better for long-range detection) 3.3μm to 3.5μm; and for CO2 4.2μm to 4.4μm.
  • 4-year warranty – This ruggedized camera was built to last, with an extended warranty.
  • Free firmware updates –Camera software upgrades and improvements regularly provided free of charge.
  • Instrument backward compatibility to enable easy upgrades from older versions of EyeCGas cameras to the EyeCGas Multi OGI Camera.
  • Integrated 64 Gb solid-state memory, snapshot button, high-definition color camera, more operability.
  • Real-time component label synchronization
  • Built-in geotagging
  • Integrated with multiple technologies such as PIF/FID Method 21 analyzer data to realize intelligent LDAR.
Certifications
  • ANSI/ISA-12.12.01-Nonincendive Electrical Equipment for Use in Class I and II, Division 2, and Class III Hazardous (Classified) Locations
  • CSA C22.2 No. 213-M1987, First Edition: Non-Incendive Electrical Equipment for Use in Class I, Division 2 Hazardous Locations.
  • IECEx EMT 19.0006X as Intrinsic Safety “ic”, protection “n”
  • EMT19ATEX0009X
  • Ex II 3 GD
  • Ex ic nA nC IIC T6 Gc Tamb = -20 °C to +50 °C
  • Ex ic IIIC T85 °C Dc    Tamb = -20 °C to +50 °C
Service and product availability may vary by country and are subjected to varying regulatory requirements. Please contact your local sales representative for availability.