ExoSAP-IT_PCR_Product_Cleanup_Reagent
High-quality, accurate results, faster turnaround time
When it comes to achieving a clean template for your Sanger sequencing reaction, without losing precious samples, our sequencing ExoSAP-IT PCR product cleanup reagent delivers time and time again. Whether you’re looking for faster turnaround times or high-throughput, you can rely on our DNA sequencing PCR cleanup regaents to deliver the data quality and sequencing accuracy you need to move forward with complete peace of mind.
Before the PCR product is ready for Sanger sequencing, you must undertake a PCR cleanup of your reaction mixtures to prevent unincorporated primers and dNTPs from interfering with your results. If you overlook this vital step, you risk compromising the Sanger sequencing process and your abililty to effectively read DNA sequence base by base.
Video Player is loading.
Current Time 0:00
Duration 3:08
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:08
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn more: watch our Seq it Out video
    ExoSAP_IT_Reagents
    Brochure: ExoSAP-IT PCR cleanup reagents
    Learn more about how our ExoSAP-IT PCR Product Cleanup Reagent ensures quality sequencing results in a fraction of the time.
    Download brochure
    By offering a straightforward protocol and 100% recovery of both short and long amplicons, ExoSAP-IT PCR product cleanup reagent serves as a trusted tool in the researcher’s kit, allowing finite samples to be sufficiently conserved and productivity and process to be fine-tuned as part of a single, end-to-end workflow.
    ExoSAP-IT_PCR_Product_Cleanup_Reagent
    Application note: Use of ExoSAP-IT PCR Product Cleanup Reagent in NGS
    While Sanger sequencing–based methods remain popular for validation and long contiguous DNA sequence reads (>500 nucleotides), many genomic analysis applications are transitioning to next-generation sequencing (NGS) technology for its scalability and affordability for sequencing a large number of targets.
    Download application note ›
    ExoSAP-IT Express—one tube, one-step
    Accelerate result accuracy with confidence with ExoSAP-IT Express reagent. Our one-step solution that delivers higher yields and full PCR product recovery.                
    • Five-minute protocol—fast turnaround time
    • One-tube, one-step PCR cleanup— add reagent directly to PCR product
    • Novel enzyme technology— irreversibly inactivated in just one minute at 80°C
    • Complete sample recovery—100% recovery of PCR products, regardless of amplicon length
    • Eliminates spin columns or beads—helps to significantly decrease time and errors while increasing yield
    • Compatible—no interference in downstream applications
    • Scalable—treat a wide range of volumes
    • Multiple formats—single-tube, 8-tube strip, and 96-well plate formats offer flexibility for manual or automated workflows
    • Optional tracking dye—helps to avoid losing track of reagent addition during pipetting
    • Consolidated ordering from one trusted partner
    Purchase
    ExoSAP-IT_PCR_Product_Cleanup_Reagent
    Applied Biosystems ExoSAP-IT Express reagents are proprietary mixtures of exonuclease I combined with shrimp alkaline phosphatase (SAP) in a specially formulated buffer. Informed by decades of results, these reagents expertly remove excess primers and dNTPs following PCR. Exonuclease I removes residual single-stranded primers and any extraneous single-stranded DNA produced during PCR, while SAP removes the remaining dNTPs from the PCR mixture that may inhibit subsequent reactions.
    ExoSAP-IT_PCR product_cleanup_reagent
    Figure 1. Enzymatic cleanup of PCR reaction with ExoSAP-IT Express reagent. Treat 5 μL of PCR product with 2 μL of ExoSAP-IT Express reagent. Treatment is carried out at 37°C for 4 minutes followed by an incubation period at 80°C for 1 minute to irreversibly inactivate both enzymes. Once enzyme inactivation is complete, your PCR products are ready for downstream applications such as sequencing (Sanger/NGS), fragment analysis, SNP analysis, in vitro transcription, or single base extension.
    Adding ExoSAP-IT Express reagent directly to the PCR product eliminates transfer steps to tubes, wells, or columns and conserves amplicons, helping to reduce the chance of cross-contamination; no further processing is needed. After enzymatic cleanup with ExoSAP-IT Express reagent, primers and dNTPs will no longer interfere with DNA sequencing or other downstream applications.
    ExoSAP-IT_PCR_product_cleanup_reagent
    Product Spotlight: ExoSAP-IT Express PCR Cleanup Agent
    In only five minutes, ExoSAP-IT Express PCR Cleanup Agent removes excess primers and unincorporated nucleotides from a PCR reaction. This enzymatic cleanup method offers precision, increased yield, and a faster turnaround time compared to alternative purification methods, such as spin columns or beads.
    Download product spotlight ›
    ExoSAP-IT Express reagent offers rapid turnaround times and improved efficiency of resource use while delivering the same superior cleanup as the original ExoSAP-IT reagent. Eliminate spin columns, magnetic beads, sedimentation, filtration, and gel purifications—the novel ExoSAP-IT technology allows for a significant reduction in sample cleanup time with minimal steps, providing the simplest workflow (see figure).
    ExoSAP-IT Express reagent vs. other common methods
    Figure 2. ExoSAP-IT Express reagent vs. other common methods.
    Complete ExoSAP-IT PCR product cleanup portfolio—comprehensive solutions
    ExoSAP-IT reagents offer a unique one-tube, one-step enzymatic method for PCR cleanup. All ExoSAP-IT reagents provide 100% recovery of PCR products regardless of the fragment sizes. This method removes excess primers and dNTPs and does not interfere with downstream applications. No matter the parmeters of your project, our suite of PCR cleanup solutions deliver the flexibility and agility you need to optimize your workflow with confidence. Use our selection guide to determine which formulation is best for your next experiment.
    Whether you are looking for a fast turnaround time or high-throughput solution, we have a PCR cleanup reagent that will work for you. Use our selection guide to determine which formulation is best for your next experiment.
     ExoSAP-IT Express reagentExoSAP-IT reagent (original formulation)HT ExoSAP-IT Fast High-Throughput reagent
    Protocol time5 minutes30 minutes14 minutes
    Format
    • Single tube
    • 8-tube strips
    Single tube
    • Single tube
    • 8-tube strips
    • 96-well plate
    Throughput levelLow to high; recommended for processing any sample sizeLow to medium; recommended for processing 1–96 samples at a timeHigh; recommended for processing ≥96 samples at a time
    PlatformSingle- or multichannel pipette, automated liquid handling platforms (lower viscosity for better handling)Single-channel pipetteAutomated liquid handling platforms (lower viscosity for better handling)
    Freezes at
    –20°C
    NoNoYes
    Stability–20°C for up to 2 years–20°C for up to 2 years–20°C for up to 2 years; once thawed, stable at 4°C for 1 month and at room temperature for 2 days
     PurchasePurchasePurchase
    Comparison to alternative PCR cleanup methods
    Learn more about the superior workflow advantages of enzymatic purification technology.
    ExoSAP_IT_Express_PCR_Product_Cleanup_Comparison
    Comparison of ExoSAP-IT and ExoSAP-IT Express reagents to alternative PCR cleanup methods
    ExoSAP-IT and ExoSAP-IT Express PCR cleanup reagents offer significant workflow advantages compared to magnetic beads or spin columns, while achieving similar results.
    Download white paper ›
    ExoSAP_IT_Express_PCR_Product_Cleanup_Comparison
    Comparison of CleanSweep and ExoSAP-IT PCR clean-up agents
    A side-by-side comparison of CleanSweep and ExoSAP-IT reagents that demonstrates ExoSAP-IT effectiveness and functional equivalency.
    Download white paper ›
    Catalog # Name Size Price (USD) Qty
    78201.1.ML ExoSAP-IT™ PCR Product Cleanup Reagent, 1 mL Each
    593.65
    Online Exclusive
    626.00
    Save 32.35 (5%)
    78205.10.ML ExoSAP-IT™ PCR Product Cleanup Reagent, 10 mL Each
    4,901.65
    Online Exclusive
    5,170.00
    Save 268.35 (5%)
    78202.4X.1.ML ExoSAP-IT™ PCR Product Cleanup Reagent, 4 x 1 mL Each
    2,128.65
    Online Exclusive
    2,245.00
    Save 116.35 (5%)
    78200.200.UL ExoSAP-IT™ PCR Product Cleanup Reagent, 200 μL Each
    151.65
    Online Exclusive
    160.00
    Save 8.35 (5%)
    75001.40.UL ExoSAP-IT™ Express PCR Product Cleanup, 40 μL Each
    60.65
    Online Exclusive
    63.50
    Save 2.85 (4%)
    75001.4X.1.ML ExoSAP-IT™ Express PCR Product Cleanup Reagent, 2000 reactions Each
    2,313.65
    Online Exclusive
    2,435.00
    Save 121.35 (5%)
    75001.1.EA ExoSAP-IT™ Express PCR Product Cleanup Reagent, 480 reactions Each
    619.65
    Online Exclusive
    646.00
    Save 26.35 (4%)
    75001.10.ML ExoSAP-IT™ Express PCR Product Cleanup Reagent, 5000 reactions Each
    5,248.65
    Online Exclusive
    5,490.00
    Save 241.35 (4%)
    75001.200.UL ExoSAP-IT™ Express PCR Product Cleanup Reagent, 100 reactions Each
    165.65
    Online Exclusive
    172.00
    Save 6.35 (4%)
    75001.1.ML ExoSAP-IT™ Express PCR Product Cleanup Reagent, 500 reactions Each
    648.65
    Online Exclusive
    676.00
    Save 27.35 (4%)
    Build your knowledge
    Build your knowledge
    Get bite-sized answers to your everyday Sanger sequencing and fragment analysis questions
    Learn about the history of sequencing and how to pick the right platform for your research needs
    For Research Use Only. Not for use in diagnostic procedures.