Quickly, accurately, and automatically dose most 96-well microtiter plates
The Thermo Scientific Sensititre AIM Automated Inoculation Delivery System automates quick and accurate dosing of most 96-well plates, including Sensititre plates, for antimicrobial susceptibility testing (AST). Offering a small footprint and user-friendly design, the system streamlines plate setup, eliminates skipped wells, and avoids costly repeat tests. It also mitigates the risks of cross-contamination, staff exposure, and biohazardous spills linked to increased sample contact.
The Sensititre AIM System is easy to use. Just select dosing volume and pattern on the icon-driven touchscreen for simple, intuitive plate inoculation.
Features
The Sensititre AIM System shares common features with all components of the Sensititre AST System: It produces accurate, reliable, consistent results; it fits seamlessly into an integrated AST workflow including manual, semi-, and fully automated options; and customers receive the benefit of premiere Thermo Scientific support and service.
In addition, the Sensititre AIM system offers these additional features for plate inoculation.
Compatible with most 96-well microtiter plates
Ensures flexibility with dry or frozen AST formats.
Unique tube design and manually applied seal
Provide additional protection against biohazardous spills.
Download our app note on aerosol containment
Workflow
Sensititre_AIM_Workflow
Specifications
Technical characteristics
Test type: MIC
Regulatory status
FDA-cleared
Physical characteristics
Instrument dimensions: 
33.8 cm (W) x 31.2 cm (D) x 28.7 cm (H)
13.3 in (W) x 12.3 in (D) x 11.3 in (H)
Weight: 8 kg; 17.6 lb
Electrical
Power requirements: 100–240 VAC; 50–60 Hz
Power consumption: 150 watts
Ordering information
* Not all products are available for sale in all territories. Please inquire.