Clinical laboratory quality control software to simplify your QC revision
Thermo Scientific LabLink xL web-based quality control software allows for QC results to be monitored with peer-to-peer comparison and automated data entry. Easy to set up, Lablink xL can be used with Thermo Scientific MAS QC products or any other QC products.*
Efficient collaboration with your affiliated laboratories
Fast troubleshooting with global peer-to-peer comparison
Electronic signature for audit-friendly reports
Secured, long-term QC data storage with AWS™
Discover our advanced quality assurance software that streamlines troubleshooting, enhances confidence in patient results, and offers real-time peer comparison. Enjoy hands-free QC data transfers, customizable reports, and manage QC processes across multiple sites from one platform. Elevate your laboratory's performance and efficiency with LabLink xL.
ThermoFisher Scientific™ LabLink xL™ web-based quality assurance software offers statistical tools to help evaluate the performance of laboratory quality control methods and troubleshoot QC issues.
Gain real-time access to internal and peer data with Auto-Connectivity software for Thermo Scientific LabLink xL™ quality control software. Automatically capture quality control (QC) data directly from your instruments for fast access to performance metrics.
Seamless integration of QC results with automated data transfer
Automatically transfer quality control data from your instrument, laboratory information system (LIS), or middleware to LabLink xL quality control software, eliminating the need for manual entry and submission cutoff dates.
Stay audit-ready thanks to real-time comparison report generation
Receive easy-to-read reports automatically to your email inbox to help monitor lab performance by shift, day, and month. Reports can be reviewed and digitally signed with LabLink xL Signature Verification.
Evaluate your method performance to reduce QC runs
Lablink xL offers all the statistical tools laboratories need, including measurement of uncertainty and Six Sigma. Six Sigma will evaluate the performance of your methods and allow specific sets of Westgard rules to help reduce QC runs.
Watch: Improved quality control revision with LabLink xL laboratory quality control software
Video Player is loading.
Current Time 0:00
/
Duration 1:36
Loaded: 0%
0:00
Stream Type LIVE
Remaining Time -1:36
1x
Chapters
descriptions off, selected
captions settings, opens captions settings dialog
captions off, selected
en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
Simplify your QC data management now with LabLink xL clinical laboratory quality control software
Connect with an expert to learn more about implementing LabLink xL software in your lab. Already using LabLink xL? Login below.
*LabLink xL only requires an internet browser to view the interface. Some autoconnectivity options may require software installation. Non-MAS products are supported by our autoconnectivity and Daily QC features. Peer data for non-MAS products is not available at this time.