Green hydrogen processes
Green hydrogen is gaining momentum as a potential fuel and energy carrier of the future. During the production of green hydrogen by electrolysis, purity is of utmost importance so the accurate analysis of hydrogen and other gases is critical for the successful creation of this potentially clean energy fuel.
Thermo Scientific process mass spectrometers offer a robust range of features and are ideal tools for gas analysis in the production of green hydrogen, including:
  • Fast and precise real-time gas analysis (1 to 20 seconds per point) for accurate tracking of process dynamics
  • Dynamic ranges from ppm to 100%
  • Ultra-stable with >90 days interval between calibration
  • Fully automated and programmable analysis and calibration methods
  • Ultra-precise magnetic sector technology
  • Reliable, fault-tolerant design for availability of greater than 99.7%
  • Multi-stream inlet for measurement of up to 64 sample points
  • User friendly interface, no mass spectrometer experience required
  • Hydrogen & Oxygen purity and mixed gas streams on a single analyzer
  • Solutions for the laboratory and industrial plant
  • Worldwide support network from the industry leaders
  • Proven track record with >1,000 installations globally
Products
Learn more about Gas Analysis and Hydrogen Purity Solutions
Learn more about gas analysis and hydrogen production
Every plant, facility, and customer is uniquely different. Our applications specialists are ready to speak to you about how our gas analysis and continuous monitoring solutions meet your distinct needs.
Learn more about the Prima Pro
Scalable Solution
Scalable solution
To support efforts from R&D through to full production scale, we have magnetic sector mass spectrometry solutions in the Thermo Scientific Prima BT Bench Top Process Mass Spectrometer and the Thermo Scientific Prima PRO Process Mass Spectrometer.
Learn more about the Prima BT  Learn more about Prima PRO
MarqMetrix All-In-One Process Raman Analyzer
Improve process understanding with an in-line analysis technique
Real-time monitoring with Raman spectroscopy can be used as a complementary measurement technique for hydrogen production processes. The Thermo Scientific MarqMetrix All-In-One Process Raman Analyzer allows for real-time, non-destructive molecular composition analysis without the need for carrier gases, providing insights into the efficiency and purity of hydrogen production methods.
Learn more about the MarqMetrix All-In-One Process Raman Analyzer
Featured resources
Improving production of green hydrogen with fast, precise gas analysis Mass Spectroscopy
Green hydrogen production with fast, precise gas analysis Mass Spectroscopy
Learn more
Thermo Fisher Scientific and CPH2 team up to give renewable hydrogen production the green light
Renewable hydrogen production gets the green light
Learn more
Video spotlight
Video Player is loading.
Current Time 0:00
Duration 2:15
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:15
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Green Hydrogen Webinar
    Thermo Fisher Scientific and CPH2 team up to give renewable hydrogen production the green light
    Most developed nations have announced clean energy strategies to reduce their CO2 emissions, aiming to reach 'net zero' within the next few decades. The use of fossil fuels must soon become a thing of the past if this goal is to be realized, with alternative energy sources making up for the inevitable shortfall.
    Attend a virtual webinar with Thermo Fisher Scientific marketer Daniel Merriman and lab technician Bridie Haxby of Clean Power Hydrogen (CPH2) on demand to learn How online gas analysis mass spec helps electrolyzer developers increase their technology's efficacy and efficiency.
    Register
    Contact us
    *
    *
    *
    *
    *
    *
    *
    *
     Please confirm you would like to receive marketing and promotional email messages about Thermo Scientific products and services.
    This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
    Style Sheet for Global Design System