Immunocytochemistry using Alpha-Tubulin Monoclonal Antibody, Mouse
Binucleate bovine pulmonary artery endothelial cell. Anti–α-tubulin, biotin-XX conjugate; Alexa Fluor® 568 streptavidin and nuclear yellow. A binucleate bovine pulmonary artery endothelial cell labeled with the biotin-XX conjugate of anti–α-tubulin antibody (Cat. no. A21371) and Alexa Fluor® 568 streptavidin (Cat. no. S11226), then counterstained with nuclear yellow (Cat. no. N21485).
Binucleate bovine pulmonary artery endothelial cell. Anti–α-tubulin, biotin-XX conjugate; Alexa Fluor® 568 streptavidin and nuclear yellow. A binucleate bovine pulmonary artery endothelial cell labeled with the biotin-XX conjugate of anti–α-tubulin antibody (Cat. no. A21371) and Alexa Fluor® 568 streptavidin (Cat. no. S11226), then counterstained with nuclear yellow (Cat. no. N21485).
Related Products
Catalog # Name Size Price (USD) Qty
A21371 alpha Tubulin Monoclonal Antibody (236-10501), Biotin 50 µg
481.00
Related Images