Immunohistochemistry using GFAP Monoclonal Antibody, Mouse, Alexa Fluor® 594 Conjugate
Immunohistochemistry using NMDA Receptor Subunit 2A Polyclonal Antibody, Rabbit: Rat brain cryosections labeled with anti–NMDA receptor, subunit 2A (rat) antibody, rabbit IgG fraction (A6473) and detected using Alexa Fluor 488 goat anti–rabbit IgG antibody (A11008). The tissue was also labeled with Alexa Fluor 594 anti–glial fibrillary acidic protein antibody (A21295) and counterstained with TOTO-3 iodide (T3604), which was pseudocolored light blue in this image.
Immunohistochemistry using NMDA Receptor Subunit 2A Polyclonal Antibody, Rabbit: Rat brain cryosections labeled with anti–NMDA receptor, subunit 2A (rat) antibody, rabbit IgG fraction (A6473) and detected using Alexa Fluor 488 goat anti–rabbit IgG antibody (A11008). The tissue was also labeled with Alexa Fluor 594 anti–glial fibrillary acidic protein antibody (A21295) and counterstained with TOTO-3 iodide (T3604), which was pseudocolored light blue in this image.
Related Products
Catalog # Name Size Price (USD) Qty
A21295 GFAP Monoclonal Antibody (131-17719), Alexa Fluor™ 594 50 µL
516.00
A6473 NMDAR2A Polyclonal Antibody 10 µg
626.00
T3604 TOTO™-3 Iodide (642/660), 1 mM Solution in DMSO, 200 μL 200 µL
826.65
Online Exclusive
852.00
Save 25.35 (3%)
Related Images