c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
Focused ion beam scanning electron microscopy
The Thermo Scientific Scios 2 DualBeam is an ultra-high-resolution analytical focused ion beam scanning electron microscopy (FIB-SEM) system that provides outstanding sample preparation and 3D characterization performance for a wide range of samples, including magnetic and non-conductive materials. With innovative features designed to increase throughput, precision, and ease of use, the Scios 2 DualBeam is an ideal solution to meet the needs of scientists and engineers in advanced research and analysis across academic, governmental, and industrial research environments.
Subsurface characterization
Subsurface or three-dimensional characterization is often required to better understand the structure and properties of a sample. The Scios 2 DualBeam, with optional Thermo Scientific Auto Slice & View 4 (AS&V4) Software, allows for high-quality, fully automated acquisition of multi-modal 3D datasets, including backscattered electron (BSE) imaging for maximum materials contrast, energy-dispersive spectroscopy (EDS) for compositional information, and electron backscatter diffraction (EBSD) for microstructural and crystallographic information. Combined with Thermo Scientific Avizo Software, the Scios 2 DualBeam delivers a unique workflow solution for high-resolution, advanced 3D characterization and analysis at the nanometer scale.
Backscattered electron and secondary electron imaging
The innovative NICol electron column provides the foundation of the system’s high-resolution imaging and detection capabilities. It offers excellent nanoscale details, with a wide range of working conditions, whether operating at 30 keV in STEM mode (to access structural information) or at lower energies (to obtain charge-free, detailed surface information). With its unique in-lens Thermo Scientific Trinity Detection System, the Scios 2 DualBeam is designed for simultaneous acquisition of angular and energy-selective secondary electron (SE) and BSE imaging. Fast access to detailed nanoscale information is possible not only top-down, but also on tilted specimens or cross-sections. Optional below-the-lens detectors and an electron-beam-deceleration mode ensure fast and easy simultaneous collection of all signals, revealing the smallest features in a material surface or cross-section. Fast, accurate, and reproducible results are obtained thanks to the unique NICol column design with full auto alignments.
TEM sample preparation
Scientists and engineers constantly face new challenges that require highly localized characterization of increasingly complex samples with ever smaller features. The latest technological innovations of the Scios 2 DualBeam, in combination with the optional, easy-to-use, comprehensive Thermo Scientific AutoTEM 4 Software and Thermo Fisher Scientific’s application expertise, allow for fast and easy preparation of site-specific high-resolution S/TEM samples for a wide range of materials. In order to achieve high-quality results, final polishing with low-energy ions is required to minimize surface damage on the sample. The Thermo Scientific Sidewinder HT Focused Ion Beam (FIB) column not only delivers high-resolution imaging and milling at high voltages but also offers good low-voltage performance, enabling the creation of high-quality TEM lamella.
Key Features
Fast and easy preparation
High-quality, site-specific, TEM and atom probe samples using the Sidewinder HT ion column.
Ultra-high resolution imaging
Using the Thermo Scientific NICol Electron Column with best-in-class performance on the widest range of samples, including magnetic and nonconductive materials.
The most complete sample information
With sharp, refined, and charge-free contrast obtained from a variety of integrated in-column and below-the-lens detectors.
High-quality, multi-modal subsurface and 3D information
Access the high-quality, multi-modal subsurface and 3D information with precise targeting of the region of interest using optional AS&V4 Software.
Precise sample navigation
Tailored to individual application needs thanks to the high flexibility 110 mm stage and in-chamber Thermo Scientific Nav-Cam Camera.
Artifact-free imaging and patterning
With dedicated modes such as DCFI, drift suppression and Thermo Scientific SmartScan Modes.
Optimize your solution
Meet specific application requirements thanks to flexible DualBeam configuration, including optional low-vacuum mode with up to 500 Pa chamber pressure.
Specifications
Style Sheet for Products Table Specifications
Electron beam resolution
  • Optimum WD
    • 0.7 nm at 30 keV STEM
    • 1.4 nm at 1 keV
    • 1.2 nm at 1 keV with beam deceleration
Electron beam parameter space
  • Beam current range: 1 pA to 400 nA
  • Landing energy range: 20* eV – 30 keV
  • Accelerating voltage range: 200 V – 30 kV
  • Maximum horizontal field width: 3.0 mm at 7 mm WD and 7.0 mm at 60 mm WD
  • Extra wide field of view (1×) available through standard navigation montage
Ion optics
  • Acceleration voltage: 500 V – 30 kV
  • Beam current range: 1.5 pA – 65 nA
  • 15-position aperture strip
  • Drift suppression mode as standard for non-conductive samples
  • Minimum source lifetime: 1,000 hours
  • Ion beam resolution: 3.0 nm at 30kV using selective edge method
Detectors
  • Trinity Detection System (in-lens and in-column)
    • T1 segmented lower in-lens detector
    • T2 upper in-lens detector
    • T3 retractable in-column detector (Optional)
    • Up to four simultaneously detected signals
  • Everhart-Thornley SE Detector (ETD)
  • High-performance ion conversion and electron (ICE) detector for secondary ions (SI) and electrons (SE) (Optional)
  • Retractable low-voltage, high-contrast, segmented solid-state backscatter electron detector (DBS) (Optional)
  • Retractable STEM 3+ detector with BF/ DF/ HAADF segments (Optional)
  • IR camera for viewing sample and chamber
  • In-chamber Nav-Cam sample navigation camera (Optional)
  • Integrated beam current measurement
Stage and sample
Flexible 5-axis motorized stage:
  • XY range: 110 mm
  • Z range: 65 mm
  • Rotation: 360° (endless)
  • Tilt range: -15° to +90°
  • XY repeatability: 3 μm
  • Max sample height: Clearance 85 mm to eucentric point
  • Max sample weight at 0° tilt: 5 kg (including sample holder)
  • Max sample size: 110 mm with full rotation (larger sample possible with limited rotation)
  • Compucentric rotation and tilt
* Available as an option, configuration dependent
Resources
Video Player is loading.
Current Time 0:00
Duration 36:03
Loaded: 0.46%
Stream Type LIVE
Remaining Time 36:03
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Introducing the Thermo Scientific Scios 2 FIB-SEM. Learn how this ultra-high resolution analytical system provides outstanding sample preparation and 3D characterization performance for the widest range of samples, including magnetic and nonconductive materials.
    Webinar: Advanced DualBeam automation for every need
    Register for our exclusive webinar to learn how easy it has become to automate daily routine tasks on your DualBeam instrument using our Python-based AutoScript 4 API. Automation can also increase throughput, reproducibility and ease of use, quicken time to data and boost efficiency.
    Register to watch
    Video Player is loading.
    Current Time 0:00
    Duration 36:03
    Loaded: 0.46%
    Stream Type LIVE
    Remaining Time 36:03
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Introducing the Thermo Scientific Scios 2 FIB-SEM. Learn how this ultra-high resolution analytical system provides outstanding sample preparation and 3D characterization performance for the widest range of samples, including magnetic and nonconductive materials.
      Webinar: Advanced DualBeam automation for every need
      Register for our exclusive webinar to learn how easy it has become to automate daily routine tasks on your DualBeam instrument using our Python-based AutoScript 4 API. Automation can also increase throughput, reproducibility and ease of use, quicken time to data and boost efficiency.
      Register to watch
      Applications
      Process control using electron microscopy
      Process control using electron microscopy
      Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
      Quality control and failure analysis using electron microscopy
      Quality control and failure analysis
      Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
      Fundamental Materials Research_R&D_Thumb_274x180_144DPI
      Fundamental Materials Research
      Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
      Semiconductor-RD_1920x1080
      Semiconductor research and development
      Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
      Semi-Metrology_1920x1080
      Semiconductor metrology
      Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
      Semiconductor Failure Analysis
      Semiconductor Failure Analysis
      Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
      Semi-Materials-Character_1920x1080
      Semiconductor materials characterization
      Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
      Techniques
      (S)TEM Sample Preparation
      DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
      Learn more ›
      3D Materials Characterization
      Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
      Learn more ›
      APT Sample Preparation
      Atom probe tomography (APT) provides atomic-resolution 3D compositional analysis of materials. Focused ion beam (FIB) microscopy is an essential technique for high-quality, orientation, and site-specific sample preparation for APT characterization.
      Learn more ›
      Cross-sectioning
      Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
      Learn more ›
      In Situ experimentation
      Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
      Learn more ›
      Multi-scale analysis
      Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
      Learn more ›
      Sample Preparation of Semiconductor Devices
      Thermo Scientific DualBeam systems provide accurate TEM sample preparation for atomic-scale analysis of semiconductor devices. Automation and advanced machine learning technologies produce high-quality samples, at the correct location, and a low cost per sample.
      Learn more ›
      (S)TEM Sample Preparation
      DualBeam microscopes enable the preparation of high-quality, ultra-thin samples for (S)TEM analysis. Thanks to advanced automation, users with any experience level can obtain expert-level results for a wide range of materials.
      Learn more ›
      3D Materials Characterization
      Development of materials often requires multi-scale 3D characterization. DualBeam instruments enable serial sectioning of large volumes and subsequent SEM imaging at nanometer scale, which can be processed into high-quality 3D reconstructions of the sample.
      Learn more ›
      APT Sample Preparation
      Atom probe tomography (APT) provides atomic-resolution 3D compositional analysis of materials. Focused ion beam (FIB) microscopy is an essential technique for high-quality, orientation, and site-specific sample preparation for APT characterization.
      Learn more ›
      Cross-sectioning
      Cross sectioning provides extra insight by revealing sub-surface information. DualBeam instruments feature superior focused ion beam columns for high-quality cross sectioning. With automation, unattended high-throughput processing of samples is possible.
      Learn more ›
      In Situ experimentation
      Direct, real-time observation of microstructural changes with electron microscopy is necessary to understand the underlying principles of dynamic processes such as recrystallization, grain growth, and phase transformation during heating, cooling, and wetting.
      Learn more ›
      Multi-scale analysis
      Novel materials must be analyzed at ever higher resolution while retaining the larger context of the sample. Multi-scale analysis allows for the correlation of various imaging tools and modalities such as X-ray microCT, DualBeam, Laser PFIB, SEM and TEM.
      Learn more ›
      Sample Preparation of Semiconductor Devices
      Thermo Scientific DualBeam systems provide accurate TEM sample preparation for atomic-scale analysis of semiconductor devices. Automation and advanced machine learning technologies produce high-quality samples, at the correct location, and a low cost per sample.
      Learn more ›
      Contact us
      Style Sheet to change Applications H3 to p with em-h3-header class
      Style Sheet to change H3 to p with em-h3-header class
      Style Sheet to change H2 style to p with em-h2-header class
      Style Sheet for Komodo Tabs
      Electron microscopy services
      To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
      Learn more ›
      Style Sheet for Support and Service footer
      Style Sheet for Fonts
      Style Sheet for Cards