Accessories for Luminex instruments and multiplex assays
Thermo Fisher Scientific offers accessory equipment and reagents including sheath fluid, calibration kits and performance verification kits for use with assays run on Luminex systems. We are committed to ensuring that you have all the necessary components you need to run successful Luminex gene expression and protein quantitation assays on Luminex xMAP INTELLIFLEX, FLEXMAP 3D, LX200 or MAGPIX Systems.
Hand-Held Magnetic Plate Washer
The Hand-Held Magnetic Plate Washer is designed for use with the ProcartaPlex and QuantiGene Plex assays configured for magnetic separation. The hand-held magnetic separator enables you to remove the liquid contents of the 96-well plate by simply decanting or flicking the contents into a sink and blotting off the remainder on a paper towel.
Video Player is loading.
Current Time 0:00
Duration 3:02
Loaded: 0%
Stream Type LIVE
Remaining Time 3:02
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Ordering information
    Sheath fluid
    INTELLIFLEX system accessories
    Luminex 200 system accessories
    FLEXMAP 3D system accessories
    MAGPIX system accessories