Featured article
Adoptive cell therapies in immuno-oncology research
Recent advances in immuno-oncology (I-O) research are paving the way for the discovery of novel classes of cancer therapeutics that enhance or enable antitumor immune responses, overcome tumor evasion mechanisms, and promote conditions that favor immune protection. Two principal areas of research include the development of biotherapeutic antibodies that target immune-checkpoint pathways and the expansion of cellular approaches such as adoptive cell therapies (ACT). Thermo Fisher Scientific is developing 3D cell culture techniques and low–cell attachment microplates, along with assays and instruments for analyzing cell function. Here we demonstrate the use of several of these techniques and reagents to study immune cell infiltration in 3D tumor spheroid cultures and the subsequent death, by apoptosis, of spheroid cells.
Read the full article ›
 Download the full issue, BioProbes 79 PDF (3.73 MB)
About the cover
First place winner of the Thermo Fisher Scientific Cell-ebrate Science Imaging Contest in Singapore, Taiwan, and the Southeast Asia region. A human skin cancer section was probed with antibodies for keratin 14 and diaphanous 1, followed by detection with Invitrogen Anti–Mouse IgG Secondary Antibody, Alexa Fluor 488 (for keratin 14) and Invitrogen Anti–Rabbit IgG Secondary Antibody, Alexa Fluor 568 (for diaphanous 1). The section was counterstained with DAPI nucleic acid stain. The image was acquired using an Olympus™ FLUOVIEW™ FV1000 confocal laser-scanning microscope. Image contributed by Mohsin Bin Bashir, A*STAR Institute of Medical Biology (IMB), Singapore.
Featured new products
Just released
Our newest cell and protein analysis products and technologies
Immuno-oncology research
Adoptive cell therapies in immuno-oncology research
Analyze immune cell cytotoxicity in 3D tumor models
Tools for flow cytometry
Best practices for multiparameter flow cytometry
Experimental setup and panel design for accurate results
Detect bacteria in both research and industrial samples
New applications for flow cytometric analysis
Cell function assays
Fluorescent detection of senescence for imaging and flow cytometry applications
Multiplex with the CellEvent Senescence Green Probe for a more complete picture
Monitor drug-induced calcium flux in iPSC-derived cardiomyocytes
Fluo-4 meets the EVOS FL Auto 2 Imaging System and Celleste Image Analysis Software
Visualize microtubule dynamics in live cells
With the highly photostable Tubulin Tracker Deep Red reagent
Assess the cell viability of Staphylococcus aureus biofilms
PrestoBlue HS and alamarBlue HS reagents for microplate viability assays
Antibodies and immunoassays for pathway research
SNAP-ChIP: A robust method for determining histone antibody specificity in ChIP
Barcoded synthetic nucleosomes serve as ChIP internal controls
Hunting for Hippo proteins
With highly specific ABfinity recombinant monoclonal antibodies
Role of metabolic pathways in cancer
Using siRNA knockdowns to verify antibody specificity
Dissect signaling pathways with multiplex western blots
Using fluorescent antibody conjugates in combination with the iBright Imaging System
ELISA workflows just got easier
With Instant ELISA kits, E1-ClipTip electronic pipette, and Wellwash Versa Microplate Washer
Measure secreted protein and mRNA levels with a single instrument platform
ProcartaPlex and ProQuantum immunoassays augment mRNA quantitation
Journal club
Therapeutic targeting of sunitinib-induced androgen receptor (AR) phosphorylation in renal cell carcinoma
Across the spectrum: Supporting spectral flow cytometry applications
Spectral flow cytometry exploits the inherent emission pattern of each fluorescent molecule to generate a unique spectral signature. By relying on the discrimination of unique spectral signatures rather than specific emission channels for detection, spectral flow cytometry enables the use of many fluorescent combinations that were previously difficult or impossible to separate, allowing for increasingly complex multicolor experiments.
The Invitrogen portfolio of flow cytometry antibodies, assays, and reagents addresses the needs of your spectral flow cytometry experiment, whether you are an immunologist investigating the role of regulatory T cells (Tregs), a cell biologist interested in fluorescently labeling RNA for downstream applications, or a microbiologist studying endocytosis, viability, or proliferation.
Learn more about spectral flow cytometry assays and reagents
Behind the Bench blog: Macrophages and more macrophages
Want to learn a few basics about macrophages? Interested in hearing about the newest antibody markers available for studying macrophages? The Invitrogen Behind the Bench blog has four new blog posts that cater to your macrophage research.
Get macrophage basics directly from a Thermo Fisher Scientific R&D scientist in the blog post “Macrophages by flow cytometry: New insights through novel antibody reagents”. Or, for a description of available antibody markers for studying macrophages, read three new blog posts from the R&D scientists who developed them:
Each blog post provides insights into the performance of the antibody markers and their applications in the immunophenotyping of macrophages using flow cytometry.
Read the flow cytometry–tagged subset of the Behind the Bench blog
Watch: How to use the Pierce Chromogenic Endotoxin Quantitation Kit
The video “How to use the Pierce Chromogenic Endotoxin Quantitation Kit” brings to life the simple protocol of the Thermo Scientific Pierce Chromogenic Endotoxin Quant Kit. In just over 10 minutes, you will learn how this kit accurately detects and measures endotoxin (lipopolysaccharide) in a protein, peptide, nucleic acid, or antibody sample using an endpoint amebocyte lysate assay. The Pierce Chromogenic Endotoxin Quant Kit enables endotoxin contamination detection with two linear sensitivity ranges: 0.01–0.1 EU/mL and 0.1–1.0 EU/mL. It is also:
  • Highly specific—no interference from β-glucans and suitable for a wide range of samples
  • Fast—perform assay in as little as 20 minutes
  • Chromogenic—measure this endpoint assay using a standard spectrophotometer or plate reader at 405–410 nm
Watch the video How to use the Pierce Chromogenic Endotoxin Quantitation Kit
Product selection tool for crosslinking applications
Are you looking for ways to improve your protein crosslinking results? Our interactive Crosslinker Selection Tool enables you to quickly and easily find the optimal protein crosslinker based on target functional group, cleavability, water solubility, pegylation, cell membrane permeability, spacer arm length, and packaging. Download your selections to a PDF that includes a list of key references where applicable.
Try the Crosslinker Selection Tool
Product selection tool for biotinylation applications
With so many biotinylation options to choose from, it can be time-consuming and cumbersome to find the right reagent. That’s why we created the interactive Biotinylation Reagent Selection Tool to enable you to quickly and easily find the right biotin or desthiobiotin labeling reagent. This tool allows you to download your selections to a PDF that includes a list of key references where applicable.
Try the Biotinylation Reagent Selection Tool
For Research Use Only. Not for use in diagnostic procedures.