High-Content Screening (HCS) and High-Content Analysis (HCA)
High-throughput for the highest quality and fastest time-to-data possible
Our CellInsight CX7 LZR Pro, CX7 LED Pro, and CX5 high-content imaging platforms are designed for exceptional single-cell analysis capabilities and lightning-fast time-to-data. Perform unbiased spontaneous phenotyping with intact, fixed, or live cells derived from monolayers to spheroids. Acquire complex images and high-content data in seconds with the Thermo Scientific CellInsight HCS platforms powered by HCS Studio software, as featured in over 2,000 peer-reviewed publications.
Short scan times and high contrast with CX7 LED Pro
Capabilities
Professional-quality cell painting with the CellInsight CX7 LZR Pro platform
Cell painting is an advanced technique that can identify thousands of structures in screening conditions, allowing you to gain novel insights into drug discovery and safety applications.
Advanced HCS instruments/platforms designed for a broad range of advanced applications
Explore high-content screening applications ranging from 2D cell-based assays to 3D spheroid biology using our HCS platforms and multiplexable fluorescent reagents.
3D visualization and analyses with Amira software
Amira 3D Software from Thermo Scientific offers 3D analysis capabilities of the most challenging samples, including 3D neuroscience morphology and immuno-oncology colocalization assessment of cancer and immune cells.
Add the Orbitor RS plate handler for automated high-throughput screening
Equip CellInsight HCS Platforms with automated high-throughput screening of up to 80 plates using the Orbitor RS plate handler. Operate the microplate mover directly using HCS Studio software.
“Seek-and-find” functionality
Thermo Scientific EurekaScan Finder on the CellInsight CX7 LED Pro and CX7 LZR Pro HCS platforms is designed to automate the identification and subsequent capture of irregularly seeded samples at progressively higher magnifications.
Quantifiably brilliant data
Our proprietary HCS Studio quantitative analysis software saves time by performing simultaneous imaging with single cell-level quantification and advanced onboard performance statistics, including automated Z’ measurements.