Search Thermo Fisher Scientific
The new Thermo Scientific Dionex Inuvion IC system makes ion analysis simpler and more intuitive than ever before. Designed specifically for ease of use, this IC system provides convenient access to all necessary components, connections, and consumables for streamlined operation and maintenance. It can take advantage of both a wide range of application-specific column chemistries and the flexibility and convenience of electrolytic suppression and automated eluent generation.
The Dionex Inuvion IC system is available in three system configurations: the Dionex Inuvion Core IC system, Dionex Inuvion IC system, and Dionex Inuvion IC system with Reagent-Free IC (RFIC) capabilities. This flexible, adaptable platform is designed to allow you to easily upgrade from the Inuvion Core to the Inuvion (with RFIC) at any time. The system also has a variety of user-installable optional accessories that can be added to address specific application needs.
Experience the next level of IC reliability with a system that can help your lab meet its analysis needs and budget both now and in the future.
Simple, intuitive user experience | Reliable performance |
---|---|
The Dionex Inuvion system allows operators of all skill levels to achieve consistent, excellent results. With automated startup and shutdown routines, it runs samples with minimal user intervention. Additional automated capabilities, such as eluent generation, save time and improve reproducibility by eliminating manual errors. All instrument components are easily accessible for easy maintenance and to quickly swap out accessories. | The Dionex Inuvion IC system is engineered for maximum productivity. State-of-the-art pump technology and system self-diagnostics enable dependable performance. This system helps you expand your application capabilities by offering Reagent-Free IC (RFIC) operation for gradient separations and a broad range of column chemistries, including fast columns for increased speed and quality of results. |
Easily configurable and upgradeable | Comprehensive support for unmatched performance |
The Dionex Inuvion IC system is available in multiple configurations, so you can configure the exact system your lab needs today, with confidence that you can easily grow and adapt as future needs arise. The Inuvion Core configuration can meet your needs today and be upgraded later, as budget allows, to include the automation and RFIC capabilities that the Inuvion configuration provides. | Enhance your Dionex Inuvion IC system with the Unity™ Lab Services IonCare™ service plan*. This comprehensive 5-year plan includes parts, labor, and travel costs, along with preventive maintenance visits in years 2 and 4. Experience worry-free, cost-effective support that ensures your system operates smoothly and efficiently, backed by exceptional service and expertise.
*IonCare service plan is available only in North America and Europe |
High-pressure pump (up to 5000 psi) | ![]() |
![]() |
![]() |
Chemical suppression | ![]() |
![]() |
![]() |
Vacuum pump and degasser | + | ![]() |
![]() |
Electrolytic suppression | + | ![]() |
![]() |
RFIC automatic eluent generation | + | + | ![]() |
Standard + Upgradeable
With multiple options and peripherals to extend IC analysis capabilities, you can easily and cost-effectively adapt to changing sample types and workflow requirements.
Accessory | Benefit |
---|---|
Auxiliary valve (6-port or 10-port) | Automate in-line sample preparation |
Column heater | Minimize the impact of lab temperature fluctuations for more consistent results |
Digital gas pressure regulator | Increase precision of gas delivery for maximum consistency |
IC PEEK Viper fittings | Ensure confident, low dead volume connections |
Integrated regenerant pump | Save benchtop space and allow for easier control of the flow rate |
Seal wash pump | Reduce maintenance costs when using manually prepared eluents |
Eluent monitor (2L or 4L) | Maximize uptime and reduce maintenance costs |
The Dionex Inuvion IC system can be used for a variety of applications including the analysis of environmental, food and beverage, pharmaceutical, and industrial samples. It supports a full range of column chemistries to improve the speed and quality of results for both anion and cation analyses. Additional features such as electrolytic suppression and automated eluent generation allow for more consistent results and expanded application capabilities.
Product information
Application Notes