What are HRP-conjugated secondary antibodies?
Decorative Image
Horseradish peroxidase (HRP) is a commonly used enzyme label with secondary antibodies to amplify the signal of detected targets. HRP catalyzes the oxidation of substrates in the presence of hydrogen peroxide and the result is either a colored precipitate or light. When chromogenic substrates come into contact with the enzyme a colored product precipitates which doesn’t require additional equipment to visualize. When a chemiluminescent substrate comes into contact with HRP light is emitted as a byproduct and must be captured by an imaging instrument.
Horseradish peroxidase (HRP)-conjugated secondary antibodies are commonly used for chemiluminescent, colorimetric, or fluorescent detection in applications, such as western blotting, ELISA, immuno-histochemistry, and others. For each application, HRP-conjugated secondary antibodies need substrates that can deliver high signal-to-noise ratios for the chosen detection method.
HRP-conjugated secondary antibodies by application
Search our high-activity HRP-conjugated secondary antibodies by application.
HRP-conjugated secondary antibodies by popular target species
Our extensive portfolio of HRP-conjugated secondary antibodies offers many options for each target species. Search by popular target species to find the antibody that meets your needs.
Featured HRP-conjugated secondary antibody categories
Decorative image
Invitrogen Superclonal recombinant secondary antibodies represent a technology designed to provide precise and accurate detection of mouse, rabbit, and goat primary antibodies in a variety of applications.
Learn more about Superclonal recombinant secondary antibodies
Decorative image
F(ab’)2 fragments result from digestion of a whole antibody with pepsin. These antibody fragments contain the strong divalent binding found in the variable regions but lack the Fc portion of the antibody.
Search HRP conjugated F(ab')2 secondary antibodies
Complementary products and services
Product photo
Chemiluminescent detection offers exceptional sensitivity and the ability to reprobe western blots.
Product photo
Chromogenic substrates offer simple and cost-effect western blot detection.
Product photo
Invitrogen SuperBoost tyramide signal amplification kits and reagents offer exceptional signal amplification for fluorescent imaging of low-abundance targets. SuperBoost EverRed and EverBlue substrates provide permanent colorimetric staining that is also fluorescent.
Product photo
The iBright Imaging Systems offer up to five imaging modes to support multiple applications.