Speedvac rotors
Thermo Scientific™ Savant™ SpeedVac™ Rotors are available in a wide variety of styles and sizes to accommodate all your sample requirements. They are designed to efficiently handle and accommodate microtubes, bottles, glass and plastic tubes, flasks and even microplates. Individual tube volumes range from 0.4 up to 500 mL. The polypropylene rotors are fully autoclavable.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, microwell plates.
Upgrade to PRO
An alternative to traditional rotors is our PRO rotor system. This is a highly versatile rotor system consists of a four-position rotor along with mix-and-match carriers and blocks for your SPD120, SPD2030, and SPD300 vacuum concentrator systems.
Smart Note: Savant SpeedVac System
When the quality of your results hinges on the quality of synthesized oligos
As oligos are being aggressively leveraged in research and clinical applications such as detecting, characterizing, and developing therapeutics and vaccines, the quality of synthesized oligos is critical. The Savant SpeedVac system enhances speed and reliability so you can have complete confidence in the quality of your oligos as well as the applications they serve.
Download Brochures
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.