Search Thermo Fisher Scientific
Modern battery manufacturing includes a variety of challenges, including limited resources, the need to address an increasing number of tests impacting scrap rate, and ensuring that results are reproducible and reliable across operators and production sites.
Thermo Scientific Avizo Trueput Software for Battery Quality transforms your Thermo Scientific SEM into specialized battery quality analysis solution, helping you to maximize efficiency, maintain quality standards, and achieve operational excellence.
Developed for the production floor, Avizo Trueput Software is a dedicated battery quality inspection solution designed to liberate engineering time.
Transfer the protocol to new operators at different sites
Reduce cost per analysis and use your engineering time for more valuable tasks
One clear answer, and all the data to support it
|
![]() |
![]() |
![]() |
![]() |
![]() |
Particle size distribution | Cracked particles detection |
Particles/binder/ void/cracks analysis |
Precursor primary structure analysis | Post-calendaring layers | |
Identified features |
Particles |
Particles, cracks within particles |
Particles, binder, void, cracks within particles | Primary particles in precursor | Layer of active material |
Sample preparation |
Carbon tape or electrode surface |
Carbon tape or electrode surface |
Electrode surface |
Carbon tape | Electrode surface
|
Reported information |
Particle size distribution compliance, packing factor |
Number of cracked particles, crack size analysis, compliance |
Phase area fraction |
Primary particle size distribution and orientation | Active material layer thickness |
Where to use it |
Materials acceptance, tape manufacturing |
Materials acceptance, tape manufacturing |
After calendaring, after cycling |
Materials acceptance, prior to particles curing | After calendaring |
Benefit |
Optimize cell energy density by achieving target packing factor |
Optimize battery cyclability and reduce operational costs |
Optimize battery cyclability and energy density |
Optimize curing operational costs | Optimize energy density and calendaring operational costs |
Let one of our experts tailor a solution to your needs.
Our team of experts is always available to answer any questions you may have or discuss your project.
For Research Use Only. Not for use in diagnostic procedures.